Assay Details
Gene Symbol: CNNM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017623.4 | NP_060093.3 | 5 - 6 | 1815 | 85 | |
NM_199078.2 | NP_951060.1 | 4 - 5 | 1671 | 85 | ||
XM_011510957.2 | XP_011509259.1 | 5 - 6 | 2175 | 85 | ||
XM_017003800.1 | XP_016859289.1 | 5 - 6 | 2175 | 85 | ||
GenBank mRNA | AF216965.1 | - | 5 - 6 | 914 | 85 | |
AK000025.1 | - | 4 - 5 | 733 | 85 | ||
AK126847.1 | - | 5 - 6 | 1815 | 85 | ||
BC007199.1 | - | 4 - 5 | 827 | 2964706 | 85 | |
BC018604.1 | - | 4 - 5 | 560 | 85 | ||
BC022944.1 | - | 2 - 3 | 259 | 85 | ||
BC037272.1 | - | 5 - 6 | 1824 | 4823510 | 85 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC02000587.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR02009052.hg.1 | TC02000587.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02009052.hg.1 | ENST00000305510.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02009052.hg.1 | ENST00000377060.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02009052.hg.1 | ENST00000424641.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02009052.hg.1 | ENST00000480035.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02009052.hg.1 | NM_017623.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02009052.hg.1 | NM_199078.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016496.hg.1 | ENST00000305510.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016496.hg.1 | ENST00000305510.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016496.hg.1 | ENST00000305510.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016496.hg.1 | ENST00000305510.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016496.hg.1 | ENST00000305510.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016496.hg.1 | ENST00000305510.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016496.hg.1 | ENST00000305510.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016496.hg.1 | ENST00000305510.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016496.hg.1 | ENST00000305510.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016496.hg.1 | ENST00000305510.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016496.hg.1 | ENST00000305510.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016496.hg.1 | ENST00000305510.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016496.hg.1 | ENST00000305510.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016496.hg.1 | ENST00000377060.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016496.hg.1 | ENST00000377060.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016496.hg.1 | ENST00000377060.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016496.hg.1 | ENST00000377060.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016496.hg.1 | ENST00000377060.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016496.hg.1 | ENST00000377060.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016496.hg.1 | ENST00000377060.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016496.hg.1 | ENST00000377060.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016496.hg.1 | ENST00000377060.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016496.hg.1 | ENST00000377060.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016496.hg.1 | ENST00000377060.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016496.hg.1 | ENST00000465224.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016496.hg.1 | ENST00000465224.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016496.hg.1 | ENST00000465224.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016496.hg.1 | ENST00000465224.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016496.hg.1 | ENST00000465224.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016496.hg.1 | ENST00000465224.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016496.hg.1 | ENST00000465224.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016496.hg.1 | ENST00000465224.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016496.hg.1 | ENST00000465224.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016496.hg.1 | ENST00000465224.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016496.hg.1 | ENST00000465224.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016496.hg.1 | ENST00000465224.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016496.hg.1 | ENST00000465224.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016496.hg.1 | ENST00000480035.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016496.hg.1 | ENST00000480035.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016496.hg.1 | ENST00000480035.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016496.hg.1 | ENST00000494595.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016496.hg.1 | ENST00000494595.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016496.hg.1 | ENST00000494595.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016496.hg.1 | NM_017623.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016496.hg.1 | NM_017623.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016496.hg.1 | NM_017623.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016496.hg.1 | NM_017623.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016496.hg.1 | NM_017623.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016496.hg.1 | NM_017623.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016496.hg.1 | NM_017623.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016496.hg.1 | NM_017623.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016496.hg.1 | NM_017623.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016496.hg.1 | NM_017623.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016496.hg.1 | NM_017623.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016496.hg.1 | NM_017623.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016496.hg.1 | NM_017623.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016496.hg.1 | NM_017623.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016496.hg.1 | NM_017623.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016496.hg.1 | NM_199078.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016496.hg.1 | NM_199078.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016496.hg.1 | NM_199078.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016496.hg.1 | NM_199078.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016496.hg.1 | NM_199078.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016496.hg.1 | NM_199078.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016496.hg.1 | NM_199078.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016496.hg.1 | NM_199078.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016496.hg.1 | NM_199078.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016496.hg.1 | NM_199078.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016496.hg.1 | NM_199078.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016496.hg.1 | NM_199078.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016496.hg.1 | NM_199078.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016496.hg.1 | ENST00000305510.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016496.hg.1 | ENST00000305510.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016496.hg.1 | ENST00000305510.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016496.hg.1 | ENST00000305510.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016496.hg.1 | ENST00000305510.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016496.hg.1 | ENST00000305510.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016496.hg.1 | ENST00000305510.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016496.hg.1 | ENST00000305510.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016496.hg.1 | ENST00000305510.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016496.hg.1 | ENST00000305510.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016496.hg.1 | ENST00000305510.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016496.hg.1 | ENST00000305510.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016496.hg.1 | ENST00000305510.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016496.hg.1 | ENST00000377060.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016496.hg.1 | ENST00000377060.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016496.hg.1 | ENST00000377060.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016496.hg.1 | ENST00000377060.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016496.hg.1 | ENST00000377060.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016496.hg.1 | ENST00000377060.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016496.hg.1 | ENST00000377060.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016496.hg.1 | ENST00000377060.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016496.hg.1 | ENST00000377060.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016496.hg.1 | ENST00000377060.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016496.hg.1 | ENST00000377060.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016496.hg.1 | ENST00000465224.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016496.hg.1 | ENST00000465224.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016496.hg.1 | ENST00000465224.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016496.hg.1 | ENST00000465224.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016496.hg.1 | ENST00000465224.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016496.hg.1 | ENST00000465224.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016496.hg.1 | ENST00000465224.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016496.hg.1 | ENST00000465224.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016496.hg.1 | ENST00000465224.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016496.hg.1 | ENST00000465224.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016496.hg.1 | ENST00000465224.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016496.hg.1 | ENST00000465224.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016496.hg.1 | ENST00000465224.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016496.hg.1 | ENST00000480035.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016496.hg.1 | ENST00000480035.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016496.hg.1 | ENST00000480035.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016496.hg.1 | ENST00000494595.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016496.hg.1 | ENST00000494595.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016496.hg.1 | ENST00000494595.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016496.hg.1 | NM_017623.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016496.hg.1 | NM_017623.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016496.hg.1 | NM_017623.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016496.hg.1 | NM_017623.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016496.hg.1 | NM_017623.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016496.hg.1 | NM_017623.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016496.hg.1 | NM_017623.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016496.hg.1 | NM_017623.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016496.hg.1 | NM_017623.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016496.hg.1 | NM_017623.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016496.hg.1 | NM_017623.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016496.hg.1 | NM_017623.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016496.hg.1 | NM_017623.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016496.hg.1 | NM_017623.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016496.hg.1 | NM_017623.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016496.hg.1 | NM_199078.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016496.hg.1 | NM_199078.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016496.hg.1 | NM_199078.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016496.hg.1 | NM_199078.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016496.hg.1 | NM_199078.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016496.hg.1 | NM_199078.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016496.hg.1 | NM_199078.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016496.hg.1 | NM_199078.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016496.hg.1 | NM_199078.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016496.hg.1 | NM_199078.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016496.hg.1 | NM_199078.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016496.hg.1 | NM_199078.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016496.hg.1 | NM_199078.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200222715.hg.1 | ENST00000305510.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200222715.hg.1 | ENST00000377060.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200222715.hg.1 | ENST00000465224.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200222715.hg.1 | ENST00000480035.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200222715.hg.1 | NM_017623.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200222715.hg.1 | NM_199078.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200222715.hg.1 | ENST00000305510.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200222715.hg.1 | ENST00000377060.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200222715.hg.1 | ENST00000465224.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200222715.hg.1 | ENST00000480035.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200222715.hg.1 | NM_017623.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200222715.hg.1 | NM_199078.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200222715.hg.1 | TC0200016496.hg.1 | Clariom D TC |
