Assay Details
| Gene Symbol: GET4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015949.2 | NP_057033.2 | 2 - 3 | 324 | 84 | |
| GenBank mRNA | AF132954.1 | - | 2 - 3 | 211 | 84 | |
| AK023560.1 | - | 2 - 3 | 248 | 84 | ||
| AK024305.1 | - | 1 - 2 | 1025 | 84 | ||
| AK097899.1 | - | 1 - 2 | 1985 | 84 | ||
| AK125863.1 | - | 1 - 2 | 2590 | 84 | ||
| AL133014.1 | - | 1 - 2 | 1114 | 84 | ||
| BC003550.1 | - | 2 - 3 | 266 | 3546669 | 84 | |
| BM843540.1 | - | 2 - 3 | 324 | 84 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07003316.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR07000186.hg.1 | TC07003316.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07000186.hg.1 | ENST00000265857.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07000186.hg.1 | NM_015949.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07000193.hg.1 | TC07003316.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07000193.hg.1 | ENST00000265857.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07000193.hg.1 | ENST00000407192.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07000193.hg.1 | NM_015949.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013324.hg.1 | ENST00000265857.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013324.hg.1 | ENST00000265857.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013324.hg.1 | ENST00000265857.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013324.hg.1 | ENST00000265857.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013324.hg.1 | ENST00000265857.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013324.hg.1 | ENST00000265857.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013324.hg.1 | ENST00000265857.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013324.hg.1 | ENST00000265857.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013324.hg.1 | ENST00000265857.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013324.hg.1 | ENST00000265857.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013324.hg.1 | ENST00000265857.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013324.hg.1 | ENST00000265857.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013324.hg.1 | ENST00000265857.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013324.hg.1 | ENST00000265857.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013324.hg.1 | ENST00000265857.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013324.hg.1 | ENST00000407192.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013324.hg.1 | ENST00000407192.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013324.hg.1 | ENST00000407192.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013324.hg.1 | ENST00000407192.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013324.hg.1 | ENST00000407192.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013324.hg.1 | ENST00000407192.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013324.hg.1 | ENST00000407192.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013324.hg.1 | ENST00000407192.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013324.hg.1 | ENST00000407192.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013324.hg.1 | ENST00000407192.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013324.hg.1 | ENST00000407192.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013324.hg.1 | ENST00000407192.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013324.hg.1 | ENST00000407192.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013324.hg.1 | ENST00000407192.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013324.hg.1 | ENST00000407192.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013324.hg.1 | ENST00000412734.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013324.hg.1 | ENST00000412734.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013324.hg.1 | ENST00000412734.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013324.hg.1 | ENST00000412734.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013324.hg.1 | ENST00000412734.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013324.hg.1 | ENST00000412734.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013324.hg.1 | ENST00000426056.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013324.hg.1 | ENST00000426056.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013324.hg.1 | ENST00000426056.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013324.hg.1 | ENST00000426056.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013324.hg.1 | ENST00000441491.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013324.hg.1 | ENST00000441491.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013324.hg.1 | ENST00000441491.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013324.hg.1 | ENST00000441491.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013324.hg.1 | ENST00000441491.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013324.hg.1 | ENST00000464468.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013324.hg.1 | ENST00000464468.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013324.hg.1 | ENST00000464468.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013324.hg.1 | ENST00000464468.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013324.hg.1 | ENST00000464468.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013324.hg.1 | ENST00000464468.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013324.hg.1 | ENST00000483469.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013324.hg.1 | ENST00000483469.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013324.hg.1 | ENST00000483469.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013324.hg.1 | ENST00000483469.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013324.hg.1 | NM_015949.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013324.hg.1 | NM_015949.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013324.hg.1 | NM_015949.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013324.hg.1 | NM_015949.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013324.hg.1 | NM_015949.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013324.hg.1 | NM_015949.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013324.hg.1 | NM_015949.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013324.hg.1 | NM_015949.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013324.hg.1 | NM_015949.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013324.hg.1 | NM_015949.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013324.hg.1 | NM_015949.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013324.hg.1 | NM_015949.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013324.hg.1 | NM_015949.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013324.hg.1 | NM_015949.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013324.hg.1 | NM_015949.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012410.hg.1 | ENST00000509201.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012410.hg.1 | ENST00000509201.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012410.hg.1 | ENST00000509201.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012410.hg.1 | ENST00000509201.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07000187.hg.1 | TC07003316.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07000187.hg.1 | ENST00000265857.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07000187.hg.1 | NM_015949.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07000192.hg.1 | TC07003316.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07000192.hg.1 | ENST00000265857.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07000192.hg.1 | ENST00000407192.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07000192.hg.1 | NM_015949.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700196124.hg.1 | ENST00000265857.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196124.hg.1 | NM_015949.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700196124.hg.1 | ENST00000265857.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196124.hg.1 | NM_015949.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700196124.hg.1 | TC0700013324.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700196125.hg.1 | ENST00000265857.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196125.hg.1 | NM_015949.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700196125.hg.1 | ENST00000265857.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196125.hg.1 | NM_015949.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700196125.hg.1 | TC0700013324.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700196130.hg.1 | ENST00000265857.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196130.hg.1 | ENST00000407192.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196130.hg.1 | ENST00000412734.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196130.hg.1 | ENST00000441491.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196130.hg.1 | NM_015949.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700196130.hg.1 | ENST00000265857.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196130.hg.1 | ENST00000407192.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196130.hg.1 | ENST00000412734.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196130.hg.1 | ENST00000441491.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196130.hg.1 | NM_015949.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700196130.hg.1 | TC0700013324.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700196131.hg.1 | ENST00000265857.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196131.hg.1 | ENST00000407192.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196131.hg.1 | ENST00000412734.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196131.hg.1 | ENST00000441491.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196131.hg.1 | NM_015949.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700196131.hg.1 | ENST00000265857.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196131.hg.1 | ENST00000407192.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196131.hg.1 | ENST00000412734.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196131.hg.1 | ENST00000441491.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196131.hg.1 | NM_015949.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700196131.hg.1 | TC0700013324.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300012410.hg.1 | ENST00000509201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012410.hg.1 | ENST00000509201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012410.hg.1 | ENST00000509201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012410.hg.1 | ENST00000509201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196123.hg.1 | ENST00000265857.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196123.hg.1 | NM_015949.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700196123.hg.1 | ENST00000265857.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196123.hg.1 | NM_015949.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700196123.hg.1 | TC0700013324.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0700013324.hg.1 | ENST00000265857.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013324.hg.1 | ENST00000265857.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013324.hg.1 | ENST00000265857.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013324.hg.1 | ENST00000265857.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013324.hg.1 | ENST00000265857.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013324.hg.1 | ENST00000265857.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013324.hg.1 | ENST00000265857.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013324.hg.1 | ENST00000265857.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013324.hg.1 | ENST00000265857.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013324.hg.1 | ENST00000265857.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013324.hg.1 | ENST00000265857.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013324.hg.1 | ENST00000265857.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013324.hg.1 | ENST00000265857.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013324.hg.1 | ENST00000265857.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013324.hg.1 | ENST00000265857.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013324.hg.1 | ENST00000407192.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013324.hg.1 | ENST00000407192.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013324.hg.1 | ENST00000407192.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013324.hg.1 | ENST00000407192.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013324.hg.1 | ENST00000407192.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013324.hg.1 | ENST00000407192.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013324.hg.1 | ENST00000407192.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013324.hg.1 | ENST00000407192.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013324.hg.1 | ENST00000407192.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013324.hg.1 | ENST00000407192.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013324.hg.1 | ENST00000407192.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013324.hg.1 | ENST00000407192.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013324.hg.1 | ENST00000407192.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013324.hg.1 | ENST00000407192.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013324.hg.1 | ENST00000407192.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013324.hg.1 | ENST00000412734.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013324.hg.1 | ENST00000412734.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013324.hg.1 | ENST00000412734.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013324.hg.1 | ENST00000412734.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013324.hg.1 | ENST00000412734.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013324.hg.1 | ENST00000412734.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013324.hg.1 | ENST00000426056.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013324.hg.1 | ENST00000426056.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013324.hg.1 | ENST00000426056.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013324.hg.1 | ENST00000426056.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013324.hg.1 | ENST00000441491.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013324.hg.1 | ENST00000441491.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013324.hg.1 | ENST00000441491.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013324.hg.1 | ENST00000441491.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013324.hg.1 | ENST00000441491.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013324.hg.1 | ENST00000464468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013324.hg.1 | ENST00000464468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013324.hg.1 | ENST00000464468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013324.hg.1 | ENST00000464468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013324.hg.1 | ENST00000464468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013324.hg.1 | ENST00000464468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013324.hg.1 | ENST00000483469.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013324.hg.1 | ENST00000483469.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013324.hg.1 | ENST00000483469.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013324.hg.1 | ENST00000483469.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013324.hg.1 | NM_015949.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013324.hg.1 | NM_015949.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013324.hg.1 | NM_015949.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013324.hg.1 | NM_015949.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013324.hg.1 | NM_015949.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013324.hg.1 | NM_015949.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013324.hg.1 | NM_015949.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013324.hg.1 | NM_015949.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013324.hg.1 | NM_015949.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013324.hg.1 | NM_015949.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013324.hg.1 | NM_015949.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013324.hg.1 | NM_015949.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013324.hg.1 | NM_015949.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013324.hg.1 | NM_015949.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013324.hg.1 | NM_015949.2 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 156662 | - | - |

GET4
Genomic Map