Assay Details
Gene Symbol: TAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000353.2 | NP_000344.1 | 1 - 2 | 93 | 78 | |
GenBank mRNA | AK313380.1 | - | 1 - 2 | 94 | 78 | |
BC020707.1 | - | 1 - 2 | 90 | 78 | ||
BC130534.1 | - | 1 - 2 | 64 | 40146565 | 78 | |
X52520.1 | - | 1 - 2 | 90 | 78 | ||
X55675.1 | - | 1 - 2 | 44 | 78 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC16001249.hg
Additional Information:
This assay is in the 5'UTR region of the AK313380.1,BC020707.1,BC130534.1,NM_000353.2,X52520.1,X55675.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
2-oxoglutarate metabolic process
glutamate metabolic process
L-phenylalanine catabolic process
tyrosine catabolic process
response to oxidative stress
biosynthetic process
response to mercury ion
response to glucocorticoid
L-tyrosine:2-oxoglutarate aminotransferase activity
amino acid binding
pyridoxal phosphate binding
L-phenylalanine:2-oxoglutarate aminotransferase activity
2-oxoglutarate metabolic process
glutamate metabolic process
L-phenylalanine catabolic process
tyrosine catabolic process
response to oxidative stress
biosynthetic process
response to mercury ion
response to glucocorticoid
L-tyrosine:2-oxoglutarate aminotransferase activity
amino acid binding
pyridoxal phosphate binding
L-phenylalanine:2-oxoglutarate aminotransferase activity
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR16017352.hg.1 | ENST00000355962.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR16017352.hg.1 | NM_000353.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16017352.hg.1 | TC16001249.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR16017354.hg.1 | ENST00000355962.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR16017354.hg.1 | NM_000353.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16017354.hg.1 | TC16001249.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | ENST00000355962.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | ENST00000355962.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | ENST00000355962.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | ENST00000355962.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | ENST00000355962.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | ENST00000355962.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | ENST00000355962.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | ENST00000355962.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | ENST00000355962.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | ENST00000355962.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | ENST00000355962.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | ENST00000355962.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | ENST00000355962.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | ENST00000355962.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | ENST00000355962.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | ENST00000355962.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | ENST00000355962.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | ENST00000355962.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | ENST00000564007.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | ENST00000564007.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | ENST00000564007.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | ENST00000564007.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | ENST00000564007.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | ENST00000566010.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | ENST00000566010.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | ENST00000566010.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | ENST00000566010.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | ENST00000566010.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | ENST00000566010.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | ENST00000566094.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | ENST00000566094.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | ENST00000566094.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | ENST00000566094.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | ENST00000566094.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | ENST00000566094.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | NM_000353.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | NM_000353.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | NM_000353.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | NM_000353.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | NM_000353.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | NM_000353.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | NM_000353.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | NM_000353.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | NM_000353.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | NM_000353.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | NM_000353.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | NM_000353.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | NM_000353.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | NM_000353.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | NM_000353.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | NM_000353.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16017353.hg.1 | ENST00000355962.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR16017353.hg.1 | NM_000353.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16017353.hg.1 | TC16001249.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1600183646.hg.1 | ENST00000355962.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600183646.hg.1 | ENST00000566010.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600183646.hg.1 | ENST00000566094.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600183646.hg.1 | NM_000353.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600183646.hg.1 | ENST00000355962.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600183646.hg.1 | ENST00000566010.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600183646.hg.1 | ENST00000566094.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600183646.hg.1 | NM_000353.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600183646.hg.1 | TC1600010789.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1600183645.hg.1 | ENST00000355962.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600183645.hg.1 | ENST00000566010.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600183645.hg.1 | ENST00000566094.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600183645.hg.1 | NM_000353.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600183645.hg.1 | ENST00000355962.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600183645.hg.1 | ENST00000566010.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600183645.hg.1 | ENST00000566094.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600183645.hg.1 | NM_000353.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600183645.hg.1 | TC1600010789.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1600183643.hg.1 | ENST00000355962.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600183643.hg.1 | ENST00000566010.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600183643.hg.1 | ENST00000566094.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600183643.hg.1 | NM_000353.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600183643.hg.1 | ENST00000355962.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600183643.hg.1 | ENST00000566010.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600183643.hg.1 | ENST00000566094.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600183643.hg.1 | NM_000353.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600183643.hg.1 | TC1600010789.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | ENST00000355962.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | ENST00000355962.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | ENST00000355962.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | ENST00000355962.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | ENST00000355962.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | ENST00000355962.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | ENST00000355962.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | ENST00000355962.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | ENST00000355962.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | ENST00000355962.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | ENST00000355962.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | ENST00000355962.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | ENST00000355962.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | ENST00000355962.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | ENST00000355962.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | ENST00000355962.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | ENST00000355962.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | ENST00000355962.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | ENST00000564007.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | ENST00000564007.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | ENST00000564007.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | ENST00000564007.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | ENST00000564007.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | ENST00000566010.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | ENST00000566010.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | ENST00000566010.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | ENST00000566010.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | ENST00000566010.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | ENST00000566010.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | ENST00000566094.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | ENST00000566094.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | ENST00000566094.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | ENST00000566094.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | ENST00000566094.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | ENST00000566094.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | NM_000353.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | NM_000353.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | NM_000353.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | NM_000353.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | NM_000353.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | NM_000353.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | NM_000353.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | NM_000353.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | NM_000353.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | NM_000353.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | NM_000353.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | NM_000353.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | NM_000353.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | NM_000353.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | NM_000353.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | NM_000353.2 | Refseq |
