Assay Details
Gene Symbol: TAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000353.2 | NP_000344.1 | 7 - 8 | 860 | 73 | |
GenBank mRNA | AK313380.1 | - | 7 - 8 | 861 | 73 | |
BC130534.1 | - | 7 - 8 | 831 | 40146565 | 73 | |
X52520.1 | - | 7 - 8 | 857 | 73 | ||
X55675.1 | - | 7 - 8 | 811 | 73 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC16001249.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
2-oxoglutarate metabolic process
glutamate metabolic process
L-phenylalanine catabolic process
tyrosine catabolic process
response to oxidative stress
biosynthetic process
response to mercury ion
response to glucocorticoid
L-tyrosine:2-oxoglutarate aminotransferase activity
amino acid binding
pyridoxal phosphate binding
L-phenylalanine:2-oxoglutarate aminotransferase activity
2-oxoglutarate metabolic process
glutamate metabolic process
L-phenylalanine catabolic process
tyrosine catabolic process
response to oxidative stress
biosynthetic process
response to mercury ion
response to glucocorticoid
L-tyrosine:2-oxoglutarate aminotransferase activity
amino acid binding
pyridoxal phosphate binding
L-phenylalanine:2-oxoglutarate aminotransferase activity
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR16017343.hg.1 | ENST00000355962.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR16017343.hg.1 | NM_000353.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16017343.hg.1 | TC16001249.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | ENST00000355962.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | ENST00000355962.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | ENST00000355962.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | ENST00000355962.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | ENST00000355962.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | ENST00000355962.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | ENST00000355962.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | ENST00000355962.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | ENST00000355962.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | ENST00000355962.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | ENST00000355962.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | ENST00000355962.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | ENST00000355962.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | ENST00000355962.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | ENST00000355962.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | ENST00000355962.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | ENST00000355962.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | ENST00000355962.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | ENST00000564007.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | ENST00000564007.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | ENST00000564007.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | ENST00000564007.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | ENST00000564007.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | ENST00000566010.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | ENST00000566010.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | ENST00000566010.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | ENST00000566010.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | ENST00000566010.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | ENST00000566010.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | ENST00000566094.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | ENST00000566094.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | ENST00000566094.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | ENST00000566094.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | ENST00000566094.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | ENST00000566094.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | NM_000353.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | NM_000353.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | NM_000353.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | NM_000353.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | NM_000353.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | NM_000353.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | NM_000353.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | NM_000353.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | NM_000353.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | NM_000353.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | NM_000353.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | NM_000353.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | NM_000353.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | NM_000353.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | NM_000353.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010789.hg.1 | NM_000353.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600183634.hg.1 | ENST00000355962.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600183634.hg.1 | NM_000353.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600183634.hg.1 | ENST00000355962.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600183634.hg.1 | NM_000353.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600183634.hg.1 | TC1600010789.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | ENST00000355962.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | ENST00000355962.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | ENST00000355962.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | ENST00000355962.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | ENST00000355962.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | ENST00000355962.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | ENST00000355962.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | ENST00000355962.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | ENST00000355962.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | ENST00000355962.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | ENST00000355962.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | ENST00000355962.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | ENST00000355962.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | ENST00000355962.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | ENST00000355962.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | ENST00000355962.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | ENST00000355962.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | ENST00000355962.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | ENST00000564007.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | ENST00000564007.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | ENST00000564007.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | ENST00000564007.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | ENST00000564007.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | ENST00000566010.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | ENST00000566010.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | ENST00000566010.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | ENST00000566010.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | ENST00000566010.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | ENST00000566010.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | ENST00000566094.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | ENST00000566094.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | ENST00000566094.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | ENST00000566094.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | ENST00000566094.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | ENST00000566094.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | NM_000353.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | NM_000353.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | NM_000353.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | NM_000353.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | NM_000353.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | NM_000353.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | NM_000353.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | NM_000353.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | NM_000353.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | NM_000353.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | NM_000353.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | NM_000353.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | NM_000353.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | NM_000353.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | NM_000353.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010789.hg.1 | NM_000353.2 | Refseq |
