Assay Details
Gene Symbol: SFMBT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016329.3 | NP_057413.2 | 12 - 13 | 1755 | 88 | |
XM_005265221.3 | XP_005265278.1 | 12 - 13 | 1871 | 88 | ||
XM_006713203.3 | XP_006713266.1 | 13 - 14 | 1993 | 88 | ||
XM_006713204.3 | XP_006713267.1 | 12 - 13 | 1693 | 88 | ||
XM_011533824.2 | XP_011532126.1 | 13 - 14 | 1869 | 88 | ||
XM_011533825.2 | XP_011532127.1 | 13 - 14 | 1716 | 88 | ||
GenBank mRNA | AB189472.1 | - | 11 - 12 | 1379 | 88 | |
AF168132.1 | - | 12 - 13 | 1692 | 88 | ||
AK313965.1 | - | 12 - 13 | 1750 | 88 | ||
AL080140.1 | - | 9 - 10 | 1033 | 88 | ||
BC014614.2 | - | 12 - 13 | 1639 | 3029598 | 88 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC03001464.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000470575.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000470575.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000470575.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000470575.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000470575.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000470575.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000474837.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000474837.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000474837.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000474837.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000474837.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000482396.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000482396.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000482396.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000482396.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000482396.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000482396.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000482396.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000482396.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000482396.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000482396.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000483069.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000483069.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000483069.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000483069.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000483069.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000483069.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000483069.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000485494.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000485494.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000485494.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000492146.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000492146.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000492146.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000492146.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000497586.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000497586.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000497586.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000497586.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000497586.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000497586.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000497586.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000497586.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03026653.hg.1 | TC03001464.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR03026653.hg.1 | ENST00000296295.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03026653.hg.1 | ENST00000358080.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03026653.hg.1 | ENST00000394750.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03026653.hg.1 | ENST00000394752.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03026653.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000470575.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000470575.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000470575.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000470575.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000470575.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000470575.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000474837.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000474837.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000474837.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000474837.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000474837.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000482396.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000482396.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000482396.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000482396.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000482396.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000482396.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000482396.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000482396.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000482396.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000482396.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000483069.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000483069.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000483069.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000483069.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000483069.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000483069.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000483069.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000485494.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000485494.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000485494.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000492146.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000492146.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000492146.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000492146.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000497586.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000497586.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000497586.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000497586.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000497586.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000497586.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000497586.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000497586.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300208085.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300208085.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300208085.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300208085.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300208085.hg.1 | TC0300013998.hg.1 | Clariom D TC |
