Assay Details
Gene Symbol: SFMBT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016329.3 | NP_057413.2 | 16 - 17 | 2114 | 68 | |
XM_005265221.3 | XP_005265278.1 | 16 - 17 | 2230 | 68 | ||
XM_006713203.3 | XP_006713266.1 | 17 - 18 | 2352 | 68 | ||
XM_006713204.3 | XP_006713267.1 | 16 - 17 | 2052 | 68 | ||
XM_011533824.2 | XP_011532126.1 | 17 - 18 | 2228 | 68 | ||
XM_011533825.2 | XP_011532127.1 | 17 - 18 | 2075 | 68 | ||
GenBank mRNA | AB189472.1 | - | 15 - 16 | 1738 | 68 | |
AF168132.1 | - | 16 - 17 | 2051 | 68 | ||
AK313965.1 | - | 16 - 17 | 2109 | 68 | ||
AL080140.1 | - | 13 - 14 | 1392 | 68 | ||
BC014614.2 | - | 16 - 17 | 1998 | 3029598 | 68 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC03001464.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR03026649.hg.1 | TC03001464.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR03026649.hg.1 | ENST00000296295.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03026649.hg.1 | ENST00000358080.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03026649.hg.1 | ENST00000394750.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03026649.hg.1 | ENST00000394752.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03026649.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000470575.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000470575.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000470575.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000470575.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000470575.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000470575.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000474837.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000474837.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000474837.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000474837.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000474837.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000482396.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000482396.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000482396.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000482396.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000482396.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000482396.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000482396.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000482396.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000482396.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000482396.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000483069.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000483069.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000483069.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000483069.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000483069.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000483069.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000483069.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000485494.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000485494.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000485494.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000492146.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000492146.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000492146.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000492146.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000497586.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000497586.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000497586.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000497586.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000497586.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000497586.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000497586.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000497586.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000470575.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000470575.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000470575.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000470575.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000470575.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000470575.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000474837.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000474837.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000474837.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000474837.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000474837.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000482396.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000482396.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000482396.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000482396.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000482396.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000482396.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000482396.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000482396.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000482396.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000482396.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000483069.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000483069.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000483069.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000483069.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000483069.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000483069.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000483069.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000485494.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000485494.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000485494.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000492146.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000492146.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000492146.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000492146.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000497586.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000497586.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000497586.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000497586.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000497586.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000497586.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000497586.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000497586.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300208078.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300208078.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300208078.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300208078.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300208078.hg.1 | TC0300013998.hg.1 | Clariom D TC |
