Assay Details
Gene Symbol: SFMBT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016329.3 | NP_057413.2 | 18 - 19 | 2469 | 79 | |
XM_005265221.3 | XP_005265278.1 | 18 - 19 | 2585 | 79 | ||
XM_006713203.3 | XP_006713266.1 | 19 - 20 | 2707 | 79 | ||
XM_006713204.3 | XP_006713267.1 | 18 - 19 | 2407 | 79 | ||
XM_011533824.2 | XP_011532126.1 | 19 - 20 | 2583 | 79 | ||
XM_011533825.2 | XP_011532127.1 | 19 - 20 | 2430 | 79 | ||
GenBank mRNA | AB189472.1 | - | 17 - 18 | 2093 | 79 | |
AF168132.1 | - | 18 - 19 | 2406 | 79 | ||
AK313965.1 | - | 18 - 19 | 2464 | 79 | ||
AL080140.1 | - | 15 - 16 | 1747 | 79 | ||
BC014614.2 | - | 18 - 19 | 2353 | 3029598 | 79 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC03001464.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000470575.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000470575.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000470575.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000470575.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000470575.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000470575.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000474837.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000474837.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000474837.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000474837.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000474837.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000482396.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000482396.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000482396.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000482396.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000482396.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000482396.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000482396.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000482396.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000482396.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000482396.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000483069.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000483069.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000483069.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000483069.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000483069.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000483069.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000483069.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000485494.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000485494.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000485494.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000492146.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000492146.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000492146.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000492146.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000497586.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000497586.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000497586.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000497586.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000497586.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000497586.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000497586.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000497586.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03026647.hg.1 | TC03001464.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR03026647.hg.1 | ENST00000296295.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03026647.hg.1 | ENST00000358080.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03026647.hg.1 | ENST00000394750.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03026647.hg.1 | ENST00000394752.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03026647.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300208076.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300208076.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300208076.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300208076.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300208076.hg.1 | TC0300013998.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000470575.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000470575.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000470575.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000470575.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000470575.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000470575.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000474837.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000474837.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000474837.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000474837.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000474837.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000482396.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000482396.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000482396.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000482396.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000482396.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000482396.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000482396.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000482396.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000482396.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000482396.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000483069.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000483069.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000483069.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000483069.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000483069.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000483069.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000483069.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000485494.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000485494.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000485494.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000492146.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000492146.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000492146.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000492146.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000497586.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000497586.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000497586.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000497586.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000497586.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000497586.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000497586.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000497586.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
