Assay Details
| Gene Symbol: CYBRD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256909.1 | NP_001243838.1 | 2 - 3 | 247 | 76 | |
| NM_024843.3 | NP_079119.3 | 2 - 3 | 585 | 76 | ||
| GenBank mRNA | AK027115.1 | - | 2 - 3 | 427 | 76 | |
| AK225459.1 | - | 2 - 3 | 427 | 76 | ||
| AK291731.1 | - | 2 - 3 | 435 | 76 | ||
| AK301485.1 | - | 2 - 3 | 247 | 76 | ||
| AK316588.1 | - | 2 - 3 | 464 | 76 | ||
| AL136693.1 | - | 2 - 3 | 468 | 76 | ||
| BC065290.1 | - | 2 - 3 | 420 | 6063145 | 76 | |
| CX866825.1 | - | 2 - 3 | 476 | 76 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02001021.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200009955.hg.1 | ENST00000321348.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009955.hg.1 | ENST00000321348.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009955.hg.1 | ENST00000321348.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009955.hg.1 | ENST00000321348.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009955.hg.1 | ENST00000321348.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009955.hg.1 | ENST00000321348.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009955.hg.1 | ENST00000321348.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009955.hg.1 | ENST00000321348.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009955.hg.1 | ENST00000321348.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009955.hg.1 | ENST00000321348.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009955.hg.1 | ENST00000321348.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009955.hg.1 | ENST00000321348.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009955.hg.1 | ENST00000375252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009955.hg.1 | ENST00000375252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009955.hg.1 | ENST00000375252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009955.hg.1 | ENST00000375252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009955.hg.1 | ENST00000375252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009955.hg.1 | ENST00000375252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009955.hg.1 | ENST00000375252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009955.hg.1 | ENST00000375252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009955.hg.1 | ENST00000375252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009955.hg.1 | ENST00000409484.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009955.hg.1 | ENST00000409484.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009955.hg.1 | ENST00000409484.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009955.hg.1 | ENST00000409484.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009955.hg.1 | ENST00000409484.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009955.hg.1 | ENST00000409484.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009955.hg.1 | ENST00000409484.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009955.hg.1 | ENST00000409484.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009955.hg.1 | ENST00000445146.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009955.hg.1 | ENST00000445146.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009955.hg.1 | ENST00000445146.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009955.hg.1 | ENST00000445146.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009955.hg.1 | ENST00000468308.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009955.hg.1 | ENST00000468308.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009955.hg.1 | ENST00000468308.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009955.hg.1 | ENST00000468308.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009955.hg.1 | ENST00000468308.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009955.hg.1 | ENST00000474182.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009955.hg.1 | ENST00000474182.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009955.hg.1 | ENST00000474182.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009955.hg.1 | ENST00000494587.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009955.hg.1 | ENST00000494587.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009955.hg.1 | NM_001127383.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009955.hg.1 | NM_001127383.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009955.hg.1 | NM_001127383.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009955.hg.1 | NM_001127383.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009955.hg.1 | NM_001127383.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009955.hg.1 | NM_001127383.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009955.hg.1 | NM_001127383.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009955.hg.1 | NM_001127383.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009955.hg.1 | NM_001127383.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009955.hg.1 | NM_001127383.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009955.hg.1 | NM_001127383.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009955.hg.1 | NM_001256909.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009955.hg.1 | NM_001256909.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009955.hg.1 | NM_001256909.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009955.hg.1 | NM_001256909.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009955.hg.1 | NM_001256909.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009955.hg.1 | NM_001256909.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009955.hg.1 | NM_001256909.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009955.hg.1 | NM_001256909.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009955.hg.1 | NM_001256909.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009955.hg.1 | NM_001256909.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009955.hg.1 | NM_024843.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009955.hg.1 | NM_024843.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009955.hg.1 | NM_024843.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009955.hg.1 | NM_024843.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009955.hg.1 | NM_024843.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009955.hg.1 | NM_024843.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009955.hg.1 | NM_024843.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009955.hg.1 | NM_024843.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009955.hg.1 | NM_024843.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009955.hg.1 | NM_024843.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009955.hg.1 | NM_024843.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009955.hg.1 | NM_024843.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02015117.hg.1 | TC02001021.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02015117.hg.1 | ENST00000321348.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02015117.hg.1 | ENST00000375252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02015117.hg.1 | ENST00000409484.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02015117.hg.1 | NM_001127383.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02015117.hg.1 | NM_024843.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200169766.hg.1 | ENST00000409484.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200169766.hg.1 | NM_001256909.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200169766.hg.1 | ENST00000409484.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200169766.hg.1 | NM_001256909.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200169766.hg.1 | TC0200009955.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200169781.hg.1 | ENST00000321348.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200169781.hg.1 | ENST00000375252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200169781.hg.1 | ENST00000409484.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200169781.hg.1 | ENST00000445146.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200169781.hg.1 | NM_001127383.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200169781.hg.1 | NM_001256909.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200169781.hg.1 | NM_024843.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200169781.hg.1 | ENST00000321348.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200169781.hg.1 | ENST00000375252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200169781.hg.1 | ENST00000409484.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200169781.hg.1 | ENST00000445146.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200169781.hg.1 | NM_001127383.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200169781.hg.1 | NM_001256909.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200169781.hg.1 | NM_024843.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200169781.hg.1 | TC0200009955.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200009955.hg.1 | ENST00000321348.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009955.hg.1 | ENST00000321348.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009955.hg.1 | ENST00000321348.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009955.hg.1 | ENST00000321348.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009955.hg.1 | ENST00000321348.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009955.hg.1 | ENST00000321348.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009955.hg.1 | ENST00000321348.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009955.hg.1 | ENST00000321348.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009955.hg.1 | ENST00000321348.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009955.hg.1 | ENST00000321348.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009955.hg.1 | ENST00000321348.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009955.hg.1 | ENST00000321348.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009955.hg.1 | ENST00000375252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009955.hg.1 | ENST00000375252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009955.hg.1 | ENST00000375252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009955.hg.1 | ENST00000375252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009955.hg.1 | ENST00000375252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009955.hg.1 | ENST00000375252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009955.hg.1 | ENST00000375252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009955.hg.1 | ENST00000375252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009955.hg.1 | ENST00000375252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009955.hg.1 | ENST00000409484.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009955.hg.1 | ENST00000409484.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009955.hg.1 | ENST00000409484.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009955.hg.1 | ENST00000409484.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009955.hg.1 | ENST00000409484.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009955.hg.1 | ENST00000409484.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009955.hg.1 | ENST00000409484.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009955.hg.1 | ENST00000409484.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009955.hg.1 | ENST00000445146.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009955.hg.1 | ENST00000445146.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009955.hg.1 | ENST00000445146.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009955.hg.1 | ENST00000445146.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009955.hg.1 | ENST00000468308.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009955.hg.1 | ENST00000468308.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009955.hg.1 | ENST00000468308.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009955.hg.1 | ENST00000468308.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009955.hg.1 | ENST00000468308.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009955.hg.1 | ENST00000474182.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009955.hg.1 | ENST00000474182.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009955.hg.1 | ENST00000474182.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009955.hg.1 | ENST00000494587.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009955.hg.1 | ENST00000494587.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009955.hg.1 | NM_001127383.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009955.hg.1 | NM_001127383.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009955.hg.1 | NM_001127383.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009955.hg.1 | NM_001127383.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009955.hg.1 | NM_001127383.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009955.hg.1 | NM_001127383.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009955.hg.1 | NM_001127383.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009955.hg.1 | NM_001127383.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009955.hg.1 | NM_001127383.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009955.hg.1 | NM_001127383.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009955.hg.1 | NM_001127383.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009955.hg.1 | NM_001256909.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009955.hg.1 | NM_001256909.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009955.hg.1 | NM_001256909.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009955.hg.1 | NM_001256909.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009955.hg.1 | NM_001256909.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009955.hg.1 | NM_001256909.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009955.hg.1 | NM_001256909.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009955.hg.1 | NM_001256909.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009955.hg.1 | NM_001256909.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009955.hg.1 | NM_001256909.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009955.hg.1 | NM_024843.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009955.hg.1 | NM_024843.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009955.hg.1 | NM_024843.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009955.hg.1 | NM_024843.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009955.hg.1 | NM_024843.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009955.hg.1 | NM_024843.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009955.hg.1 | NM_024843.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009955.hg.1 | NM_024843.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009955.hg.1 | NM_024843.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009955.hg.1 | NM_024843.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009955.hg.1 | NM_024843.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009955.hg.1 | NM_024843.3 | Refseq |

CYBRD1
Genomic Map