Assay Details
Gene Symbol: ST8SIA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017016175.1 | XP_016871664.1 | 1 | 1023 | 90 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017016175.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
