Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016280.hg.1 |
TC08001264.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016280.hg.1 |
ENST00000356457.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016280.hg.1 |
ENST00000379454.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016280.hg.1 |
ENST00000389213.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016280.hg.1 |
ENST00000445642.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016280.hg.1 |
ENST00000517847.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016280.hg.1 |
ENST00000517903.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016280.hg.1 |
ENST00000518068.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016280.hg.1 |
ENST00000519234.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016280.hg.1 |
ENST00000522835.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016280.hg.1 |
ENST00000522919.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016280.hg.1 |
ENST00000523897.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016280.hg.1 |
ENST00000541428.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016280.hg.1 |
NM_001164750.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016280.hg.1 |
NM_001164751.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016280.hg.1 |
NM_001164752.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016280.hg.1 |
NM_001164753.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016280.hg.1 |
NM_001164754.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016280.hg.1 |
NM_001164755.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016280.hg.1 |
NM_004318.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016280.hg.1 |
NM_032466.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016280.hg.1 |
NM_032468.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016279.hg.1 |
TC08001264.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016279.hg.1 |
ENST00000356457.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016279.hg.1 |
ENST00000379454.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016279.hg.1 |
ENST00000389213.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016279.hg.1 |
ENST00000445642.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016279.hg.1 |
ENST00000517847.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016279.hg.1 |
ENST00000517903.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016279.hg.1 |
ENST00000518068.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016279.hg.1 |
ENST00000519234.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016279.hg.1 |
ENST00000522835.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016279.hg.1 |
ENST00000522919.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016279.hg.1 |
ENST00000523897.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016279.hg.1 |
ENST00000541428.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016279.hg.1 |
NM_001164750.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016279.hg.1 |
NM_001164751.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016279.hg.1 |
NM_001164752.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016279.hg.1 |
NM_001164753.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016279.hg.1 |
NM_001164754.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016279.hg.1 |
NM_001164755.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016279.hg.1 |
NM_004318.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016279.hg.1 |
NM_032466.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016279.hg.1 |
NM_032468.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016281.hg.1 |
TC08001264.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016281.hg.1 |
ENST00000356457.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016281.hg.1 |
ENST00000379454.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016281.hg.1 |
ENST00000389213.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016281.hg.1 |
ENST00000445642.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016281.hg.1 |
ENST00000517847.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016281.hg.1 |
ENST00000517903.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016281.hg.1 |
ENST00000518068.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016281.hg.1 |
ENST00000519234.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016281.hg.1 |
ENST00000522835.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016281.hg.1 |
ENST00000522919.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016281.hg.1 |
ENST00000523897.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016281.hg.1 |
ENST00000541428.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016281.hg.1 |
NM_001164750.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016281.hg.1 |
NM_001164751.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016281.hg.1 |
NM_001164752.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016281.hg.1 |
NM_001164753.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016281.hg.1 |
NM_001164754.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016281.hg.1 |
NM_001164755.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016281.hg.1 |
NM_004318.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016281.hg.1 |
NM_032466.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016281.hg.1 |
NM_032468.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016282.hg.1 |
TC08001264.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016282.hg.1 |
ENST00000356457.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016282.hg.1 |
ENST00000379454.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016282.hg.1 |
ENST00000389213.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016282.hg.1 |
ENST00000445642.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016282.hg.1 |
ENST00000517847.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016282.hg.1 |
ENST00000517903.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016282.hg.1 |
ENST00000518068.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016282.hg.1 |
ENST00000519234.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016282.hg.1 |
ENST00000522835.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016282.hg.1 |
ENST00000522919.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016282.hg.1 |
ENST00000523897.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016282.hg.1 |
ENST00000541428.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016282.hg.1 |
NM_001164750.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016282.hg.1 |
NM_001164751.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016282.hg.1 |
NM_001164752.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016282.hg.1 |
NM_001164753.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016282.hg.1 |
NM_001164754.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016282.hg.1 |
NM_001164755.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016282.hg.1 |
NM_004318.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016282.hg.1 |
NM_032466.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016282.hg.1 |
NM_032468.4 |
Refseq |