Assay Details
Gene Symbol: UAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001324113.1 | NP_001311042.1 | 6 - 7 | 1113 | 67 | |
NM_001324114.1 | NP_001311043.1 | 5 - 6 | 1001 | 67 | ||
NM_001324115.1 | NP_001311044.1 | 6 - 7 | 1340 | 67 | ||
NM_001324116.1 | NP_001311045.1 | 5 - 6 | 1228 | 67 | ||
NM_001324117.1 | NP_001311046.1 | 5 - 6 | 1228 | 67 | ||
NM_003115.4 | NP_003106.3 | 5 - 6 | 1162 | 67 | ||
XM_011509913.2 | XP_011508215.1 | 6 - 7 | 1061 | 67 | ||
XM_011509914.2 | XP_011508216.1 | 6 - 7 | 1113 | 67 | ||
GenBank mRNA | AB011004.1 | - | 4 - 5 | 833 | 67 | |
AK312685.1 | - | 5 - 6 | 1088 | 67 | ||
BC009377.2 | - | 5 - 6 | 1135 | 4121439 | 67 | |
S73498.1 | - | 5 - 6 | 1065 | 67 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01001422.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367924.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367924.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367924.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367924.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367924.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367924.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367924.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367924.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367924.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367924.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367924.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367924.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367924.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367924.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367925.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367925.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367925.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367925.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367925.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367925.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367925.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367925.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367925.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367925.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367925.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367925.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367925.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367925.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000474728.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000474728.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000474728.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000476240.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000476240.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000486089.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000486089.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000486089.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000486089.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | NM_003115.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | NM_003115.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | NM_003115.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | NM_003115.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | NM_003115.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | NM_003115.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | NM_003115.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | NM_003115.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | NM_003115.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | NM_003115.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | NM_003115.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | NM_003115.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | NM_003115.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | NM_003115.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | NM_003115.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | NM_003115.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | NM_003115.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01022220.hg.1 | TC01001422.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01022220.hg.1 | ENST00000271469.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01022220.hg.1 | ENST00000367924.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01022220.hg.1 | ENST00000367925.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01022220.hg.1 | ENST00000367926.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01022220.hg.1 | NM_003115.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01022219.hg.1 | TC01001422.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01022219.hg.1 | ENST00000271469.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01022219.hg.1 | ENST00000367924.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01022219.hg.1 | ENST00000367925.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01022219.hg.1 | ENST00000367926.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01022219.hg.1 | NM_003115.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100176498.hg.1 | ENST00000271469.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100176498.hg.1 | ENST00000367924.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100176498.hg.1 | ENST00000367925.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100176498.hg.1 | ENST00000367926.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100176498.hg.1 | ENST00000474728.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100176498.hg.1 | NM_003115.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100176498.hg.1 | ENST00000271469.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100176498.hg.1 | ENST00000367924.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100176498.hg.1 | ENST00000367925.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100176498.hg.1 | ENST00000367926.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100176498.hg.1 | ENST00000474728.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100176498.hg.1 | NM_003115.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100176498.hg.1 | TC0100010381.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367924.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367924.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367924.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367924.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367924.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367924.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367924.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367924.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367924.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367924.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367924.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367924.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367924.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367924.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367925.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367925.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367925.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367925.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367925.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367925.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367925.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367925.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367925.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367925.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367925.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367925.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367925.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367925.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000474728.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000474728.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000474728.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000476240.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000476240.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000486089.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000486089.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000486089.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000486089.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | NM_003115.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | NM_003115.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | NM_003115.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | NM_003115.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | NM_003115.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | NM_003115.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | NM_003115.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | NM_003115.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | NM_003115.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | NM_003115.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | NM_003115.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | NM_003115.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | NM_003115.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | NM_003115.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | NM_003115.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | NM_003115.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | NM_003115.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100176499.hg.1 | ENST00000271469.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100176499.hg.1 | ENST00000367924.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100176499.hg.1 | ENST00000367925.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100176499.hg.1 | ENST00000367926.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100176499.hg.1 | ENST00000474728.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100176499.hg.1 | NM_003115.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100176499.hg.1 | ENST00000271469.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100176499.hg.1 | ENST00000367924.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100176499.hg.1 | ENST00000367925.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100176499.hg.1 | ENST00000367926.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100176499.hg.1 | ENST00000474728.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100176499.hg.1 | NM_003115.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100176499.hg.1 | TC0100010381.hg.1 | Clariom D TC |
