Assay Details
| Gene Symbol: UAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001324113.1 | NP_001311042.1 | 7 - 8 | 1301 | 106 | |
| NM_001324114.1 | NP_001311043.1 | 6 - 7 | 1189 | 106 | ||
| NM_001324115.1 | NP_001311044.1 | 7 - 8 | 1528 | 106 | ||
| NM_001324116.1 | NP_001311045.1 | 6 - 7 | 1416 | 106 | ||
| NM_001324117.1 | NP_001311046.1 | 6 - 7 | 1416 | 106 | ||
| NM_003115.4 | NP_003106.3 | 6 - 7 | 1350 | 106 | ||
| XM_011509913.2 | XP_011508215.1 | 7 - 8 | 1249 | 106 | ||
| XM_011509914.2 | XP_011508216.1 | 7 - 8 | 1301 | 106 | ||
| GenBank mRNA | AB011004.1 | - | 5 - 6 | 1021 | 106 | |
| AK312685.1 | - | 6 - 7 | 1276 | 106 | ||
| BC009377.2 | - | 6 - 7 | 1323 | 4121439 | 106 | |
| S73498.1 | - | 6 - 7 | 1253 | 106 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01001422.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01022222.hg.1 | TC01001422.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022222.hg.1 | ENST00000271469.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022222.hg.1 | ENST00000367924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022222.hg.1 | ENST00000367925.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022222.hg.1 | ENST00000367926.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022222.hg.1 | NM_003115.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000474728.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000474728.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000474728.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000476240.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000476240.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000486089.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000486089.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000486089.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000486089.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | NM_003115.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | NM_003115.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | NM_003115.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | NM_003115.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | NM_003115.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | NM_003115.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | NM_003115.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | NM_003115.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | NM_003115.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | NM_003115.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | NM_003115.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | NM_003115.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | NM_003115.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | NM_003115.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | NM_003115.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | NM_003115.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | NM_003115.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022223.hg.1 | TC01001422.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022223.hg.1 | ENST00000271469.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022223.hg.1 | ENST00000367924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022223.hg.1 | ENST00000367925.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022223.hg.1 | ENST00000367926.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022223.hg.1 | NM_003115.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000474728.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000474728.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000474728.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000476240.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000476240.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000486089.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000486089.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000486089.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000486089.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | NM_003115.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | NM_003115.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | NM_003115.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | NM_003115.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | NM_003115.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | NM_003115.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | NM_003115.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | NM_003115.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | NM_003115.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | NM_003115.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | NM_003115.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | NM_003115.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | NM_003115.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | NM_003115.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | NM_003115.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | NM_003115.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | NM_003115.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100176501.hg.1 | ENST00000271469.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176501.hg.1 | ENST00000367924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176501.hg.1 | ENST00000367925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176501.hg.1 | ENST00000367926.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176501.hg.1 | NM_003115.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100176501.hg.1 | ENST00000271469.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176501.hg.1 | ENST00000367924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176501.hg.1 | ENST00000367925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176501.hg.1 | ENST00000367926.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176501.hg.1 | NM_003115.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100176501.hg.1 | TC0100010381.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100176502.hg.1 | ENST00000271469.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176502.hg.1 | ENST00000367924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176502.hg.1 | ENST00000367925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176502.hg.1 | ENST00000367926.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176502.hg.1 | NM_003115.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100176502.hg.1 | ENST00000271469.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176502.hg.1 | ENST00000367924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176502.hg.1 | ENST00000367925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176502.hg.1 | ENST00000367926.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176502.hg.1 | NM_003115.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100176502.hg.1 | TC0100010381.hg.1 | Clariom D TC |

UAP1
Genomic Map