Assay Details
| Gene Symbol: UAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001324116.1 | NP_001311045.1 | 9 - 10 | 1810 | 102 | |
| NM_001324117.1 | NP_001311046.1 | 9 - 10 | 1807 | 102 | ||
| XM_011509913.2 | XP_011508215.1 | 10 - 11 | 1643 | 102 | ||
| XM_011509914.2 | XP_011508216.1 | 10 - 11 | 1695 | 102 | ||
More Information
Set Membership: |
|

UAP1
Genomic Map