Assay Details
Gene Symbol: DNMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130823.2 | NP_001124295.1 | 14 - 15 | 1278 | 83 | |
NM_001318730.1 | NP_001305659.1 | 13 - 14 | 1149 | 83 | ||
NM_001318731.1 | NP_001305660.1 | 14 - 15 | 1157 | 83 | ||
NM_001379.3 | NP_001370.1 | 13 - 14 | 1230 | 83 | ||
GenBank mRNA | AB209413.2 | - | 13 - 14 | 956 | 83 | |
AF180682.1 | - | 11 - 12 | 737 | 83 | ||
BC092517.1 | - | 14 - 15 | 1037 | 3077831 | 83 | |
BC126227.1 | - | 14 - 15 | 1222 | 8991943 | 83 | |
BC144093.1 | - | 14 - 15 | 1222 | 9052613 | 83 | |
X63692.1 | - | 13 - 14 | 1231 | 83 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC19001159.hg.1
Additional Information:
This assay is in the 5'UTR region of the AF180682.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
