Assay Details
Gene Symbol: LGALS14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203471.1 | NP_982297.1 | 2 - 3 | 536 | 64 | |
GenBank mRNA | BG434228.1 | - | 2 - 3 | 362 | 64 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC19000555.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR19007564.hg.1 | ENST00000360675.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19007564.hg.1 | ENST00000392052.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19007564.hg.1 | NM_020129.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19007564.hg.1 | NM_203471.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19007564.hg.1 | TC19000555.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1900008078.hg.1 | ENST00000360675.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008078.hg.1 | ENST00000360675.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008078.hg.1 | ENST00000360675.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008078.hg.1 | ENST00000360675.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008078.hg.1 | ENST00000360675.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008078.hg.1 | ENST00000360675.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008078.hg.1 | ENST00000392052.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008078.hg.1 | ENST00000392052.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008078.hg.1 | ENST00000392052.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008078.hg.1 | ENST00000392052.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008078.hg.1 | ENST00000392052.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008078.hg.1 | ENST00000392052.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008078.hg.1 | ENST00000392052.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008078.hg.1 | ENST00000392052.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008078.hg.1 | ENST00000601802.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008078.hg.1 | ENST00000601802.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008078.hg.1 | ENST00000601802.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008078.hg.1 | ENST00000601802.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008078.hg.1 | NM_020129.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008078.hg.1 | NM_020129.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008078.hg.1 | NM_020129.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008078.hg.1 | NM_020129.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008078.hg.1 | NM_020129.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008078.hg.1 | NM_020129.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008078.hg.1 | NM_020129.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008078.hg.1 | NM_020129.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008078.hg.1 | NM_203471.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008078.hg.1 | NM_203471.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008078.hg.1 | NM_203471.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008078.hg.1 | NM_203471.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008078.hg.1 | NM_203471.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008078.hg.1 | NM_203471.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008078.hg.1 | NM_203471.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008078.hg.1 | NM_203471.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008078.hg.1 | NM_203471.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008078.hg.1 | NM_203471.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19007565.hg.1 | ENST00000360675.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19007565.hg.1 | NM_203471.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19007565.hg.1 | TC19000555.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR19007563.hg.1 | ENST00000360675.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19007563.hg.1 | ENST00000392052.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19007563.hg.1 | NM_020129.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19007563.hg.1 | NM_203471.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19007563.hg.1 | TC19000555.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR19007566.hg.1 | ENST00000360675.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19007566.hg.1 | ENST00000392052.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19007566.hg.1 | NM_020129.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19007566.hg.1 | NM_203471.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19007566.hg.1 | TC19000555.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1900160937.hg.1 | ENST00000360675.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900160937.hg.1 | ENST00000392052.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900160937.hg.1 | NM_020129.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900160937.hg.1 | NM_203471.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900160937.hg.1 | TC1900008078.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900160937.hg.1 | ENST00000360675.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900160937.hg.1 | ENST00000392052.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900160937.hg.1 | NM_020129.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900160937.hg.1 | NM_203471.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900160930.hg.1 | ENST00000392052.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900160930.hg.1 | NM_020129.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900160930.hg.1 | NM_203471.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900160930.hg.1 | TC1900008078.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900160930.hg.1 | ENST00000392052.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900160930.hg.1 | NM_020129.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900160930.hg.1 | NM_203471.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900160931.hg.1 | ENST00000392052.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900160931.hg.1 | NM_020129.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900160931.hg.1 | NM_203471.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900160931.hg.1 | TC1900008078.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900160931.hg.1 | ENST00000392052.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900160931.hg.1 | NM_020129.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900160931.hg.1 | NM_203471.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900160935.hg.1 | ENST00000360675.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900160935.hg.1 | NM_203471.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900160935.hg.1 | TC1900008078.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900160935.hg.1 | ENST00000360675.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900160935.hg.1 | NM_203471.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008078.hg.1 | ENST00000360675.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008078.hg.1 | ENST00000360675.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008078.hg.1 | ENST00000360675.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008078.hg.1 | ENST00000360675.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008078.hg.1 | ENST00000360675.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008078.hg.1 | ENST00000360675.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008078.hg.1 | ENST00000392052.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008078.hg.1 | ENST00000392052.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008078.hg.1 | ENST00000392052.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008078.hg.1 | ENST00000392052.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008078.hg.1 | ENST00000392052.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008078.hg.1 | ENST00000392052.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008078.hg.1 | ENST00000392052.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008078.hg.1 | ENST00000392052.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008078.hg.1 | ENST00000601802.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008078.hg.1 | ENST00000601802.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008078.hg.1 | ENST00000601802.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008078.hg.1 | ENST00000601802.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008078.hg.1 | NM_020129.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008078.hg.1 | NM_020129.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008078.hg.1 | NM_020129.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008078.hg.1 | NM_020129.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008078.hg.1 | NM_020129.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008078.hg.1 | NM_020129.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008078.hg.1 | NM_020129.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008078.hg.1 | NM_020129.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008078.hg.1 | NM_203471.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008078.hg.1 | NM_203471.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008078.hg.1 | NM_203471.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008078.hg.1 | NM_203471.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008078.hg.1 | NM_203471.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008078.hg.1 | NM_203471.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008078.hg.1 | NM_203471.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008078.hg.1 | NM_203471.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008078.hg.1 | NM_203471.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008078.hg.1 | NM_203471.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900160936.hg.1 | ENST00000360675.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900160936.hg.1 | NM_203471.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900160936.hg.1 | TC1900008078.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900160936.hg.1 | ENST00000360675.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900160936.hg.1 | NM_203471.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900160933.hg.1 | ENST00000360675.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900160933.hg.1 | ENST00000392052.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900160933.hg.1 | ENST00000601802.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900160933.hg.1 | NM_020129.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900160933.hg.1 | NM_203471.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900160933.hg.1 | TC1900008078.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900160933.hg.1 | ENST00000360675.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900160933.hg.1 | ENST00000392052.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900160933.hg.1 | ENST00000601802.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900160933.hg.1 | NM_020129.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900160933.hg.1 | NM_203471.1 | Refseq |
