Assay Details
| Gene Symbol: EPX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000502.5 | NP_000493.1 | 9 - 10 | 1662 | 54 | |
| GenBank mRNA | X14346.1 | - | 9 - 10 | 1503 | 54 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17000711.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
defense response to nematode
response to oxidative stress
negative regulation of interleukin-10 production
negative regulation of interleukin-5 production
positive regulation of interleukin-4 production
hydrogen peroxide catabolic process
oxidation-reduction process
eosinophil migration
cellular oxidant detoxification
peroxidase activity
heme binding
metal ion binding
defense response to nematode
response to oxidative stress
negative regulation of interleukin-10 production
negative regulation of interleukin-5 production
positive regulation of interleukin-4 production
hydrogen peroxide catabolic process
oxidation-reduction process
eosinophil migration
cellular oxidant detoxification
peroxidase activity
heme binding
metal ion binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR17009263.hg.1 | ENST00000225371.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17009263.hg.1 | NM_000502.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17009263.hg.1 | TC17000711.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17009262.hg.1 | ENST00000225371.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17009262.hg.1 | NM_000502.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17009262.hg.1 | TC17000711.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17009264.hg.1 | ENST00000225371.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17009264.hg.1 | NM_000502.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17009264.hg.1 | TC17000711.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17009265.hg.1 | ENST00000225371.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17009265.hg.1 | NM_000502.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17009265.hg.1 | TC17000711.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008424.hg.1 | ENST00000225371.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008424.hg.1 | ENST00000225371.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008424.hg.1 | ENST00000225371.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008424.hg.1 | ENST00000225371.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008424.hg.1 | ENST00000225371.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008424.hg.1 | ENST00000225371.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008424.hg.1 | ENST00000225371.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008424.hg.1 | ENST00000225371.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008424.hg.1 | ENST00000225371.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008424.hg.1 | ENST00000225371.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008424.hg.1 | ENST00000225371.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008424.hg.1 | ENST00000225371.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008424.hg.1 | ENST00000225371.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008424.hg.1 | ENST00000225371.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008424.hg.1 | NM_000502.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008424.hg.1 | NM_000502.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008424.hg.1 | NM_000502.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008424.hg.1 | NM_000502.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008424.hg.1 | NM_000502.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008424.hg.1 | NM_000502.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008424.hg.1 | NM_000502.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008424.hg.1 | NM_000502.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008424.hg.1 | NM_000502.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008424.hg.1 | NM_000502.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008424.hg.1 | NM_000502.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008424.hg.1 | NM_000502.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008424.hg.1 | NM_000502.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008424.hg.1 | NM_000502.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700162648.hg.1 | ENST00000225371.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700162648.hg.1 | NM_000502.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700162648.hg.1 | ENST00000225371.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700162648.hg.1 | NM_000502.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700162648.hg.1 | TC1700008424.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1700008424.hg.1 | ENST00000225371.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008424.hg.1 | ENST00000225371.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008424.hg.1 | ENST00000225371.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008424.hg.1 | ENST00000225371.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008424.hg.1 | ENST00000225371.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008424.hg.1 | ENST00000225371.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008424.hg.1 | ENST00000225371.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008424.hg.1 | ENST00000225371.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008424.hg.1 | ENST00000225371.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008424.hg.1 | ENST00000225371.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008424.hg.1 | ENST00000225371.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008424.hg.1 | ENST00000225371.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008424.hg.1 | ENST00000225371.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008424.hg.1 | ENST00000225371.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008424.hg.1 | NM_000502.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008424.hg.1 | NM_000502.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008424.hg.1 | NM_000502.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008424.hg.1 | NM_000502.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008424.hg.1 | NM_000502.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008424.hg.1 | NM_000502.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008424.hg.1 | NM_000502.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008424.hg.1 | NM_000502.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008424.hg.1 | NM_000502.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008424.hg.1 | NM_000502.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008424.hg.1 | NM_000502.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008424.hg.1 | NM_000502.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008424.hg.1 | NM_000502.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008424.hg.1 | NM_000502.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700162646.hg.1 | ENST00000225371.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700162646.hg.1 | NM_000502.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700162646.hg.1 | ENST00000225371.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700162646.hg.1 | NM_000502.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700162646.hg.1 | TC1700008424.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700162647.hg.1 | ENST00000225371.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700162647.hg.1 | NM_000502.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700162647.hg.1 | ENST00000225371.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700162647.hg.1 | NM_000502.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700162647.hg.1 | TC1700008424.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700162645.hg.1 | ENST00000225371.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700162645.hg.1 | NM_000502.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700162645.hg.1 | ENST00000225371.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700162645.hg.1 | NM_000502.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700162645.hg.1 | TC1700008424.hg.1 | Clariom D TC |

EPX
Genomic Map