Assay Details
| Gene Symbol: ECM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001197295.1 | NP_001184224.1 | 2 - 3 | 475 | 128 | |
| NM_001197296.1 | NP_001184225.1 | 2 - 3 | 475 | 128 | ||
| NM_001393.3 | NP_001384.1 | 2 - 3 | 475 | 128 | ||
| XM_017014376.1 | XP_016869865.1 | 1 - 2 | 315 | 128 | ||
| XM_017014377.1 | XP_016869866.1 | 1 - 2 | 315 | 128 | ||
| GenBank mRNA | AB011792.1 | - | 2 - 3 | 361 | 128 | |
| AK296458.1 | - | 2 - 3 | 128 | |||
| AK298278.1 | - | 3 - 4 | 387 | 128 | ||
| AK303922.1 | - | 2 - 3 | 376 | 128 | ||
| AK312820.1 | - | 2 - 3 | 373 | 128 | ||
| BC036806.2 | - | 1 - 2 | 316 | 128 | ||
| BC105958.1 | - | 1 - 2 | 40035034 | 128 | ||
| BC107493.1 | - | 1 - 2 | 318 | 40035036 | 128 | |
| BX537976.1 | - | 2 - 3 | 438 | 128 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09001337.hg
Additional Information:
This assay is in the 5'UTR region of the AK298278.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0900010800.hg.1 | ENST00000344604.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010800.hg.1 | ENST00000344604.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010800.hg.1 | ENST00000344604.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010800.hg.1 | ENST00000344604.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010800.hg.1 | ENST00000344604.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010800.hg.1 | ENST00000344604.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010800.hg.1 | ENST00000344604.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010800.hg.1 | ENST00000344604.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010800.hg.1 | ENST00000344604.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010800.hg.1 | ENST00000344604.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010800.hg.1 | ENST00000344604.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010800.hg.1 | ENST00000344604.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010800.hg.1 | ENST00000344604.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010800.hg.1 | ENST00000344604.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010800.hg.1 | ENST00000344604.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010800.hg.1 | ENST00000375540.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010800.hg.1 | ENST00000375540.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010800.hg.1 | ENST00000375540.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010800.hg.1 | ENST00000375540.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010800.hg.1 | ENST00000375540.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010800.hg.1 | ENST00000375540.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010800.hg.1 | ENST00000395534.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010800.hg.1 | ENST00000395534.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010800.hg.1 | ENST00000395534.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010800.hg.1 | ENST00000395534.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010800.hg.1 | ENST00000395534.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010800.hg.1 | ENST00000444490.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010800.hg.1 | ENST00000444490.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010800.hg.1 | ENST00000444490.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010800.hg.1 | ENST00000444490.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010800.hg.1 | ENST00000444490.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010800.hg.1 | ENST00000444490.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010800.hg.1 | ENST00000444490.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010800.hg.1 | ENST00000444490.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010800.hg.1 | ENST00000444490.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010800.hg.1 | ENST00000444490.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010800.hg.1 | ENST00000444490.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010800.hg.1 | ENST00000444490.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010800.hg.1 | ENST00000444490.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010800.hg.1 | ENST00000444490.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010800.hg.1 | NM_001197295.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010800.hg.1 | NM_001197295.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010800.hg.1 | NM_001197295.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010800.hg.1 | NM_001197295.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010800.hg.1 | NM_001197295.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010800.hg.1 | NM_001197295.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010800.hg.1 | NM_001197295.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010800.hg.1 | NM_001197295.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010800.hg.1 | NM_001197295.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010800.hg.1 | NM_001197295.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010800.hg.1 | NM_001197295.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010800.hg.1 | NM_001197295.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010800.hg.1 | NM_001197295.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010800.hg.1 | NM_001197295.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010800.hg.1 | NM_001197295.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010800.hg.1 | NM_001197296.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010800.hg.1 | NM_001197296.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010800.hg.1 | NM_001197296.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010800.hg.1 | NM_001197296.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010800.hg.1 | NM_001197296.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010800.hg.1 | NM_001197296.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010800.hg.1 | NM_001197296.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010800.hg.1 | NM_001197296.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010800.hg.1 | NM_001197296.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010800.hg.1 | NM_001197296.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010800.hg.1 | NM_001197296.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010800.hg.1 | NM_001197296.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010800.hg.1 | NM_001197296.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010800.hg.1 | NM_001197296.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010800.hg.1 | NM_001393.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010800.hg.1 | NM_001393.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010800.hg.1 | NM_001393.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010800.hg.1 | NM_001393.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010800.hg.1 | NM_001393.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010800.hg.1 | NM_001393.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010800.hg.1 | NM_001393.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010800.hg.1 | NM_001393.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010800.hg.1 | NM_001393.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010800.hg.1 | NM_001393.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010800.hg.1 | NM_001393.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010800.hg.1 | NM_001393.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010800.hg.1 | NM_001393.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010800.hg.1 | NM_001393.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010800.hg.1 | NM_001393.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010800.hg.1 | NM_001393.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09016777.hg.1 | TC09001337.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09016777.hg.1 | ENST00000344604.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09016777.hg.1 | ENST00000375540.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09016777.hg.1 | ENST00000444490.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09016777.hg.1 | NM_001197295.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09016777.hg.1 | NM_001197296.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09016777.hg.1 | NM_001393.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900173540.hg.1 | ENST00000344604.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900173540.hg.1 | ENST00000375540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900173540.hg.1 | ENST00000395534.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900173540.hg.1 | ENST00000444490.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900173540.hg.1 | NM_001197295.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900173540.hg.1 | NM_001197296.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900173540.hg.1 | NM_001393.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900173540.hg.1 | ENST00000344604.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900173540.hg.1 | ENST00000375540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900173540.hg.1 | ENST00000395534.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900173540.hg.1 | ENST00000444490.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900173540.hg.1 | NM_001197295.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900173540.hg.1 | NM_001197296.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900173540.hg.1 | NM_001393.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900173540.hg.1 | TC0900010800.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0900010800.hg.1 | ENST00000344604.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010800.hg.1 | ENST00000344604.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010800.hg.1 | ENST00000344604.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010800.hg.1 | ENST00000344604.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010800.hg.1 | ENST00000344604.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010800.hg.1 | ENST00000344604.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010800.hg.1 | ENST00000344604.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010800.hg.1 | ENST00000344604.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010800.hg.1 | ENST00000344604.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010800.hg.1 | ENST00000344604.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010800.hg.1 | ENST00000344604.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010800.hg.1 | ENST00000344604.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010800.hg.1 | ENST00000344604.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010800.hg.1 | ENST00000344604.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010800.hg.1 | ENST00000344604.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010800.hg.1 | ENST00000375540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010800.hg.1 | ENST00000375540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010800.hg.1 | ENST00000375540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010800.hg.1 | ENST00000375540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010800.hg.1 | ENST00000375540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010800.hg.1 | ENST00000375540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010800.hg.1 | ENST00000395534.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010800.hg.1 | ENST00000395534.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010800.hg.1 | ENST00000395534.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010800.hg.1 | ENST00000395534.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010800.hg.1 | ENST00000395534.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010800.hg.1 | ENST00000444490.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010800.hg.1 | ENST00000444490.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010800.hg.1 | ENST00000444490.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010800.hg.1 | ENST00000444490.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010800.hg.1 | ENST00000444490.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010800.hg.1 | ENST00000444490.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010800.hg.1 | ENST00000444490.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010800.hg.1 | ENST00000444490.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010800.hg.1 | ENST00000444490.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010800.hg.1 | ENST00000444490.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010800.hg.1 | ENST00000444490.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010800.hg.1 | ENST00000444490.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010800.hg.1 | ENST00000444490.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010800.hg.1 | ENST00000444490.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010800.hg.1 | NM_001197295.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010800.hg.1 | NM_001197295.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010800.hg.1 | NM_001197295.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010800.hg.1 | NM_001197295.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010800.hg.1 | NM_001197295.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010800.hg.1 | NM_001197295.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010800.hg.1 | NM_001197295.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010800.hg.1 | NM_001197295.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010800.hg.1 | NM_001197295.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010800.hg.1 | NM_001197295.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010800.hg.1 | NM_001197295.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010800.hg.1 | NM_001197295.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010800.hg.1 | NM_001197295.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010800.hg.1 | NM_001197295.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010800.hg.1 | NM_001197295.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010800.hg.1 | NM_001197296.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010800.hg.1 | NM_001197296.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010800.hg.1 | NM_001197296.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010800.hg.1 | NM_001197296.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010800.hg.1 | NM_001197296.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010800.hg.1 | NM_001197296.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010800.hg.1 | NM_001197296.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010800.hg.1 | NM_001197296.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010800.hg.1 | NM_001197296.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010800.hg.1 | NM_001197296.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010800.hg.1 | NM_001197296.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010800.hg.1 | NM_001197296.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010800.hg.1 | NM_001197296.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010800.hg.1 | NM_001197296.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010800.hg.1 | NM_001393.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010800.hg.1 | NM_001393.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010800.hg.1 | NM_001393.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010800.hg.1 | NM_001393.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010800.hg.1 | NM_001393.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010800.hg.1 | NM_001393.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010800.hg.1 | NM_001393.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010800.hg.1 | NM_001393.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010800.hg.1 | NM_001393.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010800.hg.1 | NM_001393.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010800.hg.1 | NM_001393.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010800.hg.1 | NM_001393.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010800.hg.1 | NM_001393.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010800.hg.1 | NM_001393.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010800.hg.1 | NM_001393.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010800.hg.1 | NM_001393.3 | Refseq |

ECM2
Genomic Map