Assay Details
Gene Symbol: MNT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020310.2 | NP_064706.1 | 2 - 3 | 907 | 90 | |
XM_011523868.2 | XP_011522170.1 | 2 - 3 | 844 | 90 | ||
GenBank mRNA | AK291596.1 | - | 2 - 3 | 1004 | 90 | |
BC117563.1 | - | 2 - 3 | 657 | 3904263 | 90 | |
X96401.1 | - | 2 - 3 | 867 | 90 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC17000994.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
multicellular organism development
cell aging
negative regulation of cell proliferation
regulation of cell cycle
negative regulation of apoptotic signaling pathway
RNA polymerase II regulatory region sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
transcription coactivator activity
transcription corepressor activity
protein dimerization activity
negative regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
multicellular organism development
cell aging
negative regulation of cell proliferation
regulation of cell cycle
negative regulation of apoptotic signaling pathway
RNA polymerase II regulatory region sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
transcription coactivator activity
transcription corepressor activity
protein dimerization activity
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR17013272.hg.1 | ENST00000174618.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17013272.hg.1 | ENST00000404961.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17013272.hg.1 | NM_020310.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17013272.hg.1 | TC17000994.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1700009436.hg.1 | ENST00000174618.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009436.hg.1 | ENST00000174618.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009436.hg.1 | ENST00000174618.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009436.hg.1 | ENST00000174618.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009436.hg.1 | ENST00000174618.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009436.hg.1 | ENST00000174618.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009436.hg.1 | ENST00000174618.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009436.hg.1 | ENST00000174618.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009436.hg.1 | ENST00000174618.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009436.hg.1 | ENST00000174618.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009436.hg.1 | ENST00000174618.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009436.hg.1 | ENST00000174618.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009436.hg.1 | ENST00000174618.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009436.hg.1 | ENST00000174618.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009436.hg.1 | ENST00000174618.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009436.hg.1 | ENST00000571232.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009436.hg.1 | ENST00000571232.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009436.hg.1 | ENST00000571232.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009436.hg.1 | ENST00000571232.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009436.hg.1 | ENST00000571232.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009436.hg.1 | ENST00000571232.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009436.hg.1 | ENST00000571836.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009436.hg.1 | ENST00000571836.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009436.hg.1 | ENST00000572892.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009436.hg.1 | ENST00000572892.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009436.hg.1 | ENST00000572892.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009436.hg.1 | ENST00000572892.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009436.hg.1 | ENST00000572892.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009436.hg.1 | ENST00000572892.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009436.hg.1 | ENST00000573384.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009436.hg.1 | ENST00000573384.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009436.hg.1 | ENST00000573384.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009436.hg.1 | ENST00000573384.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009436.hg.1 | ENST00000574559.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009436.hg.1 | ENST00000574559.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009436.hg.1 | ENST00000574559.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009436.hg.1 | ENST00000575374.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009436.hg.1 | ENST00000575374.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009436.hg.1 | ENST00000575374.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009436.hg.1 | ENST00000575374.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009436.hg.1 | ENST00000575374.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009436.hg.1 | ENST00000575374.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009436.hg.1 | ENST00000575374.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009436.hg.1 | ENST00000575394.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009436.hg.1 | ENST00000575394.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009436.hg.1 | ENST00000575394.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009436.hg.1 | ENST00000575394.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009436.hg.1 | ENST00000575402.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009436.hg.1 | ENST00000575402.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009436.hg.1 | ENST00000575402.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009436.hg.1 | ENST00000575402.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009436.hg.1 | NM_020310.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009436.hg.1 | NM_020310.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009436.hg.1 | NM_020310.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009436.hg.1 | NM_020310.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009436.hg.1 | NM_020310.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009436.hg.1 | NM_020310.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009436.hg.1 | NM_020310.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009436.hg.1 | NM_020310.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009436.hg.1 | NM_020310.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009436.hg.1 | NM_020310.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009436.hg.1 | NM_020310.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009436.hg.1 | NM_020310.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009436.hg.1 | NM_020310.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009436.hg.1 | NM_020310.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700172240.hg.1 | ENST00000174618.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700172240.hg.1 | NM_020310.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700172240.hg.1 | ENST00000174618.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700172240.hg.1 | NM_020310.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700172240.hg.1 | TC1700009436.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700172238.hg.1 | ENST00000174618.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700172238.hg.1 | NM_020310.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700172238.hg.1 | ENST00000174618.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700172238.hg.1 | NM_020310.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700172238.hg.1 | TC1700009436.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1700009436.hg.1 | ENST00000174618.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009436.hg.1 | ENST00000174618.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009436.hg.1 | ENST00000174618.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009436.hg.1 | ENST00000174618.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009436.hg.1 | ENST00000174618.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009436.hg.1 | ENST00000174618.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009436.hg.1 | ENST00000174618.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009436.hg.1 | ENST00000174618.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009436.hg.1 | ENST00000174618.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009436.hg.1 | ENST00000174618.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009436.hg.1 | ENST00000174618.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009436.hg.1 | ENST00000174618.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009436.hg.1 | ENST00000174618.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009436.hg.1 | ENST00000174618.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009436.hg.1 | ENST00000174618.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009436.hg.1 | ENST00000571232.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009436.hg.1 | ENST00000571232.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009436.hg.1 | ENST00000571232.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009436.hg.1 | ENST00000571232.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009436.hg.1 | ENST00000571232.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009436.hg.1 | ENST00000571232.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009436.hg.1 | ENST00000571836.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009436.hg.1 | ENST00000571836.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009436.hg.1 | ENST00000572892.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009436.hg.1 | ENST00000572892.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009436.hg.1 | ENST00000572892.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009436.hg.1 | ENST00000572892.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009436.hg.1 | ENST00000572892.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009436.hg.1 | ENST00000572892.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009436.hg.1 | ENST00000573384.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009436.hg.1 | ENST00000573384.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009436.hg.1 | ENST00000573384.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009436.hg.1 | ENST00000573384.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009436.hg.1 | ENST00000574559.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009436.hg.1 | ENST00000574559.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009436.hg.1 | ENST00000574559.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009436.hg.1 | ENST00000575374.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009436.hg.1 | ENST00000575374.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009436.hg.1 | ENST00000575374.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009436.hg.1 | ENST00000575374.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009436.hg.1 | ENST00000575374.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009436.hg.1 | ENST00000575374.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009436.hg.1 | ENST00000575374.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009436.hg.1 | ENST00000575394.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009436.hg.1 | ENST00000575394.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009436.hg.1 | ENST00000575394.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009436.hg.1 | ENST00000575394.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009436.hg.1 | ENST00000575402.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009436.hg.1 | ENST00000575402.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009436.hg.1 | ENST00000575402.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009436.hg.1 | ENST00000575402.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009436.hg.1 | NM_020310.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009436.hg.1 | NM_020310.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009436.hg.1 | NM_020310.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009436.hg.1 | NM_020310.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009436.hg.1 | NM_020310.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009436.hg.1 | NM_020310.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009436.hg.1 | NM_020310.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009436.hg.1 | NM_020310.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009436.hg.1 | NM_020310.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009436.hg.1 | NM_020310.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009436.hg.1 | NM_020310.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009436.hg.1 | NM_020310.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009436.hg.1 | NM_020310.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009436.hg.1 | NM_020310.2 | Refseq |
