Assay Details
Gene Symbol: PPP2R5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199756.1 | NP_001186685.1 | 8 - 9 | 1035 | 87 | |
NM_006243.3 | NP_006234.1 | 8 - 9 | 1503 | 87 | ||
GenBank mRNA | AK302202.1 | - | 8 - 9 | 1035 | 87 | |
AK312530.1 | - | 9 - 10 | 1318 | 87 | ||
BC022474.1 | - | 8 - 9 | 1089 | 4815091 | 87 | |
BC110883.1 | - | 8 - 9 | 1212 | 6045358 | 87 | |
L42373.1 | - | 8 - 9 | 1500 | 87 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01001771.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
signal transduction
regulation of protein phosphatase type 2A activity
positive regulation of protein dephosphorylation
negative regulation of establishment of protein localization to plasma membrane
negative regulation of lipid kinase activity
protein binding
protein phosphatase type 2A regulator activity
kinase binding
signal transduction
regulation of protein phosphatase type 2A activity
positive regulation of protein dephosphorylation
negative regulation of establishment of protein localization to plasma membrane
negative regulation of lipid kinase activity
protein binding
protein phosphatase type 2A regulator activity
kinase binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000261461.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000261461.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000261461.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000261461.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000261461.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000261461.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000261461.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000261461.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000261461.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000261461.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000261461.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000261461.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000261461.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000261461.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000261461.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000261461.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000261461.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000261461.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000479259.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000479259.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000479259.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000479259.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000479259.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000498123.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000498123.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000498129.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000498129.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000498129.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000498129.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000498129.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000498129.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000537030.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000537030.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000537030.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000537030.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000537030.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000537030.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000537030.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000537030.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000537030.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000537030.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000537030.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000537030.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000537030.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000537030.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000537030.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | NM_001199756.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | NM_001199756.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | NM_001199756.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | NM_001199756.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | NM_001199756.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | NM_001199756.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | NM_001199756.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | NM_001199756.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | NM_001199756.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | NM_001199756.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | NM_001199756.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | NM_001199756.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | NM_001199756.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | NM_001199756.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | NM_001199756.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | NM_001199756.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | NM_006243.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | NM_006243.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | NM_006243.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | NM_006243.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | NM_006243.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | NM_006243.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | NM_006243.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | NM_006243.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | NM_006243.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | NM_006243.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | NM_006243.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | NM_006243.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | NM_006243.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | NM_006243.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | NM_006243.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | NM_006243.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | NM_006243.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | NM_006243.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01027738.hg.1 | TC01001771.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01027738.hg.1 | ENST00000261461.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01027738.hg.1 | ENST00000537030.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01027738.hg.1 | ENST00000542178.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01027738.hg.1 | NM_001199756.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01027738.hg.1 | NM_006243.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000261461.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000261461.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000261461.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000261461.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000261461.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000261461.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000261461.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000261461.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000261461.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000261461.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000261461.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000261461.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000261461.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000261461.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000261461.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000261461.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000261461.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000261461.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000479259.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000479259.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000479259.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000479259.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000479259.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000498123.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000498123.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000498129.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000498129.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000498129.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000498129.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000498129.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000498129.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000537030.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000537030.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000537030.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000537030.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000537030.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000537030.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000537030.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000537030.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000537030.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000537030.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000537030.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000537030.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000537030.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000537030.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000537030.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | NM_001199756.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | NM_001199756.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | NM_001199756.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | NM_001199756.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | NM_001199756.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | NM_001199756.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | NM_001199756.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | NM_001199756.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | NM_001199756.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | NM_001199756.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | NM_001199756.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | NM_001199756.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | NM_001199756.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | NM_001199756.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | NM_001199756.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | NM_001199756.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | NM_006243.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | NM_006243.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | NM_006243.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | NM_006243.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | NM_006243.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | NM_006243.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | NM_006243.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | NM_006243.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | NM_006243.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | NM_006243.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | NM_006243.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | NM_006243.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | NM_006243.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | NM_006243.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | NM_006243.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | NM_006243.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | NM_006243.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | NM_006243.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100184861.hg.1 | ENST00000261461.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100184861.hg.1 | ENST00000537030.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100184861.hg.1 | NM_001199756.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100184861.hg.1 | NM_006243.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100184861.hg.1 | ENST00000261461.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100184861.hg.1 | ENST00000537030.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100184861.hg.1 | NM_001199756.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100184861.hg.1 | NM_006243.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100184861.hg.1 | TC0100011520.hg.1 | Clariom D TC |
