Assay Details
| Gene Symbol: CELSR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014246.1 | NP_055061.1 | 27 - 28 | 7950 | 79 | |
| XM_006724383.3 | XP_006724446.1 | 27 - 28 | 8277 | 79 | ||
| XM_011530554.2 | XP_011528856.1 | 27 - 28 | 4493 | 79 | ||
| XM_011530555.2 | XP_011528857.1 | 27 - 28 | 4574 | 79 | ||
| GenBank mRNA | AF231024.1 | - | 27 - 28 | 7950 | 79 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC22000889.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
establishment of planar polarity
neuron migration
neural tube closure
hair follicle development
homophilic cell adhesion via plasma membrane adhesion molecules
G-protein coupled receptor signaling pathway
Rho protein signal transduction
central nervous system development
locomotory behavior
anterior/posterior pattern specification
regulation of actin cytoskeleton organization
wound healing
establishment of planar polarity of embryonic epithelium
inner ear morphogenesis
apical protein localization
establishment of body hair planar orientation
Wnt signaling pathway, planar cell polarity pathway
orthogonal dichotomous subdivision of terminal units involved in lung branching morphogenesis
planar dichotomous subdivision of terminal units involved in lung branching morphogenesis
lateral sprouting involved in lung morphogenesis
planar cell polarity pathway involved in neural tube closure
protein localization involved in establishment of planar polarity
transmembrane signaling receptor activity
G-protein coupled receptor activity
calcium ion binding
protein dimerization activity
establishment of planar polarity
neuron migration
neural tube closure
hair follicle development
homophilic cell adhesion via plasma membrane adhesion molecules
G-protein coupled receptor signaling pathway
Rho protein signal transduction
central nervous system development
locomotory behavior
anterior/posterior pattern specification
regulation of actin cytoskeleton organization
wound healing
establishment of planar polarity of embryonic epithelium
inner ear morphogenesis
apical protein localization
establishment of body hair planar orientation
Wnt signaling pathway, planar cell polarity pathway
orthogonal dichotomous subdivision of terminal units involved in lung branching morphogenesis
planar dichotomous subdivision of terminal units involved in lung branching morphogenesis
lateral sprouting involved in lung morphogenesis
planar cell polarity pathway involved in neural tube closure
protein localization involved in establishment of planar polarity
transmembrane signaling receptor activity
G-protein coupled receptor activity
calcium ion binding
protein dimerization activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | ENST00000262738.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | ENST00000262738.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | ENST00000262738.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | ENST00000262738.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | ENST00000262738.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | ENST00000262738.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | ENST00000262738.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | ENST00000262738.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | ENST00000262738.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | ENST00000262738.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | ENST00000262738.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | ENST00000262738.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | ENST00000262738.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | ENST00000262738.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | ENST00000262738.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | ENST00000262738.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | ENST00000262738.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | ENST00000262738.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | ENST00000262738.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | ENST00000262738.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | ENST00000262738.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | ENST00000262738.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | ENST00000262738.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | ENST00000262738.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | ENST00000262738.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | ENST00000262738.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | ENST00000262738.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | ENST00000262738.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | ENST00000262738.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | ENST00000262738.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | ENST00000262738.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | ENST00000262738.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | ENST00000262738.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | ENST00000262738.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | ENST00000262738.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | ENST00000262738.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | ENST00000262738.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | ENST00000262738.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | ENST00000262738.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | ENST00000454637.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | ENST00000454637.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | ENST00000454637.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | ENST00000454637.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | ENST00000468025.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | ENST00000468025.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | ENST00000468025.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | ENST00000473624.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | ENST00000473624.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | ENST00000473624.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | ENST00000473624.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | ENST00000473624.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | ENST00000497509.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | ENST00000497509.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | NM_014246.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | NM_014246.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | NM_014246.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | NM_014246.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | NM_014246.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | NM_014246.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | NM_014246.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | NM_014246.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | NM_014246.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | NM_014246.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | NM_014246.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | NM_014246.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | NM_014246.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | NM_014246.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | NM_014246.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | NM_014246.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | NM_014246.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | NM_014246.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | NM_014246.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | NM_014246.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | NM_014246.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | NM_014246.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | NM_014246.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | NM_014246.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | NM_014246.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | NM_014246.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | NM_014246.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | NM_014246.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | NM_014246.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | NM_014246.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | NM_014246.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | NM_014246.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | NM_014246.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | NM_014246.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | NM_014246.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | NM_014246.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | NM_014246.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | NM_014246.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009021.hg.1 | NM_014246.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22014926.hg.1 | ENST00000262738.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22014926.hg.1 | NM_014246.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22014926.hg.1 | TC22000889.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | ENST00000262738.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | ENST00000262738.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | ENST00000262738.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | ENST00000262738.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | ENST00000262738.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | ENST00000262738.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | ENST00000262738.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | ENST00000262738.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | ENST00000262738.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | ENST00000262738.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | ENST00000262738.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | ENST00000262738.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | ENST00000262738.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | ENST00000262738.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | ENST00000262738.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | ENST00000262738.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | ENST00000262738.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | ENST00000262738.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | ENST00000262738.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | ENST00000262738.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | ENST00000262738.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | ENST00000262738.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | ENST00000262738.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | ENST00000262738.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | ENST00000262738.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | ENST00000262738.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | ENST00000262738.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | ENST00000262738.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | ENST00000262738.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | ENST00000262738.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | ENST00000262738.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | ENST00000262738.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | ENST00000262738.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | ENST00000262738.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | ENST00000262738.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | ENST00000262738.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | ENST00000262738.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | ENST00000262738.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | ENST00000262738.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | ENST00000454637.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | ENST00000454637.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | ENST00000454637.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | ENST00000454637.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | ENST00000468025.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | ENST00000468025.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | ENST00000468025.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | ENST00000473624.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | ENST00000473624.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | ENST00000473624.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | ENST00000473624.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | ENST00000473624.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | ENST00000497509.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | ENST00000497509.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | NM_014246.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | NM_014246.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | NM_014246.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | NM_014246.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | NM_014246.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | NM_014246.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | NM_014246.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | NM_014246.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | NM_014246.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | NM_014246.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | NM_014246.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | NM_014246.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | NM_014246.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | NM_014246.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | NM_014246.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | NM_014246.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | NM_014246.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | NM_014246.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | NM_014246.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | NM_014246.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | NM_014246.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | NM_014246.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | NM_014246.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | NM_014246.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | NM_014246.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | NM_014246.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | NM_014246.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | NM_014246.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | NM_014246.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | NM_014246.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | NM_014246.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | NM_014246.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | NM_014246.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | NM_014246.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | NM_014246.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | NM_014246.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | NM_014246.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | NM_014246.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009021.hg.1 | NM_014246.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200165346.hg.1 | TC2200009021.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200165346.hg.1 | ENST00000262738.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200165346.hg.1 | NM_014246.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200165346.hg.1 | ENST00000262738.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200165346.hg.1 | NM_014246.1 | Refseq |

CELSR1
Genomic Map