Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011181.hg.1 |
ENST00000262065.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011181.hg.1 |
ENST00000262065.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011181.hg.1 |
ENST00000262065.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011181.hg.1 |
ENST00000262065.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011181.hg.1 |
ENST00000262065.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011181.hg.1 |
ENST00000262065.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011181.hg.1 |
ENST00000262065.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011181.hg.1 |
ENST00000262065.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011181.hg.1 |
ENST00000262065.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011181.hg.1 |
ENST00000262065.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011181.hg.1 |
ENST00000262065.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011181.hg.1 |
ENST00000262065.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011181.hg.1 |
ENST00000571578.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011181.hg.1 |
ENST00000571578.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011181.hg.1 |
ENST00000571578.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011181.hg.1 |
ENST00000571578.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011181.hg.1 |
ENST00000571578.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011181.hg.1 |
ENST00000571578.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011181.hg.1 |
ENST00000571578.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011181.hg.1 |
ENST00000577038.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011181.hg.1 |
ENST00000577038.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011181.hg.1 |
ENST00000577038.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011181.hg.1 |
NM_012329.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011181.hg.1 |
NM_012329.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011181.hg.1 |
NM_012329.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011181.hg.1 |
NM_012329.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011181.hg.1 |
NM_012329.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011181.hg.1 |
NM_012329.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011181.hg.1 |
NM_012329.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011181.hg.1 |
NM_012329.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011181.hg.1 |
NM_012329.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011181.hg.1 |
NM_012329.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011181.hg.1 |
NM_012329.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011181.hg.1 |
NM_012329.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17022979.hg.1 |
ENST00000262065.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17022979.hg.1 |
NM_012329.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17022979.hg.1 |
TC17001711.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1700011181.hg.1 |
ENST00000262065.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011181.hg.1 |
ENST00000262065.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011181.hg.1 |
ENST00000262065.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011181.hg.1 |
ENST00000262065.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011181.hg.1 |
ENST00000262065.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011181.hg.1 |
ENST00000262065.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011181.hg.1 |
ENST00000262065.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011181.hg.1 |
ENST00000262065.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011181.hg.1 |
ENST00000262065.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011181.hg.1 |
ENST00000262065.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011181.hg.1 |
ENST00000262065.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011181.hg.1 |
ENST00000262065.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011181.hg.1 |
ENST00000571578.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011181.hg.1 |
ENST00000571578.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011181.hg.1 |
ENST00000571578.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011181.hg.1 |
ENST00000571578.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011181.hg.1 |
ENST00000571578.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011181.hg.1 |
ENST00000571578.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011181.hg.1 |
ENST00000571578.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011181.hg.1 |
ENST00000577038.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011181.hg.1 |
ENST00000577038.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011181.hg.1 |
ENST00000577038.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011181.hg.1 |
NM_012329.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011181.hg.1 |
NM_012329.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011181.hg.1 |
NM_012329.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011181.hg.1 |
NM_012329.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011181.hg.1 |
NM_012329.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011181.hg.1 |
NM_012329.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011181.hg.1 |
NM_012329.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011181.hg.1 |
NM_012329.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011181.hg.1 |
NM_012329.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011181.hg.1 |
NM_012329.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011181.hg.1 |
NM_012329.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011181.hg.1 |
NM_012329.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700189892.hg.1 |
ENST00000262065.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700189892.hg.1 |
ENST00000571578.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700189892.hg.1 |
NM_012329.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700189892.hg.1 |
ENST00000262065.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700189892.hg.1 |
ENST00000571578.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700189892.hg.1 |
NM_012329.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700189892.hg.1 |
TC1700011181.hg.1 |
Clariom D TC |