Assay Details
| Gene Symbol: SMARCA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128846.1 | NP_001122318.1 | 28 - 29 | 4336 | 99 | |
| NM_001128848.1 | NP_001122320.1 | 28 - 29 | 4327 | 99 | ||
| XM_017027160.1 | XP_016882649.1 | 32 - 33 | 4803 | 99 | ||
| XM_017027163.1 | XP_016882652.1 | 31 - 32 | 4707 | 99 | ||
| XM_017027164.1 | XP_016882653.1 | 31 - 32 | 4704 | 99 | ||
| XM_017027166.1 | XP_016882655.1 | 30 - 31 | 4617 | 99 | ||
| XM_017027168.1 | XP_016882657.1 | 30 - 31 | 4608 | 99 | ||
| GenBank mRNA | EU430756.1 | - | 28 - 29 | 4327 | 99 | |
| EU430758.1 | - | 28 - 29 | 4336 | 99 | ||
More Information
Set Membership: |
|

SMARCA4
Genomic Map