| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Gene 1.0 ST Array |
8092703 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8092704 |
- |
- |
| Affymetrix Human Gene 1.1 ST Array |
8092702 |
- |
- |
| Affymetrix HT HG-U133+ PM Array Plate |
224526_PM_at |
- |
- |
| Affymetrix Human Genome U219 Array |
11763886_PM_s_at |
- |
- |
| Affymetrix Human Genome U219 Array |
11763886_s_at |
- |
- |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013914.hg.1 |
ENST00000464152.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013914.hg.1 |
ENST00000464152.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013914.hg.1 |
ENST00000464152.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013914.hg.1 |
ENST00000464152.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013914.hg.1 |
ENST00000473790.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013914.hg.1 |
ENST00000473790.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013914.hg.1 |
ENST00000473790.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013914.hg.1 |
ENST00000473790.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013914.hg.1 |
ENST00000479720.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013914.hg.1 |
ENST00000479720.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013914.hg.1 |
ENST00000479720.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013914.hg.1 |
ENST00000479720.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013914.hg.1 |
ENST00000488841.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013914.hg.1 |
ENST00000488841.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013914.hg.1 |
ENST00000488841.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013914.hg.1 |
ENST00000488841.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013914.hg.1 |
ENST00000488841.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013914.hg.1 |
ENST00000488841.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013914.hg.1 |
ENST00000488841.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013914.hg.1 |
ENST00000497628.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013914.hg.1 |
ENST00000497628.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013914.hg.1 |
ENST00000497628.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013914.hg.1 |
ENST00000497628.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013914.hg.1 |
ENST00000464152.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013914.hg.1 |
ENST00000464152.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013914.hg.1 |
ENST00000464152.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013914.hg.1 |
ENST00000464152.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013914.hg.1 |
ENST00000473790.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013914.hg.1 |
ENST00000473790.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013914.hg.1 |
ENST00000473790.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013914.hg.1 |
ENST00000473790.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013914.hg.1 |
ENST00000479720.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013914.hg.1 |
ENST00000479720.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013914.hg.1 |
ENST00000479720.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013914.hg.1 |
ENST00000479720.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013914.hg.1 |
ENST00000488841.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013914.hg.1 |
ENST00000488841.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013914.hg.1 |
ENST00000488841.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013914.hg.1 |
ENST00000488841.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013914.hg.1 |
ENST00000488841.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013914.hg.1 |
ENST00000488841.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013914.hg.1 |
ENST00000488841.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013914.hg.1 |
ENST00000497628.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013914.hg.1 |
ENST00000497628.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013914.hg.1 |
ENST00000497628.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013914.hg.1 |
ENST00000497628.1 |
Ensembl |
| Human Genome U133 Plus 2.0 Array |
224526_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
224526_PM_at |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
224526_at11 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
224526_at3 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
224526_at6 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
224526_at2 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
224526_at4 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
224526_at9 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
224526_at7 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
224526_at1 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
224526_at8 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
224526_at10 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
224526_at5 |
- |
- |
| Human Genome U133 Set |
224526_at |
- |
- |
| Human Genome U133 Set |
224526_PM_at |
- |
- |