Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09008520.hg.1 |
TC09000695.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09008520.hg.1 |
ENST00000358161.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09008520.hg.1 |
ENST00000372719.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09008520.hg.1 |
ENST00000372721.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09008520.hg.1 |
ENST00000372731.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09008520.hg.1 |
ENST00000372739.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09008520.hg.1 |
NM_001130438.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09008520.hg.1 |
NM_001195532.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09008520.hg.1 |
NM_003127.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900184862.hg.1 |
ENST00000358161.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900184862.hg.1 |
ENST00000372731.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900184862.hg.1 |
ENST00000372739.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900184862.hg.1 |
ENST00000461855.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900184862.hg.1 |
ENST00000630804.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900184862.hg.1 |
ENST00000630866.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900184862.hg.1 |
NM_001130438.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900184862.hg.1 |
NM_001195532.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900184862.hg.1 |
NM_003127.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900184862.hg.1 |
ENST00000358161.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900184862.hg.1 |
ENST00000372731.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900184862.hg.1 |
ENST00000372739.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900184862.hg.1 |
ENST00000461855.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900184862.hg.1 |
ENST00000630804.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900184862.hg.1 |
ENST00000630866.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900184862.hg.1 |
NM_001130438.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900184862.hg.1 |
NM_001195532.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900184862.hg.1 |
NM_003127.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900184862.hg.1 |
TC0900012179.hg.1 |
Clariom D TC |