Assay Details
Gene Symbol: BUB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_004725.3 | NP_004716.1 | 7 - 8 | 1214 | 75 | |
GenBank mRNA | AF047472.1 | - | 7 - 8 | 1042 | 75 | |
AF053304.1 | - | 7 - 8 | 1010 | 75 | ||
AF081496.1 | - | 7 - 8 | 1059 | 75 | ||
AK293255.1 | - | 6 - 7 | 870 | 75 | ||
BC005138.2 | - | 7 - 8 | 1043 | 3504520 | 75 | |
BC022438.1 | - | 7 - 8 | 1035 | 4281970 | 75 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC10000900.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
mitotic sister chromatid segregation
sister chromatid cohesion
mitotic spindle assembly checkpoint
attachment of spindle microtubules to kinetochore
anaphase-promoting complex-dependent catabolic process
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
proteasome-mediated ubiquitin-dependent protein catabolic process
cell division
meiotic cell cycle
negative regulation of ubiquitin-protein ligase activity involved in mitotic cell cycle
positive regulation of ubiquitin-protein ligase activity involved in regulation of mitotic cell cycle transition
spindle assembly checkpoint
protein binding
ubiquitin binding
mitotic sister chromatid segregation
sister chromatid cohesion
mitotic spindle assembly checkpoint
attachment of spindle microtubules to kinetochore
anaphase-promoting complex-dependent catabolic process
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
proteasome-mediated ubiquitin-dependent protein catabolic process
cell division
meiotic cell cycle
negative regulation of ubiquitin-protein ligase activity involved in mitotic cell cycle
positive regulation of ubiquitin-protein ligase activity involved in regulation of mitotic cell cycle transition
spindle assembly checkpoint
protein binding
ubiquitin binding
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR10011037.hg.1 | ENST00000368865.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10011037.hg.1 | NM_004725.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10011037.hg.1 | TC10000900.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR10011036.hg.1 | ENST00000368865.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10011036.hg.1 | ENST00000538238.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10011036.hg.1 | NM_004725.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10011036.hg.1 | TC10000900.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR10011038.hg.1 | ENST00000368865.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10011038.hg.1 | NM_004725.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10011038.hg.1 | TC10000900.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR10011035.hg.1 | ENST00000368865.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10011035.hg.1 | ENST00000538238.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10011035.hg.1 | NM_004725.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10011035.hg.1 | TC10000900.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1000009172.hg.1 | ENST00000368858.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009172.hg.1 | ENST00000368858.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009172.hg.1 | ENST00000368858.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009172.hg.1 | ENST00000368858.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009172.hg.1 | ENST00000368858.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009172.hg.1 | ENST00000368858.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009172.hg.1 | ENST00000368858.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009172.hg.1 | ENST00000368858.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009172.hg.1 | ENST00000368858.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009172.hg.1 | ENST00000368858.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009172.hg.1 | ENST00000368859.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009172.hg.1 | ENST00000368859.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009172.hg.1 | ENST00000368859.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009172.hg.1 | ENST00000368859.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009172.hg.1 | ENST00000368859.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009172.hg.1 | ENST00000368865.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009172.hg.1 | ENST00000368865.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009172.hg.1 | ENST00000368865.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009172.hg.1 | ENST00000368865.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009172.hg.1 | ENST00000368865.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009172.hg.1 | ENST00000368865.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009172.hg.1 | ENST00000368865.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009172.hg.1 | ENST00000368865.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009172.hg.1 | ENST00000368865.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009172.hg.1 | ENST00000368865.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009172.hg.1 | ENST00000368865.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009172.hg.1 | ENST00000368865.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009172.hg.1 | ENST00000368865.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009172.hg.1 | ENST00000368865.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009172.hg.1 | ENST00000368865.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009172.hg.1 | ENST00000407911.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009172.hg.1 | ENST00000407911.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009172.hg.1 | ENST00000407911.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009172.hg.1 | ENST00000407911.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009172.hg.1 | ENST00000407911.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009172.hg.1 | ENST00000407911.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009172.hg.1 | ENST00000407911.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009172.hg.1 | ENST00000481952.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009172.hg.1 | ENST00000481952.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009172.hg.1 | ENST00000481952.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009172.hg.1 | ENST00000481952.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009172.hg.1 | NM_001007793.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009172.hg.1 | NM_001007793.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009172.hg.1 | NM_001007793.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009172.hg.1 | NM_001007793.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009172.hg.1 | NM_001007793.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009172.hg.1 | NM_001007793.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009172.hg.1 | NM_001007793.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009172.hg.1 | NM_001007793.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009172.hg.1 | NM_001007793.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009172.hg.1 | NM_001007793.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009172.hg.1 | NM_001007793.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009172.hg.1 | NM_001007793.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009172.hg.1 | NM_004725.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009172.hg.1 | NM_004725.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009172.hg.1 | NM_004725.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009172.hg.1 | NM_004725.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009172.hg.1 | NM_004725.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009172.hg.1 | NM_004725.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009172.hg.1 | NM_004725.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009172.hg.1 | NM_004725.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009172.hg.1 | NM_004725.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009172.hg.1 | NM_004725.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009172.hg.1 | NM_004725.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009172.hg.1 | NM_004725.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009172.hg.1 | NM_004725.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009172.hg.1 | NM_004725.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009172.hg.1 | NM_004725.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000162981.hg.1 | ENST00000368865.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000162981.hg.1 | NM_004725.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000162981.hg.1 | ENST00000368865.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000162981.hg.1 | NM_004725.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000162981.hg.1 | TC1000009172.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1000162979.hg.1 | ENST00000368865.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000162979.hg.1 | NM_004725.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000162979.hg.1 | ENST00000368865.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000162979.hg.1 | NM_004725.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000162979.hg.1 | TC1000009172.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1000162980.hg.1 | ENST00000368865.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000162980.hg.1 | NM_004725.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000162980.hg.1 | ENST00000368865.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000162980.hg.1 | NM_004725.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000162980.hg.1 | TC1000009172.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1000009172.hg.1 | ENST00000368858.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009172.hg.1 | ENST00000368858.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009172.hg.1 | ENST00000368858.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009172.hg.1 | ENST00000368858.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009172.hg.1 | ENST00000368858.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009172.hg.1 | ENST00000368858.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009172.hg.1 | ENST00000368858.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009172.hg.1 | ENST00000368858.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009172.hg.1 | ENST00000368858.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009172.hg.1 | ENST00000368858.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009172.hg.1 | ENST00000368859.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009172.hg.1 | ENST00000368859.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009172.hg.1 | ENST00000368859.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009172.hg.1 | ENST00000368859.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009172.hg.1 | ENST00000368859.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009172.hg.1 | ENST00000368865.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009172.hg.1 | ENST00000368865.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009172.hg.1 | ENST00000368865.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009172.hg.1 | ENST00000368865.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009172.hg.1 | ENST00000368865.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009172.hg.1 | ENST00000368865.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009172.hg.1 | ENST00000368865.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009172.hg.1 | ENST00000368865.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009172.hg.1 | ENST00000368865.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009172.hg.1 | ENST00000368865.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009172.hg.1 | ENST00000368865.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009172.hg.1 | ENST00000368865.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009172.hg.1 | ENST00000368865.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009172.hg.1 | ENST00000368865.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009172.hg.1 | ENST00000368865.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009172.hg.1 | ENST00000407911.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009172.hg.1 | ENST00000407911.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009172.hg.1 | ENST00000407911.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009172.hg.1 | ENST00000407911.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009172.hg.1 | ENST00000407911.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009172.hg.1 | ENST00000407911.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009172.hg.1 | ENST00000407911.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009172.hg.1 | ENST00000481952.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009172.hg.1 | ENST00000481952.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009172.hg.1 | ENST00000481952.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009172.hg.1 | ENST00000481952.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009172.hg.1 | NM_001007793.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009172.hg.1 | NM_001007793.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009172.hg.1 | NM_001007793.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009172.hg.1 | NM_001007793.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009172.hg.1 | NM_001007793.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009172.hg.1 | NM_001007793.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009172.hg.1 | NM_001007793.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009172.hg.1 | NM_001007793.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009172.hg.1 | NM_001007793.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009172.hg.1 | NM_001007793.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009172.hg.1 | NM_001007793.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009172.hg.1 | NM_001007793.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009172.hg.1 | NM_004725.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009172.hg.1 | NM_004725.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009172.hg.1 | NM_004725.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009172.hg.1 | NM_004725.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009172.hg.1 | NM_004725.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009172.hg.1 | NM_004725.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009172.hg.1 | NM_004725.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009172.hg.1 | NM_004725.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009172.hg.1 | NM_004725.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009172.hg.1 | NM_004725.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009172.hg.1 | NM_004725.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009172.hg.1 | NM_004725.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009172.hg.1 | NM_004725.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009172.hg.1 | NM_004725.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009172.hg.1 | NM_004725.3 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 108339 | - | - |
