Assay Details
Gene Symbol: IPO9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018085.4 | NP_060555.2 | 9 - 10 | 1019 | 108 | |
GenBank mRNA | AF410465.1 | - | 9 - 10 | 969 | 108 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01001664.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR01025760.hg.1 | TC01001664.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01025760.hg.1 | ENST00000361565.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01025760.hg.1 | NM_018085.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011192.hg.1 | ENST00000361565.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011192.hg.1 | ENST00000361565.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011192.hg.1 | ENST00000361565.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011192.hg.1 | ENST00000361565.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011192.hg.1 | ENST00000361565.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011192.hg.1 | ENST00000361565.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011192.hg.1 | ENST00000361565.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011192.hg.1 | ENST00000361565.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011192.hg.1 | ENST00000361565.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011192.hg.1 | ENST00000361565.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011192.hg.1 | ENST00000361565.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011192.hg.1 | ENST00000361565.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011192.hg.1 | ENST00000361565.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011192.hg.1 | ENST00000361565.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011192.hg.1 | ENST00000361565.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011192.hg.1 | ENST00000361565.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011192.hg.1 | ENST00000361565.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011192.hg.1 | ENST00000361565.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011192.hg.1 | ENST00000361565.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011192.hg.1 | ENST00000361565.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011192.hg.1 | ENST00000361565.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011192.hg.1 | ENST00000361565.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011192.hg.1 | ENST00000361565.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011192.hg.1 | ENST00000361565.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011192.hg.1 | ENST00000361565.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011192.hg.1 | ENST00000361565.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011192.hg.1 | ENST00000361565.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011192.hg.1 | ENST00000361565.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011192.hg.1 | ENST00000361565.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011192.hg.1 | ENST00000456707.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011192.hg.1 | ENST00000456707.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011192.hg.1 | ENST00000456707.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011192.hg.1 | ENST00000456707.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011192.hg.1 | ENST00000456707.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011192.hg.1 | ENST00000464348.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011192.hg.1 | ENST00000464348.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011192.hg.1 | ENST00000464348.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011192.hg.1 | ENST00000464348.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011192.hg.1 | ENST00000464348.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011192.hg.1 | ENST00000464348.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011192.hg.1 | ENST00000464348.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011192.hg.1 | ENST00000464348.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011192.hg.1 | ENST00000479374.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011192.hg.1 | ENST00000479374.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011192.hg.1 | ENST00000479374.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011192.hg.1 | ENST00000479374.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011192.hg.1 | NM_018085.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011192.hg.1 | NM_018085.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011192.hg.1 | NM_018085.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011192.hg.1 | NM_018085.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011192.hg.1 | NM_018085.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011192.hg.1 | NM_018085.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011192.hg.1 | NM_018085.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011192.hg.1 | NM_018085.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011192.hg.1 | NM_018085.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011192.hg.1 | NM_018085.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011192.hg.1 | NM_018085.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011192.hg.1 | NM_018085.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011192.hg.1 | NM_018085.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011192.hg.1 | NM_018085.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011192.hg.1 | NM_018085.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011192.hg.1 | NM_018085.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011192.hg.1 | NM_018085.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011192.hg.1 | NM_018085.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011192.hg.1 | NM_018085.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011192.hg.1 | NM_018085.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011192.hg.1 | NM_018085.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011192.hg.1 | NM_018085.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011192.hg.1 | NM_018085.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011192.hg.1 | NM_018085.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011192.hg.1 | NM_018085.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011192.hg.1 | NM_018085.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011192.hg.1 | NM_018085.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011192.hg.1 | NM_018085.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011192.hg.1 | NM_018085.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100182050.hg.1 | ENST00000361565.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100182050.hg.1 | NM_018085.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100182050.hg.1 | ENST00000361565.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100182050.hg.1 | NM_018085.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100182050.hg.1 | TC0100011192.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0100011192.hg.1 | ENST00000361565.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011192.hg.1 | ENST00000361565.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011192.hg.1 | ENST00000361565.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011192.hg.1 | ENST00000361565.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011192.hg.1 | ENST00000361565.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011192.hg.1 | ENST00000361565.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011192.hg.1 | ENST00000361565.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011192.hg.1 | ENST00000361565.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011192.hg.1 | ENST00000361565.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011192.hg.1 | ENST00000361565.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011192.hg.1 | ENST00000361565.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011192.hg.1 | ENST00000361565.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011192.hg.1 | ENST00000361565.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011192.hg.1 | ENST00000361565.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011192.hg.1 | ENST00000361565.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011192.hg.1 | ENST00000361565.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011192.hg.1 | ENST00000361565.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011192.hg.1 | ENST00000361565.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011192.hg.1 | ENST00000361565.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011192.hg.1 | ENST00000361565.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011192.hg.1 | ENST00000361565.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011192.hg.1 | ENST00000361565.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011192.hg.1 | ENST00000361565.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011192.hg.1 | ENST00000361565.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011192.hg.1 | ENST00000361565.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011192.hg.1 | ENST00000361565.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011192.hg.1 | ENST00000361565.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011192.hg.1 | ENST00000361565.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011192.hg.1 | ENST00000361565.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011192.hg.1 | ENST00000456707.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011192.hg.1 | ENST00000456707.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011192.hg.1 | ENST00000456707.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011192.hg.1 | ENST00000456707.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011192.hg.1 | ENST00000456707.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011192.hg.1 | ENST00000464348.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011192.hg.1 | ENST00000464348.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011192.hg.1 | ENST00000464348.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011192.hg.1 | ENST00000464348.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011192.hg.1 | ENST00000464348.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011192.hg.1 | ENST00000464348.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011192.hg.1 | ENST00000464348.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011192.hg.1 | ENST00000464348.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011192.hg.1 | ENST00000479374.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011192.hg.1 | ENST00000479374.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011192.hg.1 | ENST00000479374.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011192.hg.1 | ENST00000479374.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011192.hg.1 | NM_018085.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011192.hg.1 | NM_018085.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011192.hg.1 | NM_018085.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011192.hg.1 | NM_018085.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011192.hg.1 | NM_018085.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011192.hg.1 | NM_018085.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011192.hg.1 | NM_018085.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011192.hg.1 | NM_018085.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011192.hg.1 | NM_018085.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011192.hg.1 | NM_018085.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011192.hg.1 | NM_018085.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011192.hg.1 | NM_018085.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011192.hg.1 | NM_018085.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011192.hg.1 | NM_018085.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011192.hg.1 | NM_018085.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011192.hg.1 | NM_018085.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011192.hg.1 | NM_018085.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011192.hg.1 | NM_018085.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011192.hg.1 | NM_018085.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011192.hg.1 | NM_018085.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011192.hg.1 | NM_018085.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011192.hg.1 | NM_018085.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011192.hg.1 | NM_018085.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011192.hg.1 | NM_018085.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011192.hg.1 | NM_018085.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011192.hg.1 | NM_018085.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011192.hg.1 | NM_018085.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011192.hg.1 | NM_018085.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011192.hg.1 | NM_018085.4 | Refseq |
