Assay Details
Gene Symbol: NEK7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017000345.1 | XP_016855834.1 | 1 - 2 | 269 | 74 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017000345.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
cytokinesis
protein phosphorylation
regulation of mitotic cell cycle
positive regulation of telomere maintenance via telomerase
spindle assembly
positive regulation of telomerase activity
positive regulation of telomere capping
protein serine/threonine kinase activity
ATP binding
metal ion binding
cytokinesis
protein phosphorylation
regulation of mitotic cell cycle
positive regulation of telomere maintenance via telomerase
spindle assembly
positive regulation of telomerase activity
positive regulation of telomere capping
protein serine/threonine kinase activity
ATP binding
metal ion binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100011096.hg.1 | ENST00000367383.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011096.hg.1 | ENST00000367383.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011096.hg.1 | ENST00000367383.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011096.hg.1 | ENST00000367383.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011096.hg.1 | ENST00000367383.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011096.hg.1 | ENST00000367383.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011096.hg.1 | ENST00000367385.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011096.hg.1 | ENST00000367385.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011096.hg.1 | ENST00000367385.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011096.hg.1 | ENST00000367385.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011096.hg.1 | ENST00000367385.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011096.hg.1 | ENST00000367385.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011096.hg.1 | ENST00000367385.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011096.hg.1 | ENST00000367385.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011096.hg.1 | ENST00000367385.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011096.hg.1 | ENST00000367385.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011096.hg.1 | ENST00000367385.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011096.hg.1 | ENST00000367385.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011096.hg.1 | ENST00000367385.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011096.hg.1 | ENST00000367385.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011096.hg.1 | ENST00000367385.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011096.hg.1 | ENST00000391974.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011096.hg.1 | ENST00000391974.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011096.hg.1 | ENST00000391974.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011096.hg.1 | ENST00000391974.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011096.hg.1 | ENST00000391974.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011096.hg.1 | ENST00000391974.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011096.hg.1 | ENST00000391974.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011096.hg.1 | ENST00000417895.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011096.hg.1 | ENST00000417895.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011096.hg.1 | ENST00000417895.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011096.hg.1 | ENST00000417895.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011096.hg.1 | ENST00000417895.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011096.hg.1 | ENST00000442588.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011096.hg.1 | ENST00000442588.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011096.hg.1 | ENST00000442588.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011096.hg.1 | ENST00000442588.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011096.hg.1 | ENST00000442588.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011096.hg.1 | ENST00000442588.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011096.hg.1 | ENST00000493790.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011096.hg.1 | ENST00000493790.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011096.hg.1 | ENST00000493790.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011096.hg.1 | ENST00000493790.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011096.hg.1 | ENST00000493790.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011096.hg.1 | ENST00000493790.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011096.hg.1 | ENST00000493790.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011096.hg.1 | ENST00000538004.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011096.hg.1 | ENST00000538004.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011096.hg.1 | ENST00000538004.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011096.hg.1 | ENST00000538004.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011096.hg.1 | ENST00000538004.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011096.hg.1 | ENST00000538004.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011096.hg.1 | ENST00000538004.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011096.hg.1 | ENST00000538004.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011096.hg.1 | ENST00000538004.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011096.hg.1 | ENST00000538004.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011096.hg.1 | ENST00000538004.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011096.hg.1 | ENST00000538004.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011096.hg.1 | ENST00000544035.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011096.hg.1 | ENST00000544035.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011096.hg.1 | ENST00000544035.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011096.hg.1 | ENST00000544035.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011096.hg.1 | ENST00000544035.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011096.hg.1 | NM_133494.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011096.hg.1 | NM_133494.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011096.hg.1 | NM_133494.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011096.hg.1 | NM_133494.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011096.hg.1 | NM_133494.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011096.hg.1 | NM_133494.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011096.hg.1 | NM_133494.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011096.hg.1 | NM_133494.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011096.hg.1 | NM_133494.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011096.hg.1 | NM_133494.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011096.hg.1 | NM_133494.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011096.hg.1 | NM_133494.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011096.hg.1 | NM_133494.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011096.hg.1 | NM_133494.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011096.hg.1 | NM_133494.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011096.hg.1 | ENST00000367383.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011096.hg.1 | ENST00000367383.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011096.hg.1 | ENST00000367383.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011096.hg.1 | ENST00000367383.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011096.hg.1 | ENST00000367383.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011096.hg.1 | ENST00000367383.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011096.hg.1 | ENST00000367385.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011096.hg.1 | ENST00000367385.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011096.hg.1 | ENST00000367385.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011096.hg.1 | ENST00000367385.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011096.hg.1 | ENST00000367385.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011096.hg.1 | ENST00000367385.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011096.hg.1 | ENST00000367385.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011096.hg.1 | ENST00000367385.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011096.hg.1 | ENST00000367385.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011096.hg.1 | ENST00000367385.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011096.hg.1 | ENST00000367385.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011096.hg.1 | ENST00000367385.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011096.hg.1 | ENST00000367385.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011096.hg.1 | ENST00000367385.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011096.hg.1 | ENST00000367385.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011096.hg.1 | ENST00000391974.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011096.hg.1 | ENST00000391974.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011096.hg.1 | ENST00000391974.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011096.hg.1 | ENST00000391974.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011096.hg.1 | ENST00000391974.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011096.hg.1 | ENST00000391974.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011096.hg.1 | ENST00000391974.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011096.hg.1 | ENST00000417895.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011096.hg.1 | ENST00000417895.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011096.hg.1 | ENST00000417895.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011096.hg.1 | ENST00000417895.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011096.hg.1 | ENST00000417895.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011096.hg.1 | ENST00000442588.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011096.hg.1 | ENST00000442588.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011096.hg.1 | ENST00000442588.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011096.hg.1 | ENST00000442588.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011096.hg.1 | ENST00000442588.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011096.hg.1 | ENST00000442588.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011096.hg.1 | ENST00000493790.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011096.hg.1 | ENST00000493790.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011096.hg.1 | ENST00000493790.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011096.hg.1 | ENST00000493790.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011096.hg.1 | ENST00000493790.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011096.hg.1 | ENST00000493790.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011096.hg.1 | ENST00000493790.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011096.hg.1 | ENST00000538004.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011096.hg.1 | ENST00000538004.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011096.hg.1 | ENST00000538004.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011096.hg.1 | ENST00000538004.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011096.hg.1 | ENST00000538004.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011096.hg.1 | ENST00000538004.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011096.hg.1 | ENST00000538004.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011096.hg.1 | ENST00000538004.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011096.hg.1 | ENST00000538004.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011096.hg.1 | ENST00000538004.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011096.hg.1 | ENST00000538004.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011096.hg.1 | ENST00000538004.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011096.hg.1 | ENST00000544035.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011096.hg.1 | ENST00000544035.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011096.hg.1 | ENST00000544035.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011096.hg.1 | ENST00000544035.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011096.hg.1 | ENST00000544035.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011096.hg.1 | NM_133494.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011096.hg.1 | NM_133494.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011096.hg.1 | NM_133494.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011096.hg.1 | NM_133494.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011096.hg.1 | NM_133494.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011096.hg.1 | NM_133494.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011096.hg.1 | NM_133494.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011096.hg.1 | NM_133494.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011096.hg.1 | NM_133494.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011096.hg.1 | NM_133494.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011096.hg.1 | NM_133494.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011096.hg.1 | NM_133494.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011096.hg.1 | NM_133494.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011096.hg.1 | NM_133494.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011096.hg.1 | NM_133494.2 | Refseq |
