Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17023046.hg.1 |
ENST00000313608.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17023046.hg.1 |
NM_016070.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17023046.hg.1 |
TC17001720.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011233.hg.1 |
ENST00000313608.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011233.hg.1 |
ENST00000313608.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011233.hg.1 |
ENST00000313608.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011233.hg.1 |
ENST00000313608.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011233.hg.1 |
ENST00000313608.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011233.hg.1 |
ENST00000313608.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011233.hg.1 |
ENST00000313608.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011233.hg.1 |
ENST00000313608.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011233.hg.1 |
ENST00000313608.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011233.hg.1 |
ENST00000313608.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011233.hg.1 |
ENST00000578444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011233.hg.1 |
ENST00000578444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011233.hg.1 |
ENST00000578444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011233.hg.1 |
ENST00000578444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011233.hg.1 |
ENST00000578444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011233.hg.1 |
ENST00000578444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011233.hg.1 |
ENST00000578444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011233.hg.1 |
ENST00000579380.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011233.hg.1 |
ENST00000579380.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011233.hg.1 |
ENST00000579380.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011233.hg.1 |
ENST00000579380.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011233.hg.1 |
ENST00000579380.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011233.hg.1 |
ENST00000579380.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011233.hg.1 |
ENST00000579768.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011233.hg.1 |
ENST00000579768.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011233.hg.1 |
ENST00000579768.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011233.hg.1 |
NM_016070.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011233.hg.1 |
NM_016070.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011233.hg.1 |
NM_016070.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011233.hg.1 |
NM_016070.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011233.hg.1 |
NM_016070.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011233.hg.1 |
NM_016070.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011233.hg.1 |
NM_016070.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011233.hg.1 |
NM_016070.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011233.hg.1 |
NM_016070.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700190175.hg.1 |
ENST00000313608.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700190175.hg.1 |
ENST00000578444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700190175.hg.1 |
ENST00000579380.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700190175.hg.1 |
NM_016070.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700190175.hg.1 |
ENST00000313608.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700190175.hg.1 |
ENST00000578444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700190175.hg.1 |
ENST00000579380.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700190175.hg.1 |
NM_016070.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700190175.hg.1 |
TC1700011233.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700011233.hg.1 |
ENST00000313608.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011233.hg.1 |
ENST00000313608.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011233.hg.1 |
ENST00000313608.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011233.hg.1 |
ENST00000313608.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011233.hg.1 |
ENST00000313608.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011233.hg.1 |
ENST00000313608.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011233.hg.1 |
ENST00000313608.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011233.hg.1 |
ENST00000313608.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011233.hg.1 |
ENST00000313608.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011233.hg.1 |
ENST00000313608.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011233.hg.1 |
ENST00000578444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011233.hg.1 |
ENST00000578444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011233.hg.1 |
ENST00000578444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011233.hg.1 |
ENST00000578444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011233.hg.1 |
ENST00000578444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011233.hg.1 |
ENST00000578444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011233.hg.1 |
ENST00000578444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011233.hg.1 |
ENST00000579380.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011233.hg.1 |
ENST00000579380.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011233.hg.1 |
ENST00000579380.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011233.hg.1 |
ENST00000579380.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011233.hg.1 |
ENST00000579380.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011233.hg.1 |
ENST00000579380.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011233.hg.1 |
ENST00000579768.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011233.hg.1 |
ENST00000579768.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011233.hg.1 |
ENST00000579768.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011233.hg.1 |
NM_016070.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011233.hg.1 |
NM_016070.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011233.hg.1 |
NM_016070.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011233.hg.1 |
NM_016070.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011233.hg.1 |
NM_016070.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011233.hg.1 |
NM_016070.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011233.hg.1 |
NM_016070.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011233.hg.1 |
NM_016070.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011233.hg.1 |
NM_016070.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700190174.hg.1 |
ENST00000313608.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700190174.hg.1 |
ENST00000578444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700190174.hg.1 |
NM_016070.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700190174.hg.1 |
ENST00000313608.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700190174.hg.1 |
ENST00000578444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700190174.hg.1 |
NM_016070.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700190174.hg.1 |
TC1700011233.hg.1 |
Clariom D TC |