Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007352.hg.1 |
ENST00000222248.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007352.hg.1 |
ENST00000222248.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007352.hg.1 |
ENST00000222248.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007352.hg.1 |
ENST00000222248.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007352.hg.1 |
ENST00000222248.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007352.hg.1 |
ENST00000222248.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007352.hg.1 |
ENST00000222248.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007352.hg.1 |
ENST00000222248.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007352.hg.1 |
ENST00000222248.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007352.hg.1 |
ENST00000222248.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007352.hg.1 |
ENST00000222248.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007352.hg.1 |
ENST00000222248.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007352.hg.1 |
ENST00000222248.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007352.hg.1 |
ENST00000222248.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007352.hg.1 |
ENST00000222248.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007352.hg.1 |
ENST00000222248.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007352.hg.1 |
ENST00000222248.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007352.hg.1 |
ENST00000222248.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007352.hg.1 |
ENST00000222248.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007352.hg.1 |
ENST00000222248.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007352.hg.1 |
ENST00000597109.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007352.hg.1 |
ENST00000597109.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007352.hg.1 |
ENST00000597109.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007352.hg.1 |
ENST00000597109.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007352.hg.1 |
ENST00000597109.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007352.hg.1 |
ENST00000597109.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007352.hg.1 |
NM_000453.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007352.hg.1 |
NM_000453.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007352.hg.1 |
NM_000453.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007352.hg.1 |
NM_000453.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007352.hg.1 |
NM_000453.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007352.hg.1 |
NM_000453.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007352.hg.1 |
NM_000453.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007352.hg.1 |
NM_000453.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007352.hg.1 |
NM_000453.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007352.hg.1 |
NM_000453.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007352.hg.1 |
NM_000453.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007352.hg.1 |
NM_000453.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007352.hg.1 |
NM_000453.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007352.hg.1 |
NM_000453.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007352.hg.1 |
NM_000453.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007352.hg.1 |
NM_000453.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007352.hg.1 |
NM_000453.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007352.hg.1 |
NM_000453.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007352.hg.1 |
NM_000453.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007352.hg.1 |
NM_000453.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19004571.hg.1 |
ENST00000222248.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19004571.hg.1 |
NM_000453.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19004571.hg.1 |
TC19000346.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19004572.hg.1 |
ENST00000222248.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19004572.hg.1 |
NM_000453.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19004572.hg.1 |
TC19000346.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19004570.hg.1 |
ENST00000222248.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19004570.hg.1 |
NM_000453.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19004570.hg.1 |
TC19000346.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1900153811.hg.1 |
ENST00000222248.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900153811.hg.1 |
NM_000453.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900153811.hg.1 |
TC1900007352.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900153811.hg.1 |
ENST00000222248.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900153811.hg.1 |
NM_000453.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900153810.hg.1 |
ENST00000222248.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900153810.hg.1 |
NM_000453.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900153810.hg.1 |
TC1900007352.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900153810.hg.1 |
ENST00000222248.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900153810.hg.1 |
NM_000453.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007352.hg.1 |
ENST00000222248.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007352.hg.1 |
ENST00000222248.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007352.hg.1 |
ENST00000222248.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007352.hg.1 |
ENST00000222248.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007352.hg.1 |
ENST00000222248.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007352.hg.1 |
ENST00000222248.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007352.hg.1 |
ENST00000222248.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007352.hg.1 |
ENST00000222248.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007352.hg.1 |
ENST00000222248.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007352.hg.1 |
ENST00000222248.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007352.hg.1 |
ENST00000222248.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007352.hg.1 |
ENST00000222248.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007352.hg.1 |
ENST00000222248.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007352.hg.1 |
ENST00000222248.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007352.hg.1 |
ENST00000222248.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007352.hg.1 |
ENST00000222248.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007352.hg.1 |
ENST00000222248.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007352.hg.1 |
ENST00000222248.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007352.hg.1 |
ENST00000222248.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007352.hg.1 |
ENST00000222248.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007352.hg.1 |
ENST00000597109.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007352.hg.1 |
ENST00000597109.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007352.hg.1 |
ENST00000597109.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007352.hg.1 |
ENST00000597109.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007352.hg.1 |
ENST00000597109.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007352.hg.1 |
ENST00000597109.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007352.hg.1 |
NM_000453.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007352.hg.1 |
NM_000453.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007352.hg.1 |
NM_000453.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007352.hg.1 |
NM_000453.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007352.hg.1 |
NM_000453.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007352.hg.1 |
NM_000453.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007352.hg.1 |
NM_000453.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007352.hg.1 |
NM_000453.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007352.hg.1 |
NM_000453.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007352.hg.1 |
NM_000453.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007352.hg.1 |
NM_000453.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007352.hg.1 |
NM_000453.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007352.hg.1 |
NM_000453.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007352.hg.1 |
NM_000453.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007352.hg.1 |
NM_000453.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007352.hg.1 |
NM_000453.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007352.hg.1 |
NM_000453.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007352.hg.1 |
NM_000453.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007352.hg.1 |
NM_000453.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007352.hg.1 |
NM_000453.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900153812.hg.1 |
ENST00000222248.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900153812.hg.1 |
NM_000453.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900153812.hg.1 |
TC1900007352.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900153812.hg.1 |
ENST00000222248.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900153812.hg.1 |
NM_000453.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900153809.hg.1 |
ENST00000222248.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900153809.hg.1 |
NM_000453.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900153809.hg.1 |
TC1900007352.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900153809.hg.1 |
ENST00000222248.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900153809.hg.1 |
NM_000453.2 |
Refseq |