Assay Details
Gene Symbol: CNNM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020184.3 | NP_064569.3 | 1 - 2 | 1496 | 83 | |
XM_005263914.3 | XP_005263971.1 | 1 - 2 | 1413 | 83 | ||
XM_005263915.3 | XP_005263972.1 | 1 - 2 | 1413 | 83 | ||
XM_011510955.2 | XP_011509257.1 | 1 - 2 | 1413 | 83 | ||
GenBank mRNA | AB046812.1 | - | 1 - 2 | 1226 | 83 | |
AF202777.1 | - | 1 - 2 | 1257 | 83 | ||
BC063295.1 | - | 1 - 2 | 1496 | 30340626 | 83 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC02000585.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
visual perception
magnesium ion homeostasis
magnesium ion transport
sodium ion transmembrane transport
response to stimulus
enamel mineralization
magnesium ion transmembrane transport
sodium ion transmembrane transporter activity
magnesium ion transmembrane transporter activity
visual perception
magnesium ion homeostasis
magnesium ion transport
sodium ion transmembrane transport
response to stimulus
enamel mineralization
magnesium ion transmembrane transport
sodium ion transmembrane transporter activity
magnesium ion transmembrane transporter activity
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR02009025.hg.1 | TC02000585.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02009025.hg.1 | ENST00000377075.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02009025.hg.1 | ENST00000496186.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02009025.hg.1 | ENST00000540067.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02009025.hg.1 | NM_020184.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02009019.hg.1 | TC02000585.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02009019.hg.1 | ENST00000377075.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02009019.hg.1 | NM_020184.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016494.hg.1 | ENST00000377075.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016494.hg.1 | ENST00000377075.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016494.hg.1 | ENST00000377075.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016494.hg.1 | ENST00000377075.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016494.hg.1 | ENST00000377075.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016494.hg.1 | ENST00000377075.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016494.hg.1 | ENST00000377075.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016494.hg.1 | ENST00000377075.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016494.hg.1 | ENST00000377075.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016494.hg.1 | ENST00000377075.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016494.hg.1 | ENST00000377075.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016494.hg.1 | ENST00000377075.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016494.hg.1 | ENST00000377075.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016494.hg.1 | ENST00000377075.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016494.hg.1 | ENST00000482716.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016494.hg.1 | ENST00000482716.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016494.hg.1 | ENST00000482716.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016494.hg.1 | ENST00000482716.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016494.hg.1 | ENST00000482716.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016494.hg.1 | ENST00000482716.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016494.hg.1 | ENST00000493384.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016494.hg.1 | ENST00000493384.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016494.hg.1 | ENST00000493384.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016494.hg.1 | ENST00000493384.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016494.hg.1 | ENST00000493384.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016494.hg.1 | ENST00000493384.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016494.hg.1 | ENST00000496186.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016494.hg.1 | ENST00000496186.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016494.hg.1 | ENST00000496186.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016494.hg.1 | ENST00000496186.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016494.hg.1 | ENST00000496186.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016494.hg.1 | ENST00000496186.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016494.hg.1 | ENST00000496186.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016494.hg.1 | ENST00000496186.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016494.hg.1 | ENST00000496186.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016494.hg.1 | ENST00000630969.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016494.hg.1 | ENST00000630969.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016494.hg.1 | NM_020184.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016494.hg.1 | NM_020184.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016494.hg.1 | NM_020184.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016494.hg.1 | NM_020184.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016494.hg.1 | NM_020184.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016494.hg.1 | NM_020184.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016494.hg.1 | NM_020184.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016494.hg.1 | NM_020184.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016494.hg.1 | NM_020184.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016494.hg.1 | NM_020184.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016494.hg.1 | NM_020184.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016494.hg.1 | NM_020184.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016494.hg.1 | NM_020184.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016494.hg.1 | NM_020184.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200222680.hg.1 | ENST00000377075.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200222680.hg.1 | NM_020184.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200222680.hg.1 | ENST00000377075.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200222680.hg.1 | NM_020184.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200222680.hg.1 | TC0200016494.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0200016494.hg.1 | ENST00000377075.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016494.hg.1 | ENST00000377075.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016494.hg.1 | ENST00000377075.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016494.hg.1 | ENST00000377075.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016494.hg.1 | ENST00000377075.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016494.hg.1 | ENST00000377075.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016494.hg.1 | ENST00000377075.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016494.hg.1 | ENST00000377075.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016494.hg.1 | ENST00000377075.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016494.hg.1 | ENST00000377075.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016494.hg.1 | ENST00000377075.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016494.hg.1 | ENST00000377075.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016494.hg.1 | ENST00000377075.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016494.hg.1 | ENST00000377075.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016494.hg.1 | ENST00000482716.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016494.hg.1 | ENST00000482716.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016494.hg.1 | ENST00000482716.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016494.hg.1 | ENST00000482716.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016494.hg.1 | ENST00000482716.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016494.hg.1 | ENST00000482716.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016494.hg.1 | ENST00000493384.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016494.hg.1 | ENST00000493384.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016494.hg.1 | ENST00000493384.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016494.hg.1 | ENST00000493384.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016494.hg.1 | ENST00000493384.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016494.hg.1 | ENST00000493384.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016494.hg.1 | ENST00000496186.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016494.hg.1 | ENST00000496186.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016494.hg.1 | ENST00000496186.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016494.hg.1 | ENST00000496186.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016494.hg.1 | ENST00000496186.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016494.hg.1 | ENST00000496186.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016494.hg.1 | ENST00000496186.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016494.hg.1 | ENST00000496186.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016494.hg.1 | ENST00000496186.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016494.hg.1 | ENST00000630969.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016494.hg.1 | ENST00000630969.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016494.hg.1 | NM_020184.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016494.hg.1 | NM_020184.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016494.hg.1 | NM_020184.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016494.hg.1 | NM_020184.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016494.hg.1 | NM_020184.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016494.hg.1 | NM_020184.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016494.hg.1 | NM_020184.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016494.hg.1 | NM_020184.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016494.hg.1 | NM_020184.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016494.hg.1 | NM_020184.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016494.hg.1 | NM_020184.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016494.hg.1 | NM_020184.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016494.hg.1 | NM_020184.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016494.hg.1 | NM_020184.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200222685.hg.1 | ENST00000377075.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200222685.hg.1 | ENST00000482716.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200222685.hg.1 | ENST00000493384.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200222685.hg.1 | ENST00000496186.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200222685.hg.1 | NM_020184.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200222685.hg.1 | ENST00000377075.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200222685.hg.1 | ENST00000482716.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200222685.hg.1 | ENST00000493384.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200222685.hg.1 | ENST00000496186.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200222685.hg.1 | NM_020184.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200222685.hg.1 | TC0200016494.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0200222679.hg.1 | ENST00000377075.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200222679.hg.1 | NM_020184.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200222679.hg.1 | ENST00000377075.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200222679.hg.1 | NM_020184.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200222679.hg.1 | TC0200016494.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 113091 | - | - |
