Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02009027.hg.1 |
TC02000585.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02009027.hg.1 |
ENST00000377075.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02009027.hg.1 |
ENST00000496186.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02009027.hg.1 |
ENST00000540067.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02009027.hg.1 |
NM_020184.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02009028.hg.1 |
TC02000585.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02009028.hg.1 |
ENST00000377075.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02009028.hg.1 |
ENST00000496186.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02009028.hg.1 |
ENST00000540067.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02009028.hg.1 |
NM_020184.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016494.hg.1 |
ENST00000377075.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016494.hg.1 |
ENST00000377075.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016494.hg.1 |
ENST00000377075.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016494.hg.1 |
ENST00000377075.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016494.hg.1 |
ENST00000377075.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016494.hg.1 |
ENST00000377075.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016494.hg.1 |
ENST00000377075.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016494.hg.1 |
ENST00000377075.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016494.hg.1 |
ENST00000377075.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016494.hg.1 |
ENST00000377075.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016494.hg.1 |
ENST00000377075.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016494.hg.1 |
ENST00000377075.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016494.hg.1 |
ENST00000377075.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016494.hg.1 |
ENST00000377075.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016494.hg.1 |
ENST00000482716.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016494.hg.1 |
ENST00000482716.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016494.hg.1 |
ENST00000482716.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016494.hg.1 |
ENST00000482716.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016494.hg.1 |
ENST00000482716.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016494.hg.1 |
ENST00000482716.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016494.hg.1 |
ENST00000493384.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016494.hg.1 |
ENST00000493384.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016494.hg.1 |
ENST00000493384.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016494.hg.1 |
ENST00000493384.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016494.hg.1 |
ENST00000493384.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016494.hg.1 |
ENST00000493384.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016494.hg.1 |
ENST00000496186.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016494.hg.1 |
ENST00000496186.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016494.hg.1 |
ENST00000496186.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016494.hg.1 |
ENST00000496186.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016494.hg.1 |
ENST00000496186.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016494.hg.1 |
ENST00000496186.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016494.hg.1 |
ENST00000496186.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016494.hg.1 |
ENST00000496186.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016494.hg.1 |
ENST00000496186.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016494.hg.1 |
ENST00000630969.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016494.hg.1 |
ENST00000630969.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016494.hg.1 |
NM_020184.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016494.hg.1 |
NM_020184.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016494.hg.1 |
NM_020184.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016494.hg.1 |
NM_020184.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016494.hg.1 |
NM_020184.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016494.hg.1 |
NM_020184.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016494.hg.1 |
NM_020184.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016494.hg.1 |
NM_020184.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016494.hg.1 |
NM_020184.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016494.hg.1 |
NM_020184.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016494.hg.1 |
NM_020184.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016494.hg.1 |
NM_020184.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016494.hg.1 |
NM_020184.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016494.hg.1 |
NM_020184.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200222688.hg.1 |
ENST00000377075.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200222688.hg.1 |
ENST00000493384.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200222688.hg.1 |
ENST00000496186.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200222688.hg.1 |
NM_020184.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200222688.hg.1 |
ENST00000377075.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200222688.hg.1 |
ENST00000493384.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200222688.hg.1 |
ENST00000496186.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200222688.hg.1 |
NM_020184.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200222688.hg.1 |
TC0200016494.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200222687.hg.1 |
ENST00000377075.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200222687.hg.1 |
ENST00000493384.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200222687.hg.1 |
ENST00000496186.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200222687.hg.1 |
NM_020184.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200222687.hg.1 |
ENST00000377075.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200222687.hg.1 |
ENST00000493384.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200222687.hg.1 |
ENST00000496186.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200222687.hg.1 |
NM_020184.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200222687.hg.1 |
TC0200016494.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200016494.hg.1 |
ENST00000377075.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016494.hg.1 |
ENST00000377075.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016494.hg.1 |
ENST00000377075.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016494.hg.1 |
ENST00000377075.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016494.hg.1 |
ENST00000377075.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016494.hg.1 |
ENST00000377075.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016494.hg.1 |
ENST00000377075.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016494.hg.1 |
ENST00000377075.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016494.hg.1 |
ENST00000377075.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016494.hg.1 |
ENST00000377075.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016494.hg.1 |
ENST00000377075.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016494.hg.1 |
ENST00000377075.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016494.hg.1 |
ENST00000377075.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016494.hg.1 |
ENST00000377075.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016494.hg.1 |
ENST00000482716.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016494.hg.1 |
ENST00000482716.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016494.hg.1 |
ENST00000482716.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016494.hg.1 |
ENST00000482716.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016494.hg.1 |
ENST00000482716.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016494.hg.1 |
ENST00000482716.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016494.hg.1 |
ENST00000493384.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016494.hg.1 |
ENST00000493384.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016494.hg.1 |
ENST00000493384.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016494.hg.1 |
ENST00000493384.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016494.hg.1 |
ENST00000493384.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016494.hg.1 |
ENST00000493384.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016494.hg.1 |
ENST00000496186.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016494.hg.1 |
ENST00000496186.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016494.hg.1 |
ENST00000496186.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016494.hg.1 |
ENST00000496186.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016494.hg.1 |
ENST00000496186.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016494.hg.1 |
ENST00000496186.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016494.hg.1 |
ENST00000496186.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016494.hg.1 |
ENST00000496186.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016494.hg.1 |
ENST00000496186.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016494.hg.1 |
ENST00000630969.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016494.hg.1 |
ENST00000630969.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016494.hg.1 |
NM_020184.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016494.hg.1 |
NM_020184.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016494.hg.1 |
NM_020184.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016494.hg.1 |
NM_020184.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016494.hg.1 |
NM_020184.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016494.hg.1 |
NM_020184.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016494.hg.1 |
NM_020184.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016494.hg.1 |
NM_020184.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016494.hg.1 |
NM_020184.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016494.hg.1 |
NM_020184.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016494.hg.1 |
NM_020184.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016494.hg.1 |
NM_020184.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016494.hg.1 |
NM_020184.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016494.hg.1 |
NM_020184.3 |
Refseq |