Assay Details
| Gene Symbol: XPNPEP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003399.5 | NP_003390.4 | 8 - 9 | 985 | 58 | |
| GenBank mRNA | BC126174.1 | - | 8 - 9 | 902 | 8991890 | 58 |
| BC143901.1 | - | 8 - 9 | 902 | 9052419 | 58 | |
| BF830085.1 | - | 2 - 3 | 230 | 58 | ||
| U90724.2 | - | 8 - 9 | 1005 | 58 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X000617.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR0X008111.hg.1 | TC0X000617.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X008111.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X008111.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371105.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371105.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371105.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371105.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371105.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371105.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371105.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371105.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371105.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371105.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00157095.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00157095.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00157095.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00157095.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00157095.hg.1 | TC0X00008387.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371105.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371105.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371105.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371105.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371105.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371105.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371105.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371105.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371105.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371105.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | NM_003399.5 | Refseq |

XPNPEP2
Genomic Map