Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08018450.hg.1 |
TC08001423.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08018450.hg.1 |
ENST00000378861.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08018450.hg.1 |
ENST00000517858.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08018450.hg.1 |
ENST00000518748.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08018450.hg.1 |
ENST00000519069.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08018450.hg.1 |
ENST00000519969.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08018450.hg.1 |
ENST00000520430.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08018450.hg.1 |
ENST00000520686.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08018450.hg.1 |
ENST00000521617.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08018450.hg.1 |
ENST00000521988.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08018450.hg.1 |
ENST00000523580.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08018450.hg.1 |
ENST00000524037.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08018450.hg.1 |
ENST00000524107.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08018450.hg.1 |
ENST00000537541.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08018450.hg.1 |
NM_001171795.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08018450.hg.1 |
NM_001171796.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08018450.hg.1 |
NM_001171797.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08018450.hg.1 |
NM_001171798.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08018450.hg.1 |
NM_001171799.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08018450.hg.1 |
NM_001191035.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08018450.hg.1 |
NM_001191036.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08018447.hg.1 |
TC08001423.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08018447.hg.1 |
ENST00000378861.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08018447.hg.1 |
ENST00000517540.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08018447.hg.1 |
ENST00000517858.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08018447.hg.1 |
ENST00000518748.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08018447.hg.1 |
ENST00000519069.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08018447.hg.1 |
ENST00000519969.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08018447.hg.1 |
ENST00000520430.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08018447.hg.1 |
ENST00000520686.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08018447.hg.1 |
ENST00000521617.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08018447.hg.1 |
ENST00000521988.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08018447.hg.1 |
ENST00000523580.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08018447.hg.1 |
ENST00000524037.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08018447.hg.1 |
ENST00000524107.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08018447.hg.1 |
ENST00000537541.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08018447.hg.1 |
NM_001171795.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08018447.hg.1 |
NM_001171796.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08018447.hg.1 |
NM_001171797.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08018447.hg.1 |
NM_001171798.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08018447.hg.1 |
NM_001171799.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08018447.hg.1 |
NM_001191035.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08018447.hg.1 |
NM_001191036.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175626.hg.1 |
ENST00000378861.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175626.hg.1 |
ENST00000517751.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175626.hg.1 |
ENST00000517858.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175626.hg.1 |
ENST00000518748.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175626.hg.1 |
ENST00000519069.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175626.hg.1 |
ENST00000519969.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175626.hg.1 |
ENST00000520430.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175626.hg.1 |
ENST00000520686.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175626.hg.1 |
ENST00000521617.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175626.hg.1 |
ENST00000521988.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175626.hg.1 |
ENST00000522925.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175626.hg.1 |
ENST00000523580.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175626.hg.1 |
ENST00000524037.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175626.hg.1 |
ENST00000524107.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175626.hg.1 |
ENST00000537541.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175626.hg.1 |
NM_001171795.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175626.hg.1 |
NM_001171796.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175626.hg.1 |
NM_001171797.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175626.hg.1 |
NM_001171798.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175626.hg.1 |
NM_001171799.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175626.hg.1 |
NM_001191035.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175626.hg.1 |
NM_001191036.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175626.hg.1 |
ENST00000378861.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175626.hg.1 |
ENST00000517751.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175626.hg.1 |
ENST00000517858.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175626.hg.1 |
ENST00000518748.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175626.hg.1 |
ENST00000519069.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175626.hg.1 |
ENST00000519969.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175626.hg.1 |
ENST00000520430.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175626.hg.1 |
ENST00000520686.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175626.hg.1 |
ENST00000521617.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175626.hg.1 |
ENST00000521988.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175626.hg.1 |
ENST00000522925.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175626.hg.1 |
ENST00000523580.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175626.hg.1 |
ENST00000524037.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175626.hg.1 |
ENST00000524107.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175626.hg.1 |
ENST00000537541.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175626.hg.1 |
NM_001171795.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175626.hg.1 |
NM_001171796.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175626.hg.1 |
NM_001171797.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175626.hg.1 |
NM_001171798.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175626.hg.1 |
NM_001171799.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175626.hg.1 |
NM_001191035.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175626.hg.1 |
NM_001191036.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175626.hg.1 |
TC0800011105.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800175623.hg.1 |
ENST00000378861.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175623.hg.1 |
ENST00000517540.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175623.hg.1 |
ENST00000517751.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175623.hg.1 |
ENST00000517858.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175623.hg.1 |
ENST00000518400.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175623.hg.1 |
ENST00000518748.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175623.hg.1 |
ENST00000519069.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175623.hg.1 |
ENST00000519969.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175623.hg.1 |
ENST00000520384.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175623.hg.1 |
ENST00000520430.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175623.hg.1 |
ENST00000520686.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175623.hg.1 |
ENST00000521617.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175623.hg.1 |
ENST00000521988.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175623.hg.1 |
ENST00000522925.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175623.hg.1 |
ENST00000523580.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175623.hg.1 |
ENST00000524037.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175623.hg.1 |
ENST00000524107.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175623.hg.1 |
ENST00000537541.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175623.hg.1 |
NM_001171795.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175623.hg.1 |
NM_001171796.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175623.hg.1 |
NM_001171797.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175623.hg.1 |
NM_001171798.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175623.hg.1 |
NM_001171799.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175623.hg.1 |
NM_001191035.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175623.hg.1 |
NM_001191036.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175623.hg.1 |
ENST00000378861.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175623.hg.1 |
ENST00000517540.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175623.hg.1 |
ENST00000517751.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175623.hg.1 |
ENST00000517858.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175623.hg.1 |
ENST00000518400.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175623.hg.1 |
ENST00000518748.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175623.hg.1 |
ENST00000519069.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175623.hg.1 |
ENST00000519969.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175623.hg.1 |
ENST00000520384.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175623.hg.1 |
ENST00000520430.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175623.hg.1 |
ENST00000520686.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175623.hg.1 |
ENST00000521617.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175623.hg.1 |
ENST00000521988.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175623.hg.1 |
ENST00000522925.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175623.hg.1 |
ENST00000523580.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175623.hg.1 |
ENST00000524037.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175623.hg.1 |
ENST00000524107.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175623.hg.1 |
ENST00000537541.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175623.hg.1 |
NM_001171795.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175623.hg.1 |
NM_001171796.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175623.hg.1 |
NM_001171797.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175623.hg.1 |
NM_001171798.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175623.hg.1 |
NM_001171799.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175623.hg.1 |
NM_001191035.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175623.hg.1 |
NM_001191036.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175623.hg.1 |
TC0800011105.hg.1 |
Clariom D TC |