Assay Details
Gene Symbol: HSPE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_002157.2 | NP_002148.1 | 3 - 4 | 695 | 133 | |
GenBank mRNA | BC023518.2 | - | 3 - 4 | 369 | 4304224 | 133 |
BG188453.1 | - | 2 - 3 | 256 | 133 | ||
U07550.1 | - | 3 - 4 | 296 | 133 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC02005004.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
osteoblast differentiation
protein folding
activation of cysteine-type endopeptidase activity involved in apoptotic process
response to unfolded protein
chaperone mediated protein folding requiring cofactor
protein binding
ATP binding
poly(A) RNA binding
metal ion binding
unfolded protein binding
chaperone binding
osteoblast differentiation
protein folding
activation of cysteine-type endopeptidase activity involved in apoptotic process
response to unfolded protein
chaperone mediated protein folding requiring cofactor
protein binding
ATP binding
poly(A) RNA binding
metal ion binding
unfolded protein binding
chaperone binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR02017352.hg.1 | TC02005004.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02017352.hg.1 | ENST00000233893.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02017352.hg.1 | ENST00000409468.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02017352.hg.1 | ENST00000409729.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02017352.hg.1 | ENST00000465573.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02017352.hg.1 | NM_002157.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02017354.hg.1 | TC02005004.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02017354.hg.1 | ENST00000233893.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02017354.hg.1 | ENST00000409468.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02017354.hg.1 | ENST00000409729.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02017354.hg.1 | ENST00000465573.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02017354.hg.1 | NM_002157.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007867.hg.1 | ENST00000509838.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007867.hg.1 | ENST00000509838.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000233893.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000233893.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000233893.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000233893.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000233893.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000233893.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000233893.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000233893.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000233893.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000233893.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000233893.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000233893.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000233893.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000409468.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000409468.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000409468.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000409468.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000409468.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000409468.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000409468.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000409468.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000409468.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000409729.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000409729.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000409729.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000409729.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000409729.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000409729.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000409729.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000463841.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000463841.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000463841.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000463841.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000463841.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000463841.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000463841.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000463841.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000463841.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000463841.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000465573.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000465573.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000465573.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000465573.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000465573.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000465573.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000465573.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000473395.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000473395.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000473395.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000473395.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000473395.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000495200.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000495200.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000495200.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | ENST00000495200.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | NM_002157.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | NM_002157.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | NM_002157.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | NM_002157.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | NM_002157.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | NM_002157.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | NM_002157.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | NM_002157.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | NM_002157.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | NM_002157.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | NM_002157.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | NM_002157.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016584.hg.1 | NM_002157.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000233893.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000233893.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000233893.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000233893.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000233893.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000233893.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000233893.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000233893.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000233893.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000233893.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000233893.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000233893.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000233893.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000409468.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000409468.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000409468.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000409468.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000409468.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000409468.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000409468.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000409468.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000409468.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000409729.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000409729.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000409729.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000409729.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000409729.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000409729.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000409729.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000463841.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000463841.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000463841.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000463841.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000463841.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000463841.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000463841.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000463841.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000463841.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000463841.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000465573.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000465573.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000465573.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000465573.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000465573.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000465573.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000465573.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000473395.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000473395.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000473395.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000473395.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000473395.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000495200.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000495200.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000495200.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | ENST00000495200.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | NM_002157.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | NM_002157.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | NM_002157.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | NM_002157.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | NM_002157.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | NM_002157.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | NM_002157.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | NM_002157.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | NM_002157.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | NM_002157.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | NM_002157.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | NM_002157.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016584.hg.1 | NM_002157.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200224577.hg.1 | ENST00000233893.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200224577.hg.1 | ENST00000409468.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200224577.hg.1 | ENST00000409729.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200224577.hg.1 | ENST00000463841.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200224577.hg.1 | ENST00000465573.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200224577.hg.1 | NM_002157.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200224577.hg.1 | ENST00000233893.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200224577.hg.1 | ENST00000409468.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200224577.hg.1 | ENST00000409729.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200224577.hg.1 | ENST00000463841.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200224577.hg.1 | ENST00000465573.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200224577.hg.1 | NM_002157.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200224577.hg.1 | TC0200016584.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0200224579.hg.1 | ENST00000233893.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200224579.hg.1 | ENST00000409468.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200224579.hg.1 | ENST00000409729.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200224579.hg.1 | ENST00000463841.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200224579.hg.1 | ENST00000465573.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200224579.hg.1 | NM_002157.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200224579.hg.1 | ENST00000233893.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200224579.hg.1 | ENST00000409468.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200224579.hg.1 | ENST00000409729.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200224579.hg.1 | ENST00000463841.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200224579.hg.1 | ENST00000465573.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200224579.hg.1 | NM_002157.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200224579.hg.1 | TC0200016584.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0400007867.hg.1 | ENST00000509838.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007867.hg.1 | ENST00000509838.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200224580.hg.1 | ENST00000233893.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200224580.hg.1 | ENST00000409468.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200224580.hg.1 | ENST00000409729.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200224580.hg.1 | ENST00000463841.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200224580.hg.1 | ENST00000465573.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200224580.hg.1 | NM_002157.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200224580.hg.1 | ENST00000233893.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200224580.hg.1 | ENST00000409468.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200224580.hg.1 | ENST00000409729.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200224580.hg.1 | ENST00000463841.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200224580.hg.1 | ENST00000465573.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200224580.hg.1 | NM_002157.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200224580.hg.1 | TC0200016584.hg.1 | Clariom D TC |
