Assay Details
| Gene Symbol: PLEKHD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161498.1 | NP_001154970.1 | 8 - 9 | 955 | 68 | |
| XM_011536762.2 | XP_011535064.1 | 5 - 6 | 401 | 68 | ||
| XM_011536763.2 | XP_011535065.1 | 3 - 4 | 261 | 68 | ||
| XM_017021290.1 | XP_016876779.1 | 8 - 9 | 1317 | 68 | ||
| GenBank mRNA | BC146872.1 | - | 8 - 9 | 860 | 9056800 | 68 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC14000416.hg.1
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1400007555.hg.1 | ENST00000322564.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007555.hg.1 | ENST00000322564.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007555.hg.1 | ENST00000322564.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007555.hg.1 | ENST00000322564.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007555.hg.1 | ENST00000322564.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007555.hg.1 | ENST00000322564.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007555.hg.1 | ENST00000322564.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007555.hg.1 | ENST00000322564.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007555.hg.1 | ENST00000322564.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007555.hg.1 | ENST00000322564.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007555.hg.1 | ENST00000322564.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007555.hg.1 | ENST00000322564.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007555.hg.1 | ENST00000322564.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007555.hg.1 | ENST00000322564.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007555.hg.1 | ENST00000322564.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007555.hg.1 | ENST00000322564.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007555.hg.1 | ENST00000322564.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007555.hg.1 | ENST00000322564.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007555.hg.1 | ENST00000556123.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007555.hg.1 | ENST00000556123.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007555.hg.1 | ENST00000556123.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007555.hg.1 | ENST00000556123.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007555.hg.1 | ENST00000556123.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007555.hg.1 | ENST00000556123.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007555.hg.1 | ENST00000556123.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007555.hg.1 | ENST00000556123.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007555.hg.1 | ENST00000556123.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007555.hg.1 | ENST00000556123.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007555.hg.1 | ENST00000556123.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007555.hg.1 | ENST00000556123.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007555.hg.1 | ENST00000556123.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007555.hg.1 | ENST00000556123.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007555.hg.1 | NM_001161498.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007555.hg.1 | NM_001161498.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007555.hg.1 | NM_001161498.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007555.hg.1 | NM_001161498.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007555.hg.1 | NM_001161498.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007555.hg.1 | NM_001161498.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007555.hg.1 | NM_001161498.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007555.hg.1 | NM_001161498.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007555.hg.1 | NM_001161498.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007555.hg.1 | NM_001161498.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007555.hg.1 | NM_001161498.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007555.hg.1 | NM_001161498.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007555.hg.1 | NM_001161498.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007555.hg.1 | NM_001161498.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007555.hg.1 | NM_001161498.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007555.hg.1 | NM_001161498.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007555.hg.1 | NM_001161498.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14005026.hg.1 | ENST00000322564.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14005026.hg.1 | NM_001161498.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14005026.hg.1 | TC14000416.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1400152975.hg.1 | ENST00000322564.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400152975.hg.1 | NM_001161498.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400152975.hg.1 | ENST00000322564.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400152975.hg.1 | NM_001161498.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400152975.hg.1 | TC1400007555.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1400007555.hg.1 | ENST00000322564.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007555.hg.1 | ENST00000322564.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007555.hg.1 | ENST00000322564.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007555.hg.1 | ENST00000322564.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007555.hg.1 | ENST00000322564.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007555.hg.1 | ENST00000322564.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007555.hg.1 | ENST00000322564.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007555.hg.1 | ENST00000322564.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007555.hg.1 | ENST00000322564.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007555.hg.1 | ENST00000322564.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007555.hg.1 | ENST00000322564.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007555.hg.1 | ENST00000322564.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007555.hg.1 | ENST00000322564.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007555.hg.1 | ENST00000322564.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007555.hg.1 | ENST00000322564.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007555.hg.1 | ENST00000322564.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007555.hg.1 | ENST00000322564.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007555.hg.1 | ENST00000322564.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007555.hg.1 | ENST00000556123.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007555.hg.1 | ENST00000556123.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007555.hg.1 | ENST00000556123.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007555.hg.1 | ENST00000556123.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007555.hg.1 | ENST00000556123.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007555.hg.1 | ENST00000556123.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007555.hg.1 | ENST00000556123.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007555.hg.1 | ENST00000556123.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007555.hg.1 | ENST00000556123.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007555.hg.1 | ENST00000556123.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007555.hg.1 | ENST00000556123.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007555.hg.1 | ENST00000556123.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007555.hg.1 | ENST00000556123.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007555.hg.1 | ENST00000556123.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007555.hg.1 | NM_001161498.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007555.hg.1 | NM_001161498.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007555.hg.1 | NM_001161498.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007555.hg.1 | NM_001161498.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007555.hg.1 | NM_001161498.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007555.hg.1 | NM_001161498.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007555.hg.1 | NM_001161498.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007555.hg.1 | NM_001161498.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007555.hg.1 | NM_001161498.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007555.hg.1 | NM_001161498.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007555.hg.1 | NM_001161498.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007555.hg.1 | NM_001161498.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007555.hg.1 | NM_001161498.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007555.hg.1 | NM_001161498.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007555.hg.1 | NM_001161498.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007555.hg.1 | NM_001161498.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007555.hg.1 | NM_001161498.1 | Refseq |

PLEKHD1
Genomic Map