Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01010994.hg.1 |
TC01000685.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01010994.hg.1 |
ENST00000371244.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01010994.hg.1 |
NM_006252.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008450.hg.1 |
ENST00000371244.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008450.hg.1 |
ENST00000371244.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008450.hg.1 |
ENST00000371244.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008450.hg.1 |
ENST00000371244.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008450.hg.1 |
ENST00000371244.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008450.hg.1 |
ENST00000371244.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008450.hg.1 |
ENST00000371244.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008450.hg.1 |
ENST00000371244.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008450.hg.1 |
ENST00000371244.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008450.hg.1 |
ENST00000371244.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008450.hg.1 |
ENST00000371244.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008450.hg.1 |
ENST00000371244.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008450.hg.1 |
ENST00000371244.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008450.hg.1 |
ENST00000371244.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008450.hg.1 |
ENST00000610361.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008450.hg.1 |
ENST00000610361.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008450.hg.1 |
ENST00000610361.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008450.hg.1 |
ENST00000610361.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008450.hg.1 |
ENST00000610361.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008450.hg.1 |
ENST00000610361.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008450.hg.1 |
ENST00000610361.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008450.hg.1 |
ENST00000610361.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008450.hg.1 |
ENST00000610361.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008450.hg.1 |
ENST00000610361.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008450.hg.1 |
ENST00000610361.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008450.hg.1 |
ENST00000610361.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008450.hg.1 |
NM_006252.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008450.hg.1 |
NM_006252.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008450.hg.1 |
NM_006252.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008450.hg.1 |
NM_006252.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008450.hg.1 |
NM_006252.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008450.hg.1 |
NM_006252.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008450.hg.1 |
NM_006252.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008450.hg.1 |
NM_006252.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008450.hg.1 |
NM_006252.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008450.hg.1 |
NM_006252.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008450.hg.1 |
NM_006252.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008450.hg.1 |
NM_006252.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008450.hg.1 |
NM_006252.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008450.hg.1 |
NM_006252.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01010995.hg.1 |
TC01000685.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01010995.hg.1 |
ENST00000371244.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01010995.hg.1 |
NM_006252.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100160063.hg.1 |
ENST00000371244.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100160063.hg.1 |
NM_006252.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100160063.hg.1 |
ENST00000371244.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100160063.hg.1 |
NM_006252.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100160063.hg.1 |
TC0100008450.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100008450.hg.1 |
ENST00000371244.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008450.hg.1 |
ENST00000371244.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008450.hg.1 |
ENST00000371244.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008450.hg.1 |
ENST00000371244.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008450.hg.1 |
ENST00000371244.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008450.hg.1 |
ENST00000371244.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008450.hg.1 |
ENST00000371244.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008450.hg.1 |
ENST00000371244.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008450.hg.1 |
ENST00000371244.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008450.hg.1 |
ENST00000371244.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008450.hg.1 |
ENST00000371244.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008450.hg.1 |
ENST00000371244.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008450.hg.1 |
ENST00000371244.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008450.hg.1 |
ENST00000371244.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008450.hg.1 |
ENST00000610361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008450.hg.1 |
ENST00000610361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008450.hg.1 |
ENST00000610361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008450.hg.1 |
ENST00000610361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008450.hg.1 |
ENST00000610361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008450.hg.1 |
ENST00000610361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008450.hg.1 |
ENST00000610361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008450.hg.1 |
ENST00000610361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008450.hg.1 |
ENST00000610361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008450.hg.1 |
ENST00000610361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008450.hg.1 |
ENST00000610361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008450.hg.1 |
ENST00000610361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008450.hg.1 |
NM_006252.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008450.hg.1 |
NM_006252.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008450.hg.1 |
NM_006252.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008450.hg.1 |
NM_006252.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008450.hg.1 |
NM_006252.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008450.hg.1 |
NM_006252.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008450.hg.1 |
NM_006252.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008450.hg.1 |
NM_006252.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008450.hg.1 |
NM_006252.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008450.hg.1 |
NM_006252.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008450.hg.1 |
NM_006252.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008450.hg.1 |
NM_006252.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008450.hg.1 |
NM_006252.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008450.hg.1 |
NM_006252.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100160062.hg.1 |
ENST00000371244.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100160062.hg.1 |
NM_006252.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100160062.hg.1 |
ENST00000371244.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100160062.hg.1 |
NM_006252.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100160062.hg.1 |
TC0100008450.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100160064.hg.1 |
ENST00000371244.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100160064.hg.1 |
ENST00000610361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100160064.hg.1 |
NM_006252.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100160064.hg.1 |
ENST00000371244.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100160064.hg.1 |
ENST00000610361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100160064.hg.1 |
NM_006252.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100160064.hg.1 |
TC0100008450.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100160067.hg.1 |
ENST00000371244.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100160067.hg.1 |
ENST00000610361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100160067.hg.1 |
NM_006252.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100160067.hg.1 |
ENST00000371244.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100160067.hg.1 |
ENST00000610361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100160067.hg.1 |
NM_006252.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100160067.hg.1 |
TC0100008450.hg.1 |
Clariom D TC |