Assay Details
| Gene Symbol: PAPOLA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252006.1 | NP_001238935.1 | 4 - 5 | 548 | 154 | |
| NM_001252007.1 | NP_001238936.1 | 4 - 5 | 548 | 154 | ||
| NM_001293627.1 | NP_001280556.1 | 4 - 5 | 548 | 154 | ||
| NM_001293628.1 | NP_001280557.1 | 4 - 5 | 413 | 154 | ||
| NM_032632.4 | NP_116021.2 | 4 - 5 | 548 | 154 | ||
| XM_005267281.1 | XP_005267338.1 | 4 - 5 | 544 | 154 | ||
| XM_005267282.2 | XP_005267339.1 | 4 - 5 | 544 | 154 | ||
| XM_017020935.1 | XP_016876424.1 | 4 - 5 | 544 | 154 | ||
| XM_017020936.1 | XP_016876425.1 | 4 - 5 | 544 | 154 | ||
| GenBank mRNA | AK295020.1 | - | 4 - 5 | 379 | 154 | |
| AK302406.1 | - | 5 - 6 | 409 | 154 | ||
| BC000927.1 | - | 4 - 5 | 499 | 154 | ||
| BC036014.1 | - | 4 - 5 | 543 | 5271668 | 154 | |
| BX161482.1 | - | 4 - 5 | 483 | 154 | ||
| BX248301.1 | - | 4 - 5 | 413 | 154 | ||
| BX248753.1 | - | 4 - 5 | 523 | 154 | ||
| X76770.1 | - | 3 - 4 | 307 | 154 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001293628.1 transcripts.
Set Membership: |
|

PAPOLA
Genomic Map