Assay Details
| Gene Symbol: YIPF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005264599.3 | XP_005264656.1 | 5 - 6 | 865 | 123 | |
More Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200007181.hg.1 | ENST00000238831.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007181.hg.1 | ENST00000238831.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007181.hg.1 | ENST00000238831.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007181.hg.1 | ENST00000238831.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007181.hg.1 | ENST00000238831.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007181.hg.1 | ENST00000238831.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007181.hg.1 | ENST00000238831.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007181.hg.1 | ENST00000238831.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007181.hg.1 | ENST00000238831.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007181.hg.1 | ENST00000238831.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007181.hg.1 | ENST00000238831.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007181.hg.1 | ENST00000238831.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007181.hg.1 | ENST00000238831.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007181.hg.1 | ENST00000238831.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007181.hg.1 | ENST00000238831.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007181.hg.1 | ENST00000238831.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007181.hg.1 | ENST00000238831.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007181.hg.1 | ENST00000238831.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007181.hg.1 | ENST00000238831.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007181.hg.1 | ENST00000437765.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007181.hg.1 | ENST00000437765.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007181.hg.1 | ENST00000437765.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007181.hg.1 | ENST00000437765.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007181.hg.1 | ENST00000437765.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007181.hg.1 | ENST00000437765.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007181.hg.1 | ENST00000441084.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007181.hg.1 | ENST00000441084.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007181.hg.1 | ENST00000495355.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007181.hg.1 | ENST00000495355.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007181.hg.1 | ENST00000495355.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007181.hg.1 | ENST00000495355.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007181.hg.1 | NM_032312.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007181.hg.1 | NM_032312.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007181.hg.1 | NM_032312.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007181.hg.1 | NM_032312.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007181.hg.1 | NM_032312.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007181.hg.1 | NM_032312.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007181.hg.1 | NM_032312.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007181.hg.1 | NM_032312.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007181.hg.1 | NM_032312.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007181.hg.1 | NM_032312.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007181.hg.1 | NM_032312.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007181.hg.1 | NM_032312.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007181.hg.1 | ENST00000238831.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007181.hg.1 | ENST00000238831.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007181.hg.1 | ENST00000238831.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007181.hg.1 | ENST00000238831.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007181.hg.1 | ENST00000238831.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007181.hg.1 | ENST00000238831.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007181.hg.1 | ENST00000238831.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007181.hg.1 | ENST00000238831.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007181.hg.1 | ENST00000238831.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007181.hg.1 | ENST00000238831.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007181.hg.1 | ENST00000238831.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007181.hg.1 | ENST00000238831.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007181.hg.1 | ENST00000238831.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007181.hg.1 | ENST00000238831.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007181.hg.1 | ENST00000238831.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007181.hg.1 | ENST00000238831.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007181.hg.1 | ENST00000238831.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007181.hg.1 | ENST00000238831.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007181.hg.1 | ENST00000238831.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007181.hg.1 | ENST00000437765.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007181.hg.1 | ENST00000437765.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007181.hg.1 | ENST00000437765.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007181.hg.1 | ENST00000437765.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007181.hg.1 | ENST00000437765.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007181.hg.1 | ENST00000437765.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007181.hg.1 | ENST00000441084.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007181.hg.1 | ENST00000441084.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007181.hg.1 | ENST00000495355.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007181.hg.1 | ENST00000495355.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007181.hg.1 | ENST00000495355.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007181.hg.1 | ENST00000495355.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007181.hg.1 | NM_032312.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007181.hg.1 | NM_032312.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007181.hg.1 | NM_032312.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007181.hg.1 | NM_032312.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007181.hg.1 | NM_032312.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007181.hg.1 | NM_032312.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007181.hg.1 | NM_032312.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007181.hg.1 | NM_032312.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007181.hg.1 | NM_032312.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007181.hg.1 | NM_032312.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007181.hg.1 | NM_032312.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007181.hg.1 | NM_032312.3 | Refseq |

YIPF4
Genomic Map