Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06006135.hg.1 |
TC06000556.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06006135.hg.1 |
ENST00000359357.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06006135.hg.1 |
ENST00000441566.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06006135.hg.1 |
ENST00000449981.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06006135.hg.1 |
NM_001206927.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154982.hg.1 |
ENST00000327475.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154982.hg.1 |
ENST00000359357.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154982.hg.1 |
ENST00000449981.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154982.hg.1 |
NM_001206927.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154982.hg.1 |
ENST00000327475.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154982.hg.1 |
ENST00000359357.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154982.hg.1 |
ENST00000449981.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154982.hg.1 |
NM_001206927.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154982.hg.1 |
TC0600007940.hg.1 |
Clariom D TC |