Assay Details
| Gene Symbol: ZCCHC17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282569.1 | NP_001269498.1 | 4 - 5 | 339 | 152 | |
| NM_001282573.1 | NP_001269502.1 | 4 - 5 | 339 | 152 | ||
| XM_006710681.3 | XP_006710744.1 | 4 - 5 | 413 | 152 | ||
| GenBank mRNA | AK302248.1 | - | 4 - 5 | 289 | 152 | |
| DA737244.1 | - | 4 - 5 | 289 | 152 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK302248.1, NM_001282569.1, NM_001282573.1, XM_006710681.3 transcripts.
Set Membership: |
|

ZCCHC17
Genomic Map