Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02045367.hg.1 |
TC02002808.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02045367.hg.1 |
ENST00000281821.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02045367.hg.1 |
ENST00000392071.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02045367.hg.1 |
ENST00000409854.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02045367.hg.1 |
ENST00000409938.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02045367.hg.1 |
NM_004438.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200216405.hg.1 |
ENST00000281821.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200216405.hg.1 |
ENST00000409854.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200216405.hg.1 |
ENST00000409938.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200216405.hg.1 |
NM_001304536.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200216405.hg.1 |
NM_001304537.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200216405.hg.1 |
NM_004438.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200216405.hg.1 |
ENST00000281821.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200216405.hg.1 |
ENST00000409854.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200216405.hg.1 |
ENST00000409938.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200216405.hg.1 |
NM_001304536.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200216405.hg.1 |
NM_001304537.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200216405.hg.1 |
NM_004438.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200216405.hg.1 |
TC0200015815.hg.1 |
Clariom D TC |