Assay Details
Gene Symbol: KRT84 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033045.3 | NP_149034.2 | 2 - 3 | 804 | 63 | |
XM_011538335.2 | XP_011536637.1 | 3 - 4 | 919 | 63 | ||
GenBank mRNA | AJ628420.1 | - | 2 - 3 | 750 | 63 | |
AK314030.1 | - | 2 - 3 | 823 | 63 | ||
BC069647.1 | - | 2 - 3 | 804 | 7262449 | 63 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC12001520.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR12019959.hg.1 | ENST00000257951.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12019959.hg.1 | NM_033045.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12019959.hg.1 | TC12001520.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1200010752.hg.1 | ENST00000257951.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010752.hg.1 | ENST00000257951.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010752.hg.1 | ENST00000257951.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010752.hg.1 | ENST00000257951.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010752.hg.1 | ENST00000257951.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010752.hg.1 | ENST00000257951.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010752.hg.1 | ENST00000257951.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010752.hg.1 | ENST00000257951.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010752.hg.1 | ENST00000257951.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010752.hg.1 | ENST00000257951.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010752.hg.1 | ENST00000257951.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010752.hg.1 | NM_033045.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010752.hg.1 | NM_033045.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010752.hg.1 | NM_033045.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010752.hg.1 | NM_033045.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010752.hg.1 | NM_033045.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010752.hg.1 | NM_033045.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010752.hg.1 | NM_033045.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010752.hg.1 | NM_033045.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010752.hg.1 | NM_033045.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010752.hg.1 | NM_033045.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010752.hg.1 | NM_033045.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12019961.hg.1 | ENST00000257951.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12019961.hg.1 | NM_033045.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12019961.hg.1 | TC12001520.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR12019960.hg.1 | ENST00000257951.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12019960.hg.1 | NM_033045.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12019960.hg.1 | TC12001520.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR12019962.hg.1 | ENST00000257951.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12019962.hg.1 | NM_033045.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12019962.hg.1 | TC12001520.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1200010745.hg.1 | ENST00000529785.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010745.hg.1 | ENST00000529785.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010745.hg.1 | ENST00000529785.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010745.hg.1 | ENST00000529785.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010745.hg.1 | ENST00000529785.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010745.hg.1 | ENST00000529785.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010745.hg.1 | ENST00000529785.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010745.hg.1 | ENST00000529785.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010745.hg.1 | ENST00000529785.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010745.hg.1 | ENST00000529785.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010745.hg.1 | ENST00000534226.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010745.hg.1 | ENST00000534226.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010745.hg.1 | ENST00000534226.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010745.hg.1 | ENST00000534226.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010745.hg.1 | ENST00000534226.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010745.hg.1 | ENST00000534226.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010745.hg.1 | ENST00000534226.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010745.hg.1 | ENST00000534226.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010745.hg.1 | ENST00000534226.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010745.hg.1 | ENST00000534226.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200181904.hg.1 | ENST00000257951.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200181904.hg.1 | NM_033045.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200181904.hg.1 | ENST00000257951.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200181904.hg.1 | NM_033045.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200181904.hg.1 | TC1200010752.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1200010752.hg.1 | ENST00000257951.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010752.hg.1 | ENST00000257951.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010752.hg.1 | ENST00000257951.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010752.hg.1 | ENST00000257951.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010752.hg.1 | ENST00000257951.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010752.hg.1 | ENST00000257951.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010752.hg.1 | ENST00000257951.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010752.hg.1 | ENST00000257951.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010752.hg.1 | ENST00000257951.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010752.hg.1 | ENST00000257951.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010752.hg.1 | ENST00000257951.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010752.hg.1 | NM_033045.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010752.hg.1 | NM_033045.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010752.hg.1 | NM_033045.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010752.hg.1 | NM_033045.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010752.hg.1 | NM_033045.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010752.hg.1 | NM_033045.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010752.hg.1 | NM_033045.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010752.hg.1 | NM_033045.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010752.hg.1 | NM_033045.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010752.hg.1 | NM_033045.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010752.hg.1 | NM_033045.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200181906.hg.1 | ENST00000257951.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200181906.hg.1 | NM_033045.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200181906.hg.1 | ENST00000257951.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200181906.hg.1 | NM_033045.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200181906.hg.1 | TC1200010752.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1200181907.hg.1 | ENST00000257951.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200181907.hg.1 | NM_033045.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200181907.hg.1 | ENST00000257951.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200181907.hg.1 | NM_033045.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200181907.hg.1 | TC1200010752.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1200010745.hg.1 | ENST00000529785.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010745.hg.1 | ENST00000529785.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010745.hg.1 | ENST00000529785.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010745.hg.1 | ENST00000529785.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010745.hg.1 | ENST00000529785.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010745.hg.1 | ENST00000529785.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010745.hg.1 | ENST00000529785.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010745.hg.1 | ENST00000529785.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010745.hg.1 | ENST00000529785.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010745.hg.1 | ENST00000529785.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010745.hg.1 | ENST00000534226.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010745.hg.1 | ENST00000534226.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010745.hg.1 | ENST00000534226.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010745.hg.1 | ENST00000534226.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010745.hg.1 | ENST00000534226.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010745.hg.1 | ENST00000534226.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010745.hg.1 | ENST00000534226.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010745.hg.1 | ENST00000534226.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010745.hg.1 | ENST00000534226.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010745.hg.1 | ENST00000534226.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200181905.hg.1 | ENST00000257951.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200181905.hg.1 | NM_033045.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200181905.hg.1 | ENST00000257951.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200181905.hg.1 | NM_033045.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200181905.hg.1 | TC1200010752.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1200181908.hg.1 | ENST00000257951.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200181908.hg.1 | NM_033045.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200181908.hg.1 | ENST00000257951.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200181908.hg.1 | NM_033045.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200181908.hg.1 | TC1200010752.hg.1 | Clariom D TC |
