| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06027185.hg.1 |
TC06002157.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06027185.hg.1 |
ENST00000265603.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06027185.hg.1 |
ENST00000421351.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06027185.hg.1 |
NM_022121.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06027186.hg.1 |
TC06002157.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06027186.hg.1 |
ENST00000421351.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06027186.hg.1 |
NM_022121.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06027184.hg.1 |
TC06002157.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06027184.hg.1 |
ENST00000265603.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06027184.hg.1 |
ENST00000421351.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06027184.hg.1 |
NM_022121.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06027189.hg.1 |
TC06002157.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06027189.hg.1 |
ENST00000421351.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06027189.hg.1 |
NM_022121.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013326.hg.1 |
ENST00000421351.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013326.hg.1 |
ENST00000421351.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013326.hg.1 |
ENST00000421351.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013326.hg.1 |
ENST00000421351.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013326.hg.1 |
ENST00000421351.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013326.hg.1 |
ENST00000421351.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013326.hg.1 |
ENST00000421351.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013326.hg.1 |
NM_022121.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013326.hg.1 |
NM_022121.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013326.hg.1 |
NM_022121.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013326.hg.1 |
NM_022121.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013326.hg.1 |
NM_022121.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013326.hg.1 |
NM_022121.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013326.hg.1 |
NM_022121.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06027187.hg.1 |
TC06002157.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06027187.hg.1 |
ENST00000265603.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06027187.hg.1 |
ENST00000421351.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06027187.hg.1 |
NM_022121.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600192187.hg.1 |
ENST00000421351.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600192187.hg.1 |
NM_022121.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600192187.hg.1 |
ENST00000421351.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600192187.hg.1 |
NM_022121.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600192187.hg.1 |
TC0600013326.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600192186.hg.1 |
ENST00000421351.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600192186.hg.1 |
NM_022121.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600192186.hg.1 |
ENST00000421351.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600192186.hg.1 |
NM_022121.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600192186.hg.1 |
TC0600013326.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0600013326.hg.1 |
ENST00000421351.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013326.hg.1 |
ENST00000421351.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013326.hg.1 |
ENST00000421351.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013326.hg.1 |
ENST00000421351.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013326.hg.1 |
ENST00000421351.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013326.hg.1 |
ENST00000421351.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013326.hg.1 |
ENST00000421351.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013326.hg.1 |
NM_022121.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600013326.hg.1 |
NM_022121.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600013326.hg.1 |
NM_022121.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600013326.hg.1 |
NM_022121.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600013326.hg.1 |
NM_022121.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600013326.hg.1 |
NM_022121.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600013326.hg.1 |
NM_022121.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600192184.hg.1 |
ENST00000421351.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600192184.hg.1 |
NM_022121.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600192184.hg.1 |
ENST00000421351.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600192184.hg.1 |
NM_022121.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600192184.hg.1 |
TC0600013326.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600192185.hg.1 |
ENST00000421351.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600192185.hg.1 |
NM_022121.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600192185.hg.1 |
ENST00000421351.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600192185.hg.1 |
NM_022121.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600192185.hg.1 |
TC0600013326.hg.1 |
Clariom D TC |