Assay Details
Gene Symbol: MTMR8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017677.3 | NP_060147.2 | 2 - 3 | 239 | 61 | |
GenBank mRNA | BC012399.1 | - | 2 - 3 | 217 | 3864128 | 61 |
DA691850.1 | - | 2 - 3 | 239 | 61 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC0X002333.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR0X014930.hg.1 | TC0X002333.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR0X014930.hg.1 | ENST00000247400.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X014930.hg.1 | ENST00000374852.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X014930.hg.1 | NM_017677.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009882.hg.1 | ENST00000374852.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009882.hg.1 | ENST00000374852.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009882.hg.1 | ENST00000374852.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009882.hg.1 | ENST00000374852.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009882.hg.1 | ENST00000374852.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009882.hg.1 | ENST00000374852.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009882.hg.1 | ENST00000374852.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009882.hg.1 | ENST00000374852.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009882.hg.1 | ENST00000374852.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009882.hg.1 | ENST00000374852.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009882.hg.1 | ENST00000374852.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009882.hg.1 | ENST00000374852.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009882.hg.1 | ENST00000374852.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009882.hg.1 | ENST00000374852.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009882.hg.1 | ENST00000374852.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009882.hg.1 | ENST00000374852.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009882.hg.1 | ENST00000461403.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009882.hg.1 | ENST00000461403.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009882.hg.1 | ENST00000461403.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009882.hg.1 | ENST00000461403.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009882.hg.1 | ENST00000461403.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009882.hg.1 | ENST00000462447.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009882.hg.1 | ENST00000462447.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009882.hg.1 | ENST00000462447.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009882.hg.1 | ENST00000462447.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009882.hg.1 | ENST00000462447.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009882.hg.1 | ENST00000478487.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009882.hg.1 | ENST00000478487.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009882.hg.1 | ENST00000478487.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009882.hg.1 | ENST00000478487.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009882.hg.1 | ENST00000478487.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009882.hg.1 | NM_017677.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009882.hg.1 | NM_017677.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009882.hg.1 | NM_017677.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009882.hg.1 | NM_017677.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009882.hg.1 | NM_017677.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009882.hg.1 | NM_017677.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009882.hg.1 | NM_017677.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009882.hg.1 | NM_017677.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009882.hg.1 | NM_017677.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009882.hg.1 | NM_017677.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009882.hg.1 | NM_017677.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009882.hg.1 | NM_017677.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009882.hg.1 | NM_017677.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009882.hg.1 | NM_017677.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009882.hg.1 | NM_017677.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009882.hg.1 | NM_017677.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009882.hg.1 | ENST00000374852.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009882.hg.1 | ENST00000374852.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009882.hg.1 | ENST00000374852.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009882.hg.1 | ENST00000374852.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009882.hg.1 | ENST00000374852.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009882.hg.1 | ENST00000374852.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009882.hg.1 | ENST00000374852.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009882.hg.1 | ENST00000374852.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009882.hg.1 | ENST00000374852.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009882.hg.1 | ENST00000374852.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009882.hg.1 | ENST00000374852.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009882.hg.1 | ENST00000374852.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009882.hg.1 | ENST00000374852.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009882.hg.1 | ENST00000374852.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009882.hg.1 | ENST00000374852.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009882.hg.1 | ENST00000374852.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009882.hg.1 | ENST00000461403.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009882.hg.1 | ENST00000461403.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009882.hg.1 | ENST00000461403.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009882.hg.1 | ENST00000461403.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009882.hg.1 | ENST00000461403.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009882.hg.1 | ENST00000462447.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009882.hg.1 | ENST00000462447.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009882.hg.1 | ENST00000462447.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009882.hg.1 | ENST00000462447.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009882.hg.1 | ENST00000462447.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009882.hg.1 | ENST00000478487.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009882.hg.1 | ENST00000478487.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009882.hg.1 | ENST00000478487.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009882.hg.1 | ENST00000478487.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009882.hg.1 | ENST00000478487.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009882.hg.1 | NM_017677.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009882.hg.1 | NM_017677.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009882.hg.1 | NM_017677.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009882.hg.1 | NM_017677.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009882.hg.1 | NM_017677.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009882.hg.1 | NM_017677.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009882.hg.1 | NM_017677.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009882.hg.1 | NM_017677.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009882.hg.1 | NM_017677.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009882.hg.1 | NM_017677.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009882.hg.1 | NM_017677.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009882.hg.1 | NM_017677.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009882.hg.1 | NM_017677.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009882.hg.1 | NM_017677.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009882.hg.1 | NM_017677.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009882.hg.1 | NM_017677.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00167599.hg.1 | ENST00000374852.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00167599.hg.1 | NM_017677.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00167599.hg.1 | ENST00000374852.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00167599.hg.1 | NM_017677.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00167599.hg.1 | TC0X00009882.hg.1 | Clariom D TC |
