Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X014911.hg.1 |
TC0X002333.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X014911.hg.1 |
ENST00000247400.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X014911.hg.1 |
ENST00000374852.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X014911.hg.1 |
NM_017677.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009882.hg.1 |
ENST00000374852.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009882.hg.1 |
ENST00000374852.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009882.hg.1 |
ENST00000374852.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009882.hg.1 |
ENST00000374852.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009882.hg.1 |
ENST00000374852.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009882.hg.1 |
ENST00000374852.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009882.hg.1 |
ENST00000374852.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009882.hg.1 |
ENST00000374852.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009882.hg.1 |
ENST00000374852.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009882.hg.1 |
ENST00000374852.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009882.hg.1 |
ENST00000374852.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009882.hg.1 |
ENST00000374852.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009882.hg.1 |
ENST00000374852.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009882.hg.1 |
ENST00000374852.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009882.hg.1 |
ENST00000374852.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009882.hg.1 |
ENST00000374852.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009882.hg.1 |
ENST00000461403.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009882.hg.1 |
ENST00000461403.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009882.hg.1 |
ENST00000461403.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009882.hg.1 |
ENST00000461403.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009882.hg.1 |
ENST00000461403.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009882.hg.1 |
ENST00000462447.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009882.hg.1 |
ENST00000462447.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009882.hg.1 |
ENST00000462447.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009882.hg.1 |
ENST00000462447.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009882.hg.1 |
ENST00000462447.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009882.hg.1 |
ENST00000478487.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009882.hg.1 |
ENST00000478487.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009882.hg.1 |
ENST00000478487.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009882.hg.1 |
ENST00000478487.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009882.hg.1 |
ENST00000478487.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009882.hg.1 |
NM_017677.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009882.hg.1 |
NM_017677.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009882.hg.1 |
NM_017677.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009882.hg.1 |
NM_017677.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009882.hg.1 |
NM_017677.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009882.hg.1 |
NM_017677.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009882.hg.1 |
NM_017677.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009882.hg.1 |
NM_017677.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009882.hg.1 |
NM_017677.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009882.hg.1 |
NM_017677.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009882.hg.1 |
NM_017677.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009882.hg.1 |
NM_017677.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009882.hg.1 |
NM_017677.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009882.hg.1 |
NM_017677.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009882.hg.1 |
NM_017677.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009882.hg.1 |
NM_017677.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00167585.hg.1 |
ENST00000374852.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00167585.hg.1 |
NM_017677.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00167585.hg.1 |
ENST00000374852.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00167585.hg.1 |
NM_017677.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00167585.hg.1 |
TC0X00009882.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00009882.hg.1 |
ENST00000374852.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009882.hg.1 |
ENST00000374852.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009882.hg.1 |
ENST00000374852.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009882.hg.1 |
ENST00000374852.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009882.hg.1 |
ENST00000374852.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009882.hg.1 |
ENST00000374852.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009882.hg.1 |
ENST00000374852.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009882.hg.1 |
ENST00000374852.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009882.hg.1 |
ENST00000374852.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009882.hg.1 |
ENST00000374852.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009882.hg.1 |
ENST00000374852.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009882.hg.1 |
ENST00000374852.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009882.hg.1 |
ENST00000374852.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009882.hg.1 |
ENST00000374852.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009882.hg.1 |
ENST00000374852.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009882.hg.1 |
ENST00000374852.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009882.hg.1 |
ENST00000461403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009882.hg.1 |
ENST00000461403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009882.hg.1 |
ENST00000461403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009882.hg.1 |
ENST00000461403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009882.hg.1 |
ENST00000461403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009882.hg.1 |
ENST00000462447.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009882.hg.1 |
ENST00000462447.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009882.hg.1 |
ENST00000462447.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009882.hg.1 |
ENST00000462447.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009882.hg.1 |
ENST00000462447.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009882.hg.1 |
ENST00000478487.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009882.hg.1 |
ENST00000478487.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009882.hg.1 |
ENST00000478487.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009882.hg.1 |
ENST00000478487.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009882.hg.1 |
ENST00000478487.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009882.hg.1 |
NM_017677.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009882.hg.1 |
NM_017677.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009882.hg.1 |
NM_017677.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009882.hg.1 |
NM_017677.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009882.hg.1 |
NM_017677.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009882.hg.1 |
NM_017677.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009882.hg.1 |
NM_017677.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009882.hg.1 |
NM_017677.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009882.hg.1 |
NM_017677.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009882.hg.1 |
NM_017677.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009882.hg.1 |
NM_017677.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009882.hg.1 |
NM_017677.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009882.hg.1 |
NM_017677.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009882.hg.1 |
NM_017677.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009882.hg.1 |
NM_017677.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009882.hg.1 |
NM_017677.3 |
Refseq |