Assay Details
| Gene Symbol: LDB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080115.1 | NP_001073584.1 | 4 - 5 | 715 | 95 | |
| NM_007078.2 | NP_009009.1 | 4 - 5 | 715 | 95 | ||
| XM_005269464.4 | XP_005269521.1 | 5 - 6 | 884 | 95 | ||
| XM_005269466.4 | XP_005269523.1 | 5 - 6 | 883 | 95 | ||
| XM_005269468.4 | XP_005269525.1 | 5 - 6 | 880 | 95 | ||
| XM_011539188.2 | XP_011537490.1 | 5 - 6 | 885 | 95 | ||
| XM_017015606.1 | XP_016871095.1 | 5 - 6 | 2779 | 95 | ||
| XM_017015608.1 | XP_016871097.1 | 5 - 6 | 2779 | 95 | ||
| GenBank mRNA | AB014513.1 | - | 4 - 5 | 713 | 95 | |
| AF276807.1 | - | 4 - 5 | 692 | 95 | ||
| AK294696.1 | - | 5 - 6 | 772 | 95 | ||
| AK297871.1 | - | 5 - 6 | 671 | 95 | ||
| KF772970.1 | - | 4 - 5 | 692 | 95 | ||
| KU178544.1 | - | 4 - 5 | 692 | 95 | ||
More Information
Set Membership: |
|

LDB3
Genomic Map