Assay Details
Gene Symbol: NOC4L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024078.2 | NP_076983.1 | 13 - 14 | 83 | ||
XM_017019960.1 | XP_016875449.1 | 12 - 13 | 83 | |||
GenBank mRNA | AW731886.1 | - | 2 - 3 | 326 | 83 | |
BC001191.1 | - | 13 - 14 | 3354945 | 83 | ||
BC007893.2 | - | 13 - 14 | 83 | |||
BC071958.1 | - | 13 - 14 | 6597539 | 83 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC12001054.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR12013959.hg.1 | ENST00000330579.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12013959.hg.1 | ENST00000538784.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12013959.hg.1 | NM_024078.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12013959.hg.1 | TC12001054.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000330579.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000330579.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000330579.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000330579.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000330579.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000330579.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000330579.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000330579.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000330579.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000330579.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000330579.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000330579.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000330579.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000330579.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000330579.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000330579.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000330579.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000330579.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000535343.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000535343.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000535343.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000535343.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000535343.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000535343.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000535343.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000538784.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000538784.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000538784.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000538784.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000538784.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000538784.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000538784.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000538784.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000538784.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000538784.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000541954.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000541954.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000541954.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000541954.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000541954.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000541954.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000541954.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000541954.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000541954.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000541954.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | NM_024078.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | NM_024078.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | NM_024078.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | NM_024078.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | NM_024078.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | NM_024078.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | NM_024078.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | NM_024078.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | NM_024078.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | NM_024078.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | NM_024078.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | NM_024078.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | NM_024078.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | NM_024078.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | NM_024078.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | NM_024078.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | NM_024078.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | NM_024078.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | NM_024078.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000330579.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000330579.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000330579.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000330579.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000330579.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000330579.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000330579.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000330579.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000330579.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000330579.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000330579.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000330579.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000330579.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000330579.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000330579.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000330579.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000330579.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000330579.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000535343.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000535343.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000535343.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000535343.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000535343.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000535343.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000535343.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000538784.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000538784.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000538784.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000538784.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000538784.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000538784.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000538784.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000538784.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000538784.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000538784.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000541954.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000541954.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000541954.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000541954.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000541954.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000541954.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000541954.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000541954.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000541954.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000541954.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | NM_024078.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | NM_024078.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | NM_024078.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | NM_024078.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | NM_024078.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | NM_024078.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | NM_024078.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | NM_024078.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | NM_024078.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | NM_024078.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | NM_024078.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | NM_024078.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | NM_024078.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | NM_024078.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | NM_024078.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | NM_024078.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | NM_024078.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | NM_024078.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | NM_024078.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200170659.hg.1 | ENST00000330579.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200170659.hg.1 | ENST00000538784.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200170659.hg.1 | NM_024078.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200170659.hg.1 | ENST00000330579.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200170659.hg.1 | ENST00000538784.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200170659.hg.1 | NM_024078.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200170659.hg.1 | TC1200009467.hg.1 | Clariom D TC |
