Assay Details
| Gene Symbol: NOC4L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024078.2 | NP_076983.1 | 10 - 11 | 1024 | 73 | |
| XM_017019960.1 | XP_016875449.1 | 9 - 10 | 978 | 73 | ||
| GenBank mRNA | BC001191.1 | - | 10 - 11 | 1003 | 3354945 | 73 |
| BC007893.2 | - | 10 - 11 | 896 | 73 | ||
| BC071958.1 | - | 10 - 11 | 967 | 6597539 | 73 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12001054.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR12013952.hg.1 | ENST00000330579.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12013952.hg.1 | ENST00000535343.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12013952.hg.1 | NM_024078.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12013952.hg.1 | TC12001054.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12013954.hg.1 | ENST00000330579.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12013954.hg.1 | ENST00000538784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12013954.hg.1 | NM_024078.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12013954.hg.1 | TC12001054.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12013955.hg.1 | ENST00000330579.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12013955.hg.1 | ENST00000535343.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12013955.hg.1 | ENST00000538784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12013955.hg.1 | NM_024078.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12013955.hg.1 | TC12001054.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000330579.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000330579.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000330579.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000330579.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000330579.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000330579.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000330579.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000330579.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000330579.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000330579.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000330579.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000330579.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000330579.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000330579.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000330579.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000330579.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000330579.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000330579.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000535343.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000535343.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000535343.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000535343.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000535343.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000535343.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000535343.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000538784.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000538784.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000538784.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000538784.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000538784.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000538784.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000538784.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000538784.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000538784.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000538784.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000541954.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000541954.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000541954.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000541954.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000541954.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000541954.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000541954.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000541954.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000541954.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | ENST00000541954.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | NM_024078.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | NM_024078.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | NM_024078.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | NM_024078.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | NM_024078.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | NM_024078.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | NM_024078.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | NM_024078.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | NM_024078.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | NM_024078.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | NM_024078.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | NM_024078.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | NM_024078.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | NM_024078.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | NM_024078.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | NM_024078.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | NM_024078.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | NM_024078.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009467.hg.1 | NM_024078.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12013950.hg.1 | ENST00000330579.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12013950.hg.1 | ENST00000535343.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12013950.hg.1 | NM_024078.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12013950.hg.1 | TC12001054.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12013951.hg.1 | ENST00000330579.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12013951.hg.1 | ENST00000535343.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12013951.hg.1 | NM_024078.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12013951.hg.1 | TC12001054.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12013956.hg.1 | ENST00000330579.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12013956.hg.1 | ENST00000535343.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12013956.hg.1 | ENST00000538784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12013956.hg.1 | NM_024078.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12013956.hg.1 | TC12001054.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1200170651.hg.1 | ENST00000330579.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200170651.hg.1 | ENST00000535343.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200170651.hg.1 | ENST00000541954.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200170651.hg.1 | NM_024078.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200170651.hg.1 | ENST00000330579.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200170651.hg.1 | ENST00000535343.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200170651.hg.1 | ENST00000541954.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200170651.hg.1 | NM_024078.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200170651.hg.1 | TC1200009467.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200170655.hg.1 | ENST00000330579.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200170655.hg.1 | ENST00000535343.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200170655.hg.1 | ENST00000538784.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200170655.hg.1 | NM_024078.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200170655.hg.1 | ENST00000330579.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200170655.hg.1 | ENST00000535343.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200170655.hg.1 | ENST00000538784.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200170655.hg.1 | NM_024078.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200170655.hg.1 | TC1200009467.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200170650.hg.1 | ENST00000330579.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200170650.hg.1 | ENST00000535343.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200170650.hg.1 | ENST00000541954.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200170650.hg.1 | NM_024078.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200170650.hg.1 | ENST00000330579.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200170650.hg.1 | ENST00000535343.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200170650.hg.1 | ENST00000541954.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200170650.hg.1 | NM_024078.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200170650.hg.1 | TC1200009467.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200170649.hg.1 | ENST00000330579.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200170649.hg.1 | ENST00000535343.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200170649.hg.1 | ENST00000541954.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200170649.hg.1 | NM_024078.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200170649.hg.1 | ENST00000330579.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200170649.hg.1 | ENST00000535343.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200170649.hg.1 | ENST00000541954.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200170649.hg.1 | NM_024078.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200170649.hg.1 | TC1200009467.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200170654.hg.1 | ENST00000330579.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200170654.hg.1 | ENST00000538784.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200170654.hg.1 | NM_024078.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200170654.hg.1 | ENST00000330579.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200170654.hg.1 | ENST00000538784.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200170654.hg.1 | NM_024078.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200170654.hg.1 | TC1200009467.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200170656.hg.1 | ENST00000330579.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200170656.hg.1 | ENST00000535343.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200170656.hg.1 | ENST00000538784.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200170656.hg.1 | NM_024078.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200170656.hg.1 | ENST00000330579.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200170656.hg.1 | ENST00000535343.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200170656.hg.1 | ENST00000538784.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200170656.hg.1 | NM_024078.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200170656.hg.1 | TC1200009467.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000330579.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000330579.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000330579.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000330579.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000330579.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000330579.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000330579.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000330579.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000330579.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000330579.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000330579.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000330579.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000330579.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000330579.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000330579.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000330579.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000330579.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000330579.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000535343.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000535343.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000535343.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000535343.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000535343.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000535343.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000535343.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000538784.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000538784.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000538784.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000538784.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000538784.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000538784.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000538784.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000538784.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000538784.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000538784.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000541954.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000541954.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000541954.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000541954.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000541954.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000541954.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000541954.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000541954.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000541954.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | ENST00000541954.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | NM_024078.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | NM_024078.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | NM_024078.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | NM_024078.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | NM_024078.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | NM_024078.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | NM_024078.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | NM_024078.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | NM_024078.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | NM_024078.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | NM_024078.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | NM_024078.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | NM_024078.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | NM_024078.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | NM_024078.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | NM_024078.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | NM_024078.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | NM_024078.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009467.hg.1 | NM_024078.2 | Refseq |

NOC4L
Genomic Map