| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13009187.hg.1 |
ENST00000376503.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13009187.hg.1 |
NM_005073.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13009187.hg.1 |
TC13000824.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1300164043.hg.1 |
ENST00000376503.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300164043.hg.1 |
ENST00000618992.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300164043.hg.1 |
NM_005073.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300164043.hg.1 |
ENST00000376503.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300164043.hg.1 |
ENST00000618992.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300164043.hg.1 |
NM_005073.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300164043.hg.1 |
TC1300009580.hg.1 |
Clariom D TC |