Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008706.hg.1 |
TC0X000659.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008706.hg.1 |
ENST00000287534.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008706.hg.1 |
ENST00000370652.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008706.hg.1 |
ENST00000394141.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008706.hg.1 |
ENST00000394143.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008706.hg.1 |
ENST00000412101.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008706.hg.1 |
NM_153834.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00158055.hg.1 |
ENST00000370652.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00158055.hg.1 |
ENST00000394141.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00158055.hg.1 |
ENST00000394143.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00158055.hg.1 |
NM_153834.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00158055.hg.1 |
ENST00000370652.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00158055.hg.1 |
ENST00000394141.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00158055.hg.1 |
ENST00000394143.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00158055.hg.1 |
NM_153834.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00158055.hg.1 |
TC0X00008528.hg.1 |
Clariom D TC |